Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74150021-74150551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLA3-AS1
Entrez Gene ID
100507171
Gene Name
BOLA3 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.503348
Gene Chromosome Location
Chr.2: 74147981-74152389 on Build GRCh38
Transcripts
NR_045634.1 NR_045636.1

SNP Detail

SNP ID Chromosome Location
rs985098607 Chr.2: 74150343-74150343 on Build GRCh38
rs912943079 Chr.2: 74150403-74150403 on Build GRCh38
rs780618636 Chr.2: 74150473-74150473 on Build GRCh38
rs1054655182 Chr.2: 74150217-74150217 on Build GRCh38
rs1024303607 Chr.2: 74150267-74150267 on Build GRCh38
rs1143500 Chr.2: 74150275-74150275 on Build GRCh38
rs1143499 Chr.2: 74150266-74150266 on Build GRCh38
rs756451029 Chr.2: 74150045-74150046 on Build GRCh38
rs1028816750 Chr.2: 74150075-74150075 on Build GRCh38
rs539458417 Chr.2: 74150410-74150410 on Build GRCh38
rs949682972 Chr.2: 74150502-74150502 on Build GRCh38
rs757391159 Chr.2: 74150159-74150159 on Build GRCh38
rs975833495 Chr.2: 74150444-74150444 on Build GRCh38
rs899236304 Chr.2: 74150132-74150132 on Build GRCh38
rs185138888 Chr.2: 74150429-74150429 on Build GRCh38
rs113504338 Chr.2: 74150062-74150062 on Build GRCh38
rs1042369623 Chr.2: 74150145-74150145 on Build GRCh38
rs7596510 Chr.2: 74150293-74150293 on Build GRCh38
rs1001373486 Chr.2: 74150090-74150090 on Build GRCh38
rs749637810 Chr.2: 74150501-74150501 on Build GRCh38
rs528779128 Chr.2: 74150146-74150146 on Build GRCh38
rs140260390 Chr.2: 74150249-74150249 on Build GRCh38
rs926140380 Chr.2: 74150264-74150264 on Build GRCh38
rs944985822 Chr.2: 74150121-74150121 on Build GRCh38
rs1012970766 Chr.2: 74150257-74150257 on Build GRCh38
rs568963750 Chr.2: 74150327-74150327 on Build GRCh38
rs1143504 Chr.2: 74150427-74150427 on Build GRCh38
rs945100104 Chr.2: 74150474-74150474 on Build GRCh38
rs776090631 Chr.2: 74150068-74150068 on Build GRCh38
rs189599547 Chr.2: 74150517-74150517 on Build GRCh38
rs869219379 Chr.2: 74150061-74150061 on Build GRCh38
rs994898659 Chr.2: 74150138-74150139 on Build GRCh38
rs1143498 Chr.2: 74150245-74150245 on Build GRCh38
rs936251163 Chr.2: 74150167-74150167 on Build GRCh38
rs869158361 Chr.2: 74150058-74150061 on Build GRCh38
rs7584195 Chr.2: 74150306-74150306 on Build GRCh38
rs564824968 Chr.2: 74150102-74150102 on Build GRCh38
rs965365451 Chr.2: 74150231-74150231 on Build GRCh38
rs1040119101 Chr.2: 74150069-74150069 on Build GRCh38
rs912993360 Chr.2: 74150414-74150414 on Build GRCh38
rs370837912 Chr.2: 74150415-74150415 on Build GRCh38
rs1045847455 Chr.2: 74150505-74150505 on Build GRCh38
rs56768895 Chr.2: 74150060-74150061 on Build GRCh38
rs1009314180 Chr.2: 74150186-74150187 on Build GRCh38
rs562620300 Chr.2: 74150443-74150443 on Build GRCh38
rs1026412310 Chr.2: 74150152-74150154 on Build GRCh38
rs142525433 Chr.2: 74150326-74150326 on Build GRCh38
rs376904044 Chr.2: 74150374-74150374 on Build GRCh38
rs907217868 Chr.2: 74150269-74150269 on Build GRCh38

View Full Product Details