Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74150165-74150703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLA3-AS1
Entrez Gene ID
100507171
Gene Name
BOLA3 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.503348
Gene Chromosome Location
Chr.2: 74147981-74152389 on Build GRCh38
Transcripts
NR_045634.1 NR_045636.1

SNP Detail

SNP ID Chromosome Location
rs985098607 Chr.2: 74150343-74150343 on Build GRCh38
rs912943079 Chr.2: 74150403-74150403 on Build GRCh38
rs189599547 Chr.2: 74150517-74150517 on Build GRCh38
rs898301951 Chr.2: 74150590-74150590 on Build GRCh38
rs1143498 Chr.2: 74150245-74150245 on Build GRCh38
rs769274648 Chr.2: 74150611-74150611 on Build GRCh38
rs780618636 Chr.2: 74150473-74150473 on Build GRCh38
rs1024303607 Chr.2: 74150267-74150267 on Build GRCh38
rs535666675 Chr.2: 74150583-74150584 on Build GRCh38
rs1054655182 Chr.2: 74150217-74150217 on Build GRCh38
rs994009226 Chr.2: 74150614-74150614 on Build GRCh38
rs1143500 Chr.2: 74150275-74150275 on Build GRCh38
rs552929301 Chr.2: 74150600-74150600 on Build GRCh38
rs1055090219 Chr.2: 74150592-74150592 on Build GRCh38
rs7584195 Chr.2: 74150306-74150306 on Build GRCh38
rs1143499 Chr.2: 74150266-74150266 on Build GRCh38
rs965365451 Chr.2: 74150231-74150231 on Build GRCh38
rs539458417 Chr.2: 74150410-74150410 on Build GRCh38
rs949682972 Chr.2: 74150502-74150502 on Build GRCh38
rs912993360 Chr.2: 74150414-74150414 on Build GRCh38
rs370837912 Chr.2: 74150415-74150415 on Build GRCh38
rs936296476 Chr.2: 74150585-74150585 on Build GRCh38
rs975833495 Chr.2: 74150444-74150444 on Build GRCh38
rs185138888 Chr.2: 74150429-74150429 on Build GRCh38
rs7596510 Chr.2: 74150293-74150293 on Build GRCh38
rs1045847455 Chr.2: 74150505-74150505 on Build GRCh38
rs749637810 Chr.2: 74150501-74150501 on Build GRCh38
rs140260390 Chr.2: 74150249-74150249 on Build GRCh38
rs562620300 Chr.2: 74150443-74150443 on Build GRCh38
rs926140380 Chr.2: 74150264-74150264 on Build GRCh38
rs534897629 Chr.2: 74150561-74150561 on Build GRCh38
rs1012970766 Chr.2: 74150257-74150257 on Build GRCh38
rs927226648 Chr.2: 74150554-74150554 on Build GRCh38
rs894647634 Chr.2: 74150681-74150681 on Build GRCh38
rs568963750 Chr.2: 74150327-74150327 on Build GRCh38
rs142525433 Chr.2: 74150326-74150326 on Build GRCh38
rs376904044 Chr.2: 74150374-74150374 on Build GRCh38
rs907217868 Chr.2: 74150269-74150269 on Build GRCh38
rs1143504 Chr.2: 74150427-74150427 on Build GRCh38
rs945100104 Chr.2: 74150474-74150474 on Build GRCh38

View Full Product Details