Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58841825-58842329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_001309861.1 NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_001309861.1 NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs529624732 Chr.20: 58842138-58842138 on Build GRCh38
rs756247425 Chr.20: 58842225-58842225 on Build GRCh38
rs187566315 Chr.20: 58841894-58841894 on Build GRCh38
rs907096149 Chr.20: 58842080-58842080 on Build GRCh38
rs780343508 Chr.20: 58842274-58842274 on Build GRCh38
rs557127211 Chr.20: 58841954-58841954 on Build GRCh38
rs535218689 Chr.20: 58842264-58842264 on Build GRCh38
rs1010987431 Chr.20: 58842127-58842127 on Build GRCh38
rs540677999 Chr.20: 58842104-58842104 on Build GRCh38
rs1001880476 Chr.20: 58842081-58842081 on Build GRCh38
rs564459918 Chr.20: 58842039-58842039 on Build GRCh38
rs946356852 Chr.20: 58842273-58842276 on Build GRCh38
rs562359997 Chr.20: 58842130-58842130 on Build GRCh38
rs975435326 Chr.20: 58842278-58842278 on Build GRCh38
rs6100248 Chr.20: 58842031-58842031 on Build GRCh38
rs1049324871 Chr.20: 58841996-58841996 on Build GRCh38
rs191063552 Chr.20: 58842287-58842287 on Build GRCh38
rs1034114516 Chr.20: 58841867-58841867 on Build GRCh38
rs921623721 Chr.20: 58841990-58841990 on Build GRCh38
rs566522944 Chr.20: 58842033-58842033 on Build GRCh38
rs912118996 Chr.20: 58841972-58841972 on Build GRCh38
rs781147491 Chr.20: 58841936-58841936 on Build GRCh38
rs886192730 Chr.20: 58842007-58842007 on Build GRCh38
rs758455249 Chr.20: 58841901-58841901 on Build GRCh38
rs550848950 Chr.20: 58842235-58842235 on Build GRCh38
rs920449654 Chr.20: 58841926-58841926 on Build GRCh38
rs747997073 Chr.20: 58841957-58841957 on Build GRCh38
rs1045601795 Chr.20: 58842049-58842049 on Build GRCh38
rs906453208 Chr.20: 58841959-58841959 on Build GRCh38
rs569455269 Chr.20: 58842250-58842250 on Build GRCh38
rs999732115 Chr.20: 58842278-58842280 on Build GRCh38
rs944979999 Chr.20: 58841978-58841978 on Build GRCh38
rs955585178 Chr.20: 58841854-58841854 on Build GRCh38
rs1033670589 Chr.20: 58842098-58842098 on Build GRCh38
rs966780490 Chr.20: 58842165-58842165 on Build GRCh38
rs950587363 Chr.20: 58842223-58842223 on Build GRCh38
rs902254092 Chr.20: 58842278-58842278 on Build GRCh38
rs546295853 Chr.20: 58842038-58842038 on Build GRCh38
rs577524284 Chr.20: 58842175-58842176 on Build GRCh38
rs575623277 Chr.20: 58841970-58841970 on Build GRCh38
rs769654457 Chr.20: 58842117-58842117 on Build GRCh38
rs963011989 Chr.20: 58842093-58842093 on Build GRCh38
rs955646200 Chr.20: 58842062-58842063 on Build GRCh38
rs376640570 Chr.20: 58842220-58842220 on Build GRCh38
rs200012486 Chr.20: 58842287-58842288 on Build GRCh38
rs777375142 Chr.20: 58842233-58842233 on Build GRCh38
rs983246541 Chr.20: 58842225-58842225 on Build GRCh38
rs1043472034 Chr.20: 58842036-58842036 on Build GRCh38
rs999681353 Chr.20: 58842045-58842045 on Build GRCh38
rs528475197 Chr.20: 58842046-58842046 on Build GRCh38
rs966888117 Chr.20: 58841918-58841918 on Build GRCh38
rs930403342 Chr.20: 58841991-58841991 on Build GRCh38
rs989998669 Chr.20: 58841891-58841891 on Build GRCh38
rs541613069 Chr.20: 58841885-58841885 on Build GRCh38
rs939055726 Chr.20: 58841963-58841963 on Build GRCh38
rs1038680827 Chr.20: 58841983-58841983 on Build GRCh38
rs6026552 Chr.20: 58841915-58841915 on Build GRCh38
rs941455444 Chr.20: 58841911-58841911 on Build GRCh38

View Full Product Details