Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17441576-17442077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01549
Entrez Gene ID
100505929
Gene Name
long intergenic non-protein coding RNA 1549
Gene Aliases
C21orf37
UniGene
Hs.46707
Gene Chromosome Location
Chr.21: 17438890-17449185 on Build GRCh38
Transcripts
NR_037585.1 NR_037586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841649 c.4G>T p.E2* Chr.21: 17441784-17441784 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1053017069 Chr.21: 17441605-17441605 on Build GRCh38
rs181368392 Chr.21: 17441877-17441877 on Build GRCh38
rs869186998 Chr.21: 17441932-17441933 on Build GRCh38
rs570920467 Chr.21: 17441980-17441980 on Build GRCh38
rs79753277 Chr.21: 17441941-17441941 on Build GRCh38
rs997394240 Chr.21: 17441843-17441843 on Build GRCh38
rs572498804 Chr.21: 17441897-17441897 on Build GRCh38
rs550469645 Chr.21: 17441749-17441749 on Build GRCh38
rs1035384975 Chr.21: 17442010-17442010 on Build GRCh38
rs949558923 Chr.21: 17441955-17441955 on Build GRCh38
rs1014734822 Chr.21: 17441728-17441728 on Build GRCh38
rs879249340 Chr.21: 17441992-17441992 on Build GRCh38
rs570235669 Chr.21: 17441828-17441828 on Build GRCh38
rs914038894 Chr.21: 17441953-17441953 on Build GRCh38
rs906839641 Chr.21: 17441629-17441629 on Build GRCh38
rs926854486 Chr.21: 17442003-17442003 on Build GRCh38
rs887504838 Chr.21: 17441700-17441700 on Build GRCh38
rs768071216 Chr.21: 17441708-17441708 on Build GRCh38
rs1055914307 Chr.21: 17442047-17442055 on Build GRCh38
rs986526058 Chr.21: 17441890-17441890 on Build GRCh38
rs190965846 Chr.21: 17441838-17441838 on Build GRCh38
rs1030249265 Chr.21: 17441867-17441867 on Build GRCh38
rs74362024 Chr.21: 17441769-17441769 on Build GRCh38
rs769149252 Chr.21: 17441795-17441795 on Build GRCh38
rs208893 Chr.21: 17441925-17441925 on Build GRCh38
rs772224638 Chr.21: 17441706-17441706 on Build GRCh38
rs747599496 Chr.21: 17441796-17441796 on Build GRCh38
rs983149442 Chr.21: 17441687-17441687 on Build GRCh38
rs769011234 Chr.21: 17441879-17441879 on Build GRCh38
rs758970750 Chr.21: 17441703-17441703 on Build GRCh38
rs770077589 Chr.21: 17442012-17442012 on Build GRCh38
rs1001599046 Chr.21: 17441643-17441643 on Build GRCh38
rs199750688 Chr.21: 17441737-17441737 on Build GRCh38
rs932808836 Chr.21: 17441855-17441855 on Build GRCh38
rs946675897 Chr.21: 17441887-17441887 on Build GRCh38
rs555250587 Chr.21: 17441929-17441929 on Build GRCh38
rs951679703 Chr.21: 17441641-17441641 on Build GRCh38
rs1003454536 Chr.21: 17442004-17442004 on Build GRCh38
rs1055493507 Chr.21: 17441684-17441684 on Build GRCh38
rs397942000 Chr.21: 17441940-17441941 on Build GRCh38
rs760032487 Chr.21: 17441606-17441606 on Build GRCh38
rs1046588299 Chr.21: 17441956-17441956 on Build GRCh38
rs772944740 Chr.21: 17441885-17441885 on Build GRCh38
rs902541421 Chr.21: 17441972-17441972 on Build GRCh38
rs925332831 Chr.21: 17441908-17441908 on Build GRCh38
rs777059732 Chr.21: 17441801-17441801 on Build GRCh38
rs5842640 Chr.21: 17441928-17441929 on Build GRCh38
rs537259944 Chr.21: 17441654-17441654 on Build GRCh38
rs777039559 Chr.21: 17441880-17441880 on Build GRCh38
rs779948617 Chr.21: 17441750-17441750 on Build GRCh38
rs772750254 Chr.21: 17441603-17441603 on Build GRCh38
rs574996421 Chr.21: 17441990-17441990 on Build GRCh38
rs544015518 Chr.21: 17442048-17442048 on Build GRCh38
rs977444505 Chr.21: 17441823-17441823 on Build GRCh38
rs1046639756 Chr.21: 17441611-17441611 on Build GRCh38
rs116732116 Chr.21: 17441610-17441610 on Build GRCh38

View Full Product Details