Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32728753-32729258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NR_038879.1 NR_038880.1

SNP Detail

SNP ID Chromosome Location
rs923275358 Chr.21: 32729081-32729081 on Build GRCh38
rs778185176 Chr.21: 32728966-32728966 on Build GRCh38
rs540269136 Chr.21: 32728965-32728965 on Build GRCh38
rs754555084 Chr.21: 32728795-32728795 on Build GRCh38
rs887130028 Chr.21: 32728936-32728936 on Build GRCh38
rs756671987 Chr.21: 32729035-32729035 on Build GRCh38
rs753171502 Chr.21: 32728909-32728909 on Build GRCh38
rs967397423 Chr.21: 32729063-32729063 on Build GRCh38
rs957124157 Chr.21: 32729068-32729068 on Build GRCh38
rs995118858 Chr.21: 32728999-32728999 on Build GRCh38
rs1001805359 Chr.21: 32729027-32729027 on Build GRCh38
rs931905452 Chr.21: 32728825-32728825 on Build GRCh38
rs373733226 Chr.21: 32729191-32729191 on Build GRCh38
rs144277561 Chr.21: 32728817-32728817 on Build GRCh38
rs1050174368 Chr.21: 32728842-32728842 on Build GRCh38
rs150377045 Chr.21: 32729061-32729061 on Build GRCh38
rs745445350 Chr.21: 32729072-32729072 on Build GRCh38
rs547170569 Chr.21: 32729214-32729215 on Build GRCh38
rs747581097 Chr.21: 32728990-32728990 on Build GRCh38
rs753953211 Chr.21: 32728777-32728777 on Build GRCh38
rs180682822 Chr.21: 32729037-32729037 on Build GRCh38
rs779415504 Chr.21: 32728810-32728810 on Build GRCh38
rs990231588 Chr.21: 32729060-32729060 on Build GRCh38
rs907415164 Chr.21: 32729001-32729001 on Build GRCh38
rs147775739 Chr.21: 32728958-32728958 on Build GRCh38
rs909030872 Chr.21: 32729200-32729200 on Build GRCh38
rs940059517 Chr.21: 32729215-32729215 on Build GRCh38
rs941759025 Chr.21: 32728929-32728929 on Build GRCh38
rs986022532 Chr.21: 32729109-32729109 on Build GRCh38
rs545507016 Chr.21: 32728837-32728837 on Build GRCh38
rs953270945 Chr.21: 32729105-32729105 on Build GRCh38
rs976021168 Chr.21: 32729074-32729074 on Build GRCh38
rs967044035 Chr.21: 32729052-32729052 on Build GRCh38
rs894270247 Chr.21: 32729009-32729009 on Build GRCh38
rs987018711 Chr.21: 32729162-32729162 on Build GRCh38
rs976013808 Chr.21: 32729102-32729102 on Build GRCh38
rs140051776 Chr.21: 32728960-32728960 on Build GRCh38
rs1036546785 Chr.21: 32728957-32728957 on Build GRCh38
rs780801491 Chr.21: 32728965-32728965 on Build GRCh38
rs140379018 Chr.21: 32728793-32728793 on Build GRCh38
rs887624093 Chr.21: 32728884-32728884 on Build GRCh38
rs934131204 Chr.21: 32729160-32729160 on Build GRCh38
rs550944997 Chr.21: 32729231-32729231 on Build GRCh38

View Full Product Details