Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32742690-32743224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_013329.3 NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_013329.3 NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

SNP Detail

SNP ID Chromosome Location
rs930052733 Chr.21: 32742766-32742766 on Build GRCh38
rs1055151152 Chr.21: 32742966-32742966 on Build GRCh38
rs867432568 Chr.21: 32742799-32742799 on Build GRCh38
rs915540401 Chr.21: 32742897-32742897 on Build GRCh38
rs893038165 Chr.21: 32742909-32742909 on Build GRCh38
rs564722728 Chr.21: 32742875-32742875 on Build GRCh38
rs568618381 Chr.21: 32743020-32743020 on Build GRCh38
rs539295784 Chr.21: 32743143-32743143 on Build GRCh38
rs200134558 Chr.21: 32742849-32742849 on Build GRCh38
rs1027866070 Chr.21: 32743192-32743192 on Build GRCh38
rs779828531 Chr.21: 32743043-32743043 on Build GRCh38
rs941622585 Chr.21: 32742719-32742719 on Build GRCh38
rs749176452 Chr.21: 32743135-32743135 on Build GRCh38
rs1045445449 Chr.21: 32742910-32742911 on Build GRCh38
rs962974703 Chr.21: 32743181-32743181 on Build GRCh38
rs934828657 Chr.21: 32742951-32742951 on Build GRCh38
rs971882061 Chr.21: 32743175-32743175 on Build GRCh38
rs1056210271 Chr.21: 32742906-32742906 on Build GRCh38
rs759380117 Chr.21: 32743103-32743103 on Build GRCh38
rs756868916 Chr.21: 32742728-32742728 on Build GRCh38
rs971996568 Chr.21: 32742785-32742787 on Build GRCh38
rs991177188 Chr.21: 32742894-32742894 on Build GRCh38
rs1011929478 Chr.21: 32742919-32742919 on Build GRCh38
rs780779306 Chr.21: 32742735-32742735 on Build GRCh38
rs1045758634 Chr.21: 32742829-32742829 on Build GRCh38
rs974763751 Chr.21: 32743198-32743198 on Build GRCh38
rs755963334 Chr.21: 32743076-32743076 on Build GRCh38
rs777929330 Chr.21: 32743197-32743197 on Build GRCh38
rs745538355 Chr.21: 32742771-32742771 on Build GRCh38
rs748545544 Chr.21: 32742901-32742901 on Build GRCh38
rs893397787 Chr.21: 32743016-32743016 on Build GRCh38
rs751775459 Chr.21: 32743000-32743000 on Build GRCh38
rs139339104 Chr.21: 32742733-32742733 on Build GRCh38
rs535907014 Chr.21: 32743021-32743021 on Build GRCh38
rs767590123 Chr.21: 32743029-32743029 on Build GRCh38
rs920518634 Chr.21: 32742842-32742842 on Build GRCh38
rs551005149 Chr.21: 32743039-32743039 on Build GRCh38
rs547166247 Chr.21: 32742947-32742947 on Build GRCh38
rs866453789 Chr.21: 32742844-32742844 on Build GRCh38
rs770639748 Chr.21: 32742761-32742761 on Build GRCh38
rs1016373355 Chr.21: 32743164-32743164 on Build GRCh38
rs891091669 Chr.21: 32743131-32743131 on Build GRCh38
rs528716750 Chr.21: 32742921-32742921 on Build GRCh38
rs768468584 Chr.21: 32743040-32743041 on Build GRCh38
rs918789475 Chr.21: 32742753-32742753 on Build GRCh38
rs985764855 Chr.21: 32743148-32743148 on Build GRCh38
rs182647482 Chr.21: 32742718-32742718 on Build GRCh38
rs1028341897 Chr.21: 32743077-32743077 on Build GRCh38
rs2298365 Chr.21: 32743144-32743144 on Build GRCh38

View Full Product Details