Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36141575-36141930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR3
Entrez Gene ID
874
Gene Name
carbonyl reductase 3
Gene Aliases
HEL-S-25, SDR21C2, hCBR3
UniGene
Hs.154510
Gene Chromosome Location
Chr.21: 36134901-36146562 on Build GRCh38
Transcripts
NM_001236.3 XM_011529772.2 NR_038892.1 NR_038893.1 NR_038894.1

Gene Symbol
CBR3-AS1
Entrez Gene ID
100506428
Gene Name
CBR3 antisense RNA 1
Gene Aliases
PlncRNA-1, PlncRNA1
UniGene
Hs.651214
Gene Chromosome Location
Chr.21: 36131767-36156308 on Build GRCh38
Transcripts
NM_001236.3 XM_011529772.2 NR_038892.1 NR_038893.1 NR_038894.1

SNP Detail

SNP ID Chromosome Location
rs570939615 Chr.21: 36141665-36141665 on Build GRCh38
rs181555498 Chr.21: 36141856-36141856 on Build GRCh38
rs1025598934 Chr.21: 36141770-36141770 on Build GRCh38
rs770163843 Chr.21: 36141718-36141718 on Build GRCh38
rs45568242 Chr.21: 36141766-36141766 on Build GRCh38
rs918487380 Chr.21: 36141627-36141627 on Build GRCh38
rs536594937 Chr.21: 36141626-36141626 on Build GRCh38
rs1019629693 Chr.21: 36141757-36141757 on Build GRCh38
rs1001023105 Chr.21: 36141719-36141719 on Build GRCh38
rs952549747 Chr.21: 36141638-36141638 on Build GRCh38
rs774729464 Chr.21: 36141641-36141641 on Build GRCh38
rs949681508 Chr.21: 36141683-36141683 on Build GRCh38
rs1038962394 Chr.21: 36141773-36141773 on Build GRCh38
rs73902172 Chr.21: 36141754-36141754 on Build GRCh38
rs889886108 Chr.21: 36141753-36141753 on Build GRCh38
rs1042644365 Chr.21: 36141707-36141707 on Build GRCh38
rs761737831 Chr.21: 36141643-36141643 on Build GRCh38
rs922268184 Chr.21: 36141690-36141690 on Build GRCh38
rs992965285 Chr.21: 36141899-36141899 on Build GRCh38
rs575657201 Chr.21: 36141902-36141902 on Build GRCh38
rs901191750 Chr.21: 36141765-36141765 on Build GRCh38
rs753579029 Chr.21: 36141885-36141885 on Build GRCh38
rs952602083 Chr.21: 36141804-36141804 on Build GRCh38
rs750710493 Chr.21: 36141836-36141836 on Build GRCh38
rs548064480 Chr.21: 36141795-36141798 on Build GRCh38
rs960661555 Chr.21: 36141614-36141614 on Build GRCh38
rs937049790 Chr.21: 36141657-36141657 on Build GRCh38
rs1039430715 Chr.21: 36141684-36141684 on Build GRCh38
rs138002224 Chr.21: 36141658-36141658 on Build GRCh38
rs927105384 Chr.21: 36141650-36141650 on Build GRCh38
rs925778636 Chr.21: 36141662-36141662 on Build GRCh38
rs9305578 Chr.21: 36141857-36141857 on Build GRCh38
rs45614331 Chr.21: 36141874-36141874 on Build GRCh38
rs972620697 Chr.21: 36141618-36141618 on Build GRCh38
rs983959305 Chr.21: 36141833-36141833 on Build GRCh38
rs1053842465 Chr.21: 36141752-36141752 on Build GRCh38

View Full Product Details