Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24430750-24431272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3310095 c.438G>A p.L146L Chr.22: 24431115-24431115 on Build GRCh38
5859056 c.479delG p.*160fs? Chr.22: 24431074-24431074 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188266859 Chr.22: 24431237-24431237 on Build GRCh38
rs1017534517 Chr.22: 24430855-24430855 on Build GRCh38
rs921980846 Chr.22: 24431120-24431120 on Build GRCh38
rs200338456 Chr.22: 24431190-24431190 on Build GRCh38
rs973243697 Chr.22: 24430858-24430858 on Build GRCh38
rs748590001 Chr.22: 24431056-24431056 on Build GRCh38
rs530269607 Chr.22: 24430804-24430804 on Build GRCh38
rs556220777 Chr.22: 24431045-24431045 on Build GRCh38
rs970259863 Chr.22: 24430957-24430957 on Build GRCh38
rs980107330 Chr.22: 24431213-24431213 on Build GRCh38
rs1007201619 Chr.22: 24431079-24431079 on Build GRCh38
rs1038922395 Chr.22: 24430814-24430814 on Build GRCh38
rs536083474 Chr.22: 24431170-24431170 on Build GRCh38
rs766507827 Chr.22: 24431141-24431141 on Build GRCh38
rs780296946 Chr.22: 24431241-24431241 on Build GRCh38
rs776388560 Chr.22: 24431124-24431124 on Build GRCh38
rs900334420 Chr.22: 24430826-24430826 on Build GRCh38
rs12158888 Chr.22: 24431040-24431040 on Build GRCh38
rs896274082 Chr.22: 24430894-24430894 on Build GRCh38
rs549155920 Chr.22: 24431055-24431055 on Build GRCh38
rs750494022 Chr.22: 24431209-24431209 on Build GRCh38
rs926243857 Chr.22: 24430967-24430967 on Build GRCh38
rs8141793 Chr.22: 24431111-24431111 on Build GRCh38
rs912006557 Chr.22: 24430812-24430812 on Build GRCh38
rs115664791 Chr.22: 24431248-24431248 on Build GRCh38
rs956472785 Chr.22: 24431035-24431035 on Build GRCh38
rs571205667 Chr.22: 24431080-24431080 on Build GRCh38
rs917212962 Chr.22: 24430909-24430909 on Build GRCh38
rs891999970 Chr.22: 24430994-24430994 on Build GRCh38
rs930437360 Chr.22: 24430836-24430836 on Build GRCh38
rs8142326 Chr.22: 24430813-24430813 on Build GRCh38
rs878918820 Chr.22: 24431210-24431210 on Build GRCh38
rs1034634757 Chr.22: 24430892-24430892 on Build GRCh38
rs1000393741 Chr.22: 24431249-24431249 on Build GRCh38
rs1048983201 Chr.22: 24430849-24430849 on Build GRCh38
rs992950162 Chr.22: 24430908-24430908 on Build GRCh38
rs773987488 Chr.22: 24431139-24431139 on Build GRCh38
rs772627860 Chr.22: 24431094-24431094 on Build GRCh38
rs772563472 Chr.22: 24431074-24431074 on Build GRCh38
rs886386743 Chr.22: 24431178-24431178 on Build GRCh38
rs1011833730 Chr.22: 24431229-24431229 on Build GRCh38
rs925974774 Chr.22: 24431225-24431225 on Build GRCh38
rs3761423 Chr.22: 24430895-24430895 on Build GRCh38
rs754055334 Chr.22: 24431242-24431242 on Build GRCh38
rs1004527042 Chr.22: 24430887-24430887 on Build GRCh38
rs988790161 Chr.22: 24430782-24430782 on Build GRCh38
rs970022874 Chr.22: 24431212-24431212 on Build GRCh38
rs572148473 Chr.22: 24431115-24431115 on Build GRCh38
rs768979309 Chr.22: 24431024-24431024 on Build GRCh38
rs765106649 Chr.22: 24431147-24431147 on Build GRCh38
rs138745403 Chr.22: 24430857-24430857 on Build GRCh38
rs1045014211 Chr.22: 24430927-24430927 on Build GRCh38
rs936274111 Chr.22: 24430975-24430975 on Build GRCh38
rs1049349309 Chr.22: 24431166-24431166 on Build GRCh38
rs948706222 Chr.22: 24430925-24430925 on Build GRCh38
rs766180812 Chr.22: 24431117-24431117 on Build GRCh38
rs1033286581 Chr.22: 24431021-24431021 on Build GRCh38
rs529199418 Chr.22: 24430976-24430976 on Build GRCh38
rs919115696 Chr.22: 24430869-24430869 on Build GRCh38
rs896158082 Chr.22: 24431207-24431207 on Build GRCh38
rs538370231 Chr.22: 24431083-24431083 on Build GRCh38
rs576804195 Chr.22: 24430998-24430998 on Build GRCh38
rs995195302 Chr.22: 24431146-24431146 on Build GRCh38
rs554341423 Chr.22: 24431185-24431185 on Build GRCh38

View Full Product Details