Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24437966-24438496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

SNP Detail

SNP ID Chromosome Location
rs571140317 Chr.22: 24438082-24438082 on Build GRCh38
rs1001477358 Chr.22: 24438030-24438030 on Build GRCh38
rs565547686 Chr.22: 24438158-24438158 on Build GRCh38
rs1018490560 Chr.22: 24438465-24438465 on Build GRCh38
rs974399986 Chr.22: 24438119-24438119 on Build GRCh38
rs752803125 Chr.22: 24438036-24438036 on Build GRCh38
rs778014952 Chr.22: 24438210-24438210 on Build GRCh38
rs942032714 Chr.22: 24438048-24438048 on Build GRCh38
rs769413554 Chr.22: 24438420-24438420 on Build GRCh38
rs987600376 Chr.22: 24438090-24438090 on Build GRCh38
rs549775080 Chr.22: 24438078-24438078 on Build GRCh38
rs983396105 Chr.22: 24438240-24438240 on Build GRCh38
rs1001916541 Chr.22: 24438135-24438135 on Build GRCh38
rs949301498 Chr.22: 24438463-24438463 on Build GRCh38
rs779915242 Chr.22: 24438423-24438423 on Build GRCh38
rs745661175 Chr.22: 24438419-24438419 on Build GRCh38
rs1045829385 Chr.22: 24438022-24438022 on Build GRCh38
rs923409422 Chr.22: 24438387-24438387 on Build GRCh38
rs200817845 Chr.22: 24438339-24438341 on Build GRCh38
rs746654067 Chr.22: 24438291-24438291 on Build GRCh38
rs887739462 Chr.22: 24438043-24438043 on Build GRCh38
rs547062518 Chr.22: 24438113-24438113 on Build GRCh38
rs752671076 Chr.22: 24438061-24438061 on Build GRCh38
rs923460383 Chr.22: 24438005-24438005 on Build GRCh38
rs930508747 Chr.22: 24438178-24438178 on Build GRCh38
rs116001575 Chr.22: 24438105-24438105 on Build GRCh38
rs542821974 Chr.22: 24438441-24438441 on Build GRCh38
rs1008385734 Chr.22: 24438462-24438462 on Build GRCh38
rs765614095 Chr.22: 24438007-24438007 on Build GRCh38
rs950144460 Chr.22: 24438008-24438008 on Build GRCh38
rs1013941886 Chr.22: 24438070-24438070 on Build GRCh38
rs868608620 Chr.22: 24438358-24438358 on Build GRCh38
rs967634627 Chr.22: 24438328-24438328 on Build GRCh38
rs905849385 Chr.22: 24438028-24438028 on Build GRCh38
rs932040932 Chr.22: 24438011-24438011 on Build GRCh38
rs1036805246 Chr.22: 24438054-24438054 on Build GRCh38
rs987812214 Chr.22: 24438301-24438301 on Build GRCh38
rs905948486 Chr.22: 24438361-24438361 on Build GRCh38
rs1021066060 Chr.22: 24438305-24438305 on Build GRCh38
rs1032092057 Chr.22: 24438147-24438147 on Build GRCh38
rs920298486 Chr.22: 24438142-24438142 on Build GRCh38
rs897838411 Chr.22: 24438459-24438459 on Build GRCh38
rs893144868 Chr.22: 24438397-24438397 on Build GRCh38
rs34452689 Chr.22: 24438311-24438312 on Build GRCh38
rs536081599 Chr.22: 24438366-24438366 on Build GRCh38
rs1033006989 Chr.22: 24438035-24438035 on Build GRCh38
rs573837965 Chr.22: 24438391-24438391 on Build GRCh38
rs893386801 Chr.22: 24438052-24438052 on Build GRCh38
rs907673217 Chr.22: 24438268-24438268 on Build GRCh38
rs148729189 Chr.22: 24438393-24438393 on Build GRCh38
rs949933565 Chr.22: 24438314-24438314 on Build GRCh38

View Full Product Details