Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154030174-154030687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF26-AS1
Entrez Gene ID
100507524
Gene Name
ARHGEF26 antisense RNA 1
Gene Aliases
-
UniGene
Hs.370221
Gene Chromosome Location
Chr.3: 154024401-154121210 on Build GRCh38
Transcripts
NR_037901.1

SNP Detail

SNP ID Chromosome Location
rs536268991 Chr.3: 154030348-154030348 on Build GRCh38
rs947632534 Chr.3: 154030298-154030298 on Build GRCh38
rs751567886 Chr.3: 154030648-154030648 on Build GRCh38
rs61791506 Chr.3: 154030429-154030429 on Build GRCh38
rs764119583 Chr.3: 154030591-154030591 on Build GRCh38
rs1033709753 Chr.3: 154030418-154030418 on Build GRCh38
rs899750829 Chr.3: 154030371-154030371 on Build GRCh38
rs1039503029 Chr.3: 154030362-154030362 on Build GRCh38
rs115184408 Chr.3: 154030627-154030627 on Build GRCh38
rs537508484 Chr.3: 154030438-154030438 on Build GRCh38
rs975714733 Chr.3: 154030653-154030653 on Build GRCh38
rs943879740 Chr.3: 154030346-154030346 on Build GRCh38
rs754960494 Chr.3: 154030560-154030560 on Build GRCh38
rs538443303 Chr.3: 154030283-154030283 on Build GRCh38
rs66547709 Chr.3: 154030503-154030504 on Build GRCh38
rs992075299 Chr.3: 154030605-154030605 on Build GRCh38
rs376976349 Chr.3: 154030344-154030344 on Build GRCh38
rs111825950 Chr.3: 154030502-154030503 on Build GRCh38
rs890854535 Chr.3: 154030330-154030330 on Build GRCh38
rs1035796182 Chr.3: 154030581-154030581 on Build GRCh38
rs71308473 Chr.3: 154030522-154030522 on Build GRCh38
rs547327859 Chr.3: 154030297-154030297 on Build GRCh38
rs965339612 Chr.3: 154030652-154030652 on Build GRCh38
rs142563748 Chr.3: 154030501-154030502 on Build GRCh38
rs1004325548 Chr.3: 154030570-154030570 on Build GRCh38
rs6810296 Chr.3: 154030561-154030561 on Build GRCh38
rs1464414 Chr.3: 154030234-154030234 on Build GRCh38
rs869039395 Chr.3: 154030505-154030506 on Build GRCh38
rs892669032 Chr.3: 154030278-154030278 on Build GRCh38
rs1035890084 Chr.3: 154030252-154030252 on Build GRCh38
rs541624844 Chr.3: 154030573-154030573 on Build GRCh38
rs77264317 Chr.3: 154030313-154030313 on Build GRCh38
rs960124114 Chr.3: 154030583-154030583 on Build GRCh38
rs575155771 Chr.3: 154030649-154030649 on Build GRCh38
rs760375179 Chr.3: 154030472-154030472 on Build GRCh38
rs924949662 Chr.3: 154030309-154030309 on Build GRCh38
rs1023096797 Chr.3: 154030624-154030624 on Build GRCh38
rs766400344 Chr.3: 154030643-154030643 on Build GRCh38
rs951481606 Chr.3: 154030206-154030206 on Build GRCh38
rs960467234 Chr.3: 154030265-154030265 on Build GRCh38
rs760952149 Chr.3: 154030388-154030388 on Build GRCh38
rs199795405 Chr.3: 154030505-154030505 on Build GRCh38
rs1052115257 Chr.3: 154030315-154030315 on Build GRCh38
rs554917130 Chr.3: 154030408-154030408 on Build GRCh38
rs549718792 Chr.3: 154030228-154030228 on Build GRCh38
rs745433547 Chr.3: 154030263-154030263 on Build GRCh38
rs994958357 Chr.3: 154030446-154030446 on Build GRCh38
rs3071728 Chr.3: 154030504-154030505 on Build GRCh38
rs979070991 Chr.3: 154030304-154030304 on Build GRCh38

View Full Product Details