Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79862562-79863074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCAT4
Entrez Gene ID
118425
Gene Name
prostate cancer associated transcript 4 (non-protein coding)
Gene Aliases
GDEP, PCA4, PCAN1
UniGene
Hs.104215
Gene Chromosome Location
Chr.4: 79827471-79863247 on Build GRCh38
Transcripts
NR_026555.1

SNP Detail

SNP ID Chromosome Location
rs1030908369 Chr.4: 79862852-79862852 on Build GRCh38
rs899137713 Chr.4: 79862650-79862650 on Build GRCh38
rs996291845 Chr.4: 79862830-79862830 on Build GRCh38
rs568556574 Chr.4: 79862869-79862869 on Build GRCh38
rs142086169 Chr.4: 79862686-79862686 on Build GRCh38
rs33932804 Chr.4: 79862726-79862726 on Build GRCh38
rs996395227 Chr.4: 79863013-79863014 on Build GRCh38
rs996655041 Chr.4: 79862612-79862612 on Build GRCh38
rs144063864 Chr.4: 79863015-79863016 on Build GRCh38
rs1056612542 Chr.4: 79862851-79862851 on Build GRCh38
rs368211656 Chr.4: 79862980-79862980 on Build GRCh38
rs960582136 Chr.4: 79862858-79862858 on Build GRCh38
rs112179008 Chr.4: 79862642-79862642 on Build GRCh38
rs956077894 Chr.4: 79863012-79863012 on Build GRCh38
rs1044488355 Chr.4: 79862622-79862622 on Build GRCh38
rs766477336 Chr.4: 79862725-79862726 on Build GRCh38
rs185309408 Chr.4: 79862804-79862804 on Build GRCh38
rs990420785 Chr.4: 79862715-79862715 on Build GRCh38
rs4690162 Chr.4: 79863019-79863019 on Build GRCh38
rs538576105 Chr.4: 79862826-79862826 on Build GRCh38
rs947919871 Chr.4: 79862920-79862920 on Build GRCh38
rs756911154 Chr.4: 79862663-79862663 on Build GRCh38
rs117026637 Chr.4: 79862819-79862819 on Build GRCh38
rs747400401 Chr.4: 79862833-79862833 on Build GRCh38
rs145896074 Chr.4: 79862879-79862879 on Build GRCh38
rs1031705950 Chr.4: 79862681-79862681 on Build GRCh38
rs567252530 Chr.4: 79862618-79862618 on Build GRCh38
rs1009007905 Chr.4: 79862745-79862745 on Build GRCh38
rs80057303 Chr.4: 79862997-79862997 on Build GRCh38
rs914779565 Chr.4: 79862798-79862798 on Build GRCh38
rs898341209 Chr.4: 79862608-79862608 on Build GRCh38
rs557382402 Chr.4: 79862896-79862896 on Build GRCh38
rs1040632684 Chr.4: 79863005-79863005 on Build GRCh38
rs529453270 Chr.4: 79862992-79862992 on Build GRCh38
rs920717861 Chr.4: 79862950-79862950 on Build GRCh38
rs748938321 Chr.4: 79862636-79862636 on Build GRCh38
rs760179113 Chr.4: 79862847-79862847 on Build GRCh38
rs967699560 Chr.4: 79862840-79862840 on Build GRCh38
rs557852736 Chr.4: 79862864-79862864 on Build GRCh38
rs112969345 Chr.4: 79862939-79862939 on Build GRCh38
rs890956209 Chr.4: 79863000-79863000 on Build GRCh38
rs930626671 Chr.4: 79862970-79862970 on Build GRCh38
rs1008351634 Chr.4: 79863002-79863002 on Build GRCh38
rs181055430 Chr.4: 79862714-79862714 on Build GRCh38
rs546161218 Chr.4: 79862834-79862834 on Build GRCh38
rs1032134887 Chr.4: 79863011-79863011 on Build GRCh38
rs955466770 Chr.4: 79862651-79862651 on Build GRCh38
rs367725525 Chr.4: 79862605-79862607 on Build GRCh38
rs543240822 Chr.4: 79862989-79862989 on Build GRCh38
rs898394931 Chr.4: 79862881-79862881 on Build GRCh38
rs765910629 Chr.4: 79862867-79862867 on Build GRCh38

View Full Product Details