Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 86620357-86620858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7C
Entrez Gene ID
1350
Gene Name
cytochrome c oxidase subunit 7C
Gene Aliases
-
UniGene
Hs.430075
Gene Chromosome Location
Chr.5: 86617967-86620766 on Build GRCh38
Transcripts
NM_001867.2 NR_037402.1

Gene Symbol
MIR3607
Entrez Gene ID
100500805
Gene Name
microRNA 3607
Gene Aliases
mir-3607
UniGene
Gene Chromosome Location
Chr.5: 86620497-86620575 on Build GRCh38
Transcripts
NM_001867.2 NR_037402.1

SNP Detail

SNP ID Chromosome Location
rs933190779 Chr.5: 86620758-86620758 on Build GRCh38
rs35118667 Chr.5: 86620584-86620585 on Build GRCh38
rs188162668 Chr.5: 86620754-86620754 on Build GRCh38
rs946823177 Chr.5: 86620683-86620683 on Build GRCh38
rs900358635 Chr.5: 86620748-86620748 on Build GRCh38
rs544282127 Chr.5: 86620512-86620512 on Build GRCh38
rs768644623 Chr.5: 86620504-86620504 on Build GRCh38
rs898808052 Chr.5: 86620824-86620824 on Build GRCh38
rs756265841 Chr.5: 86620778-86620778 on Build GRCh38
rs1042892172 Chr.5: 86620689-86620689 on Build GRCh38
rs116683123 Chr.5: 86620488-86620488 on Build GRCh38
rs548951630 Chr.5: 86620780-86620780 on Build GRCh38
rs1007216592 Chr.5: 86620494-86620494 on Build GRCh38
rs915219131 Chr.5: 86620609-86620609 on Build GRCh38
rs927958400 Chr.5: 86620634-86620634 on Build GRCh38
rs551657381 Chr.5: 86620764-86620764 on Build GRCh38
rs562373142 Chr.5: 86620553-86620553 on Build GRCh38
rs973564244 Chr.5: 86620574-86620574 on Build GRCh38
rs1052503115 Chr.5: 86620470-86620470 on Build GRCh38
rs1051599465 Chr.5: 86620763-86620763 on Build GRCh38
rs935400527 Chr.5: 86620464-86620464 on Build GRCh38
rs151331151 Chr.5: 86620651-86620651 on Build GRCh38
rs767881963 Chr.5: 86620595-86620595 on Build GRCh38
rs537624217 Chr.5: 86620572-86620574 on Build GRCh38
rs1015869813 Chr.5: 86620498-86620498 on Build GRCh38
rs1037348558 Chr.5: 86620796-86620796 on Build GRCh38
rs756127021 Chr.5: 86620643-86620646 on Build GRCh38
rs1030457418 Chr.5: 86620478-86620478 on Build GRCh38
rs573163827 Chr.5: 86620457-86620457 on Build GRCh38
rs183185817 Chr.5: 86620454-86620454 on Build GRCh38
rs561127382 Chr.5: 86620792-86620792 on Build GRCh38
rs982299329 Chr.5: 86620549-86620549 on Build GRCh38
rs558050094 Chr.5: 86620449-86620449 on Build GRCh38
rs988567714 Chr.5: 86620643-86620643 on Build GRCh38
rs767421652 Chr.5: 86620503-86620503 on Build GRCh38
rs902984582 Chr.5: 86620446-86620446 on Build GRCh38
rs890155269 Chr.5: 86620537-86620537 on Build GRCh38
rs1014651846 Chr.5: 86620555-86620555 on Build GRCh38
rs142485335 Chr.5: 86620596-86620596 on Build GRCh38
rs947297977 Chr.5: 86620635-86620635 on Build GRCh38
rs61188008 Chr.5: 86620720-86620720 on Build GRCh38
rs778238764 Chr.5: 86620810-86620810 on Build GRCh38
rs750211286 Chr.5: 86620739-86620739 on Build GRCh38
rs890396197 Chr.5: 86620797-86620798 on Build GRCh38
rs113797294 Chr.5: 86620505-86620505 on Build GRCh38
rs1041569153 Chr.5: 86620432-86620432 on Build GRCh38
rs758600747 Chr.5: 86620675-86620675 on Build GRCh38
rs752860963 Chr.5: 86620657-86620657 on Build GRCh38
rs527535382 Chr.5: 86620762-86620762 on Build GRCh38
rs760750723 Chr.5: 86620583-86620583 on Build GRCh38
rs947265485 Chr.5: 86620426-86620426 on Build GRCh38
rs555927484 Chr.5: 86620495-86620495 on Build GRCh38
rs777892298 Chr.5: 86620691-86620691 on Build GRCh38
rs1004564099 Chr.5: 86620541-86620541 on Build GRCh38

View Full Product Details