Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26522815-26523333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCG11
Entrez Gene ID
493812
Gene Name
HLA complex group 11 (non-protein coding)
Gene Aliases
CTA-14H9.3, bK14H9.3
UniGene
Hs.272939
Gene Chromosome Location
Chr.6: 26521706-26527393 on Build GRCh38
Transcripts
NR_026790.1

SNP Detail

SNP ID Chromosome Location
rs563970443 Chr.6: 26523063-26523063 on Build GRCh38
rs903509607 Chr.6: 26523188-26523188 on Build GRCh38
rs529656293 Chr.6: 26523067-26523067 on Build GRCh38
rs372792269 Chr.6: 26523033-26523033 on Build GRCh38
rs565092020 Chr.6: 26523305-26523305 on Build GRCh38
rs528547999 Chr.6: 26523276-26523276 on Build GRCh38
rs769876904 Chr.6: 26522953-26522953 on Build GRCh38
rs1056319991 Chr.6: 26523070-26523070 on Build GRCh38
rs543230551 Chr.6: 26523124-26523124 on Build GRCh38
rs868127423 Chr.6: 26523175-26523175 on Build GRCh38
rs775376062 Chr.6: 26523209-26523209 on Build GRCh38
rs911218727 Chr.6: 26522965-26522965 on Build GRCh38
rs759288738 Chr.6: 26522911-26522911 on Build GRCh38
rs571788306 Chr.6: 26522949-26522949 on Build GRCh38
rs76245152 Chr.6: 26523134-26523134 on Build GRCh38
rs577576153 Chr.6: 26523020-26523020 on Build GRCh38
rs557435506 Chr.6: 26523018-26523018 on Build GRCh38
rs1012156254 Chr.6: 26523142-26523142 on Build GRCh38
rs541143821 Chr.6: 26522988-26522988 on Build GRCh38
rs760457086 Chr.6: 26523139-26523139 on Build GRCh38
rs956499994 Chr.6: 26523282-26523282 on Build GRCh38
rs551865283 Chr.6: 26523286-26523286 on Build GRCh38
rs934262688 Chr.6: 26522937-26522937 on Build GRCh38
rs140580139 Chr.6: 26523169-26523169 on Build GRCh38
rs918704999 Chr.6: 26523069-26523069 on Build GRCh38
rs1044997890 Chr.6: 26523144-26523144 on Build GRCh38
rs773955908 Chr.6: 26522880-26522880 on Build GRCh38
rs896356282 Chr.6: 26523086-26523086 on Build GRCh38
rs1000514539 Chr.6: 26523263-26523263 on Build GRCh38

View Full Product Details