Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26524192-26524691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCG11
Entrez Gene ID
493812
Gene Name
HLA complex group 11 (non-protein coding)
Gene Aliases
CTA-14H9.3, bK14H9.3
UniGene
Hs.272939
Gene Chromosome Location
Chr.6: 26521706-26527393 on Build GRCh38
Transcripts
NR_026790.1

SNP Detail

SNP ID Chromosome Location
rs1051887289 Chr.6: 26524435-26524435 on Build GRCh38
rs370034487 Chr.6: 26524428-26524431 on Build GRCh38
rs144942068 Chr.6: 26524443-26524443 on Build GRCh38
rs186277542 Chr.6: 26524527-26524527 on Build GRCh38
rs750355967 Chr.6: 26524615-26524615 on Build GRCh38
rs925108731 Chr.6: 26524356-26524356 on Build GRCh38
rs565692042 Chr.6: 26524339-26524339 on Build GRCh38
rs1025864587 Chr.6: 26524553-26524553 on Build GRCh38
rs887374503 Chr.6: 26524565-26524565 on Build GRCh38
rs913838063 Chr.6: 26524422-26524422 on Build GRCh38
rs372046728 Chr.6: 26524486-26524496 on Build GRCh38
rs529119809 Chr.6: 26524308-26524308 on Build GRCh38
rs149917024 Chr.6: 26524388-26524388 on Build GRCh38
rs537173368 Chr.6: 26524470-26524470 on Build GRCh38
rs564551639 Chr.6: 26524639-26524639 on Build GRCh38
rs573692819 Chr.6: 26524548-26524548 on Build GRCh38
rs1037492631 Chr.6: 26524456-26524456 on Build GRCh38
rs1025033957 Chr.6: 26524238-26524239 on Build GRCh38
rs936548690 Chr.6: 26524390-26524390 on Build GRCh38
rs776448177 Chr.6: 26524430-26524430 on Build GRCh38
rs898643555 Chr.6: 26524457-26524458 on Build GRCh38
rs1013547680 Chr.6: 26524622-26524622 on Build GRCh38
rs995701949 Chr.6: 26524477-26524477 on Build GRCh38
rs560969988 Chr.6: 26524304-26524304 on Build GRCh38
rs988000167 Chr.6: 26524413-26524413 on Build GRCh38
rs765197032 Chr.6: 26524526-26524526 on Build GRCh38
rs575541074 Chr.6: 26524272-26524272 on Build GRCh38
rs373465369 Chr.6: 26524446-26524446 on Build GRCh38
rs548874432 Chr.6: 26524312-26524312 on Build GRCh38
rs969446981 Chr.6: 26524307-26524307 on Build GRCh38
rs943108839 Chr.6: 26524449-26524449 on Build GRCh38
rs550544710 Chr.6: 26524301-26524301 on Build GRCh38
rs768342672 Chr.6: 26524378-26524378 on Build GRCh38

View Full Product Details