Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26525336-26525876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCG11
Entrez Gene ID
493812
Gene Name
HLA complex group 11 (non-protein coding)
Gene Aliases
CTA-14H9.3, bK14H9.3
UniGene
Hs.272939
Gene Chromosome Location
Chr.6: 26521706-26527393 on Build GRCh38
Transcripts
NR_026790.1

SNP Detail

SNP ID Chromosome Location
rs145154602 Chr.6: 26525717-26525717 on Build GRCh38
rs35998206 Chr.6: 26525636-26525637 on Build GRCh38
rs971174606 Chr.6: 26525711-26525711 on Build GRCh38
rs1027596329 Chr.6: 26525596-26525596 on Build GRCh38
rs574635728 Chr.6: 26525586-26525586 on Build GRCh38
rs950422238 Chr.6: 26525598-26525598 on Build GRCh38
rs370613572 Chr.6: 26525499-26525499 on Build GRCh38
rs41267935 Chr.6: 26525839-26525839 on Build GRCh38
rs183207654 Chr.6: 26525464-26525464 on Build GRCh38
rs398039009 Chr.6: 26525657-26525658 on Build GRCh38
rs776114573 Chr.6: 26525447-26525447 on Build GRCh38
rs982263994 Chr.6: 26525492-26525492 on Build GRCh38
rs536250012 Chr.6: 26525372-26525372 on Build GRCh38
rs1054000358 Chr.6: 26525501-26525501 on Build GRCh38
rs551814881 Chr.6: 26525828-26525828 on Build GRCh38
rs374349878 Chr.6: 26525442-26525442 on Build GRCh38
rs773143542 Chr.6: 26525498-26525498 on Build GRCh38
rs752863257 Chr.6: 26525587-26525587 on Build GRCh38
rs560315427 Chr.6: 26525734-26525734 on Build GRCh38
rs938222507 Chr.6: 26525793-26525793 on Build GRCh38
rs767824387 Chr.6: 26525562-26525562 on Build GRCh38
rs964624216 Chr.6: 26525367-26525367 on Build GRCh38
rs983151096 Chr.6: 26525615-26525615 on Build GRCh38
rs1039175055 Chr.6: 26525840-26525840 on Build GRCh38
rs950452508 Chr.6: 26525437-26525437 on Build GRCh38
rs869078363 Chr.6: 26525636-26525638 on Build GRCh38
rs552733500 Chr.6: 26525411-26525411 on Build GRCh38
rs11963559 Chr.6: 26525543-26525543 on Build GRCh38
rs774437777 Chr.6: 26525531-26525531 on Build GRCh38
rs945344190 Chr.6: 26525827-26525827 on Build GRCh38
rs71274287 Chr.6: 26525637-26525638 on Build GRCh38
rs572381139 Chr.6: 26525478-26525478 on Build GRCh38
rs567137243 Chr.6: 26525422-26525422 on Build GRCh38
rs753123516 Chr.6: 26525382-26525382 on Build GRCh38
rs199627535 Chr.6: 26525754-26525754 on Build GRCh38
rs35355150 Chr.6: 26525811-26525811 on Build GRCh38
rs529544932 Chr.6: 26525553-26525553 on Build GRCh38
rs544251944 Chr.6: 26525534-26525534 on Build GRCh38
rs766474500 Chr.6: 26525530-26525530 on Build GRCh38
rs187667437 Chr.6: 26525508-26525508 on Build GRCh38
rs994753770 Chr.6: 26525573-26525573 on Build GRCh38
rs1017455499 Chr.6: 26525550-26525550 on Build GRCh38
rs1027315485 Chr.6: 26525391-26525391 on Build GRCh38
rs567359335 Chr.6: 26525829-26525829 on Build GRCh38
rs893798128 Chr.6: 26525502-26525502 on Build GRCh38
rs77690026 Chr.6: 26525636-26525636 on Build GRCh38
rs1024452148 Chr.6: 26525616-26525616 on Build GRCh38
rs528361709 Chr.6: 26525736-26525736 on Build GRCh38
rs565048533 Chr.6: 26525790-26525790 on Build GRCh38
rs191495157 Chr.6: 26525779-26525779 on Build GRCh38
rs989674695 Chr.6: 26525809-26525809 on Build GRCh38

View Full Product Details