Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 72967269-72967771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
UniGene
Hs.445324
Gene Chromosome Location
Chr.6: 72621843-73198851 on Build GRCh38
Transcripts
NM_001160130.1 NM_001160132.1 NM_001160133.1 NM_001160134.1 NM_019842.3 XM_017011058.1 NR_036244.1

Gene Symbol
MIR4282
Entrez Gene ID
100423005
Gene Name
microRNA 4282
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.6: 72967687-72967753 on Build GRCh38
Transcripts
NM_001160130.1 NM_001160132.1 NM_001160133.1 NM_001160134.1 NM_019842.3 XM_017011058.1 NR_036244.1

SNP Detail

SNP ID Chromosome Location
rs147415448 Chr.6: 72967425-72967425 on Build GRCh38
rs896233463 Chr.6: 72967631-72967631 on Build GRCh38
rs754079034 Chr.6: 72967527-72967527 on Build GRCh38
rs139667853 Chr.6: 72967668-72967668 on Build GRCh38
rs559031129 Chr.6: 72967514-72967514 on Build GRCh38
rs867054707 Chr.6: 72967592-72967592 on Build GRCh38
rs897109852 Chr.6: 72967395-72967395 on Build GRCh38
rs868762201 Chr.6: 72967725-72967725 on Build GRCh38
rs369812453 Chr.6: 72967607-72967607 on Build GRCh38
rs965149514 Chr.6: 72967300-72967300 on Build GRCh38
rs538304574 Chr.6: 72967392-72967392 on Build GRCh38
rs879641747 Chr.6: 72967616-72967616 on Build GRCh38
rs925508117 Chr.6: 72967489-72967489 on Build GRCh38
rs925570854 Chr.6: 72967366-72967366 on Build GRCh38
rs751344325 Chr.6: 72967421-72967421 on Build GRCh38
rs143011399 Chr.6: 72967317-72967317 on Build GRCh38
rs114793984 Chr.6: 72967445-72967445 on Build GRCh38
rs1046474466 Chr.6: 72967389-72967389 on Build GRCh38
rs945734026 Chr.6: 72967328-72967328 on Build GRCh38
rs755058572 Chr.6: 72967709-72967709 on Build GRCh38
rs142743833 Chr.6: 72967422-72967422 on Build GRCh38
rs544648922 Chr.6: 72967577-72967577 on Build GRCh38
rs898176214 Chr.6: 72967648-72967648 on Build GRCh38
rs914584995 Chr.6: 72967535-72967535 on Build GRCh38
rs148227278 Chr.6: 72967407-72967407 on Build GRCh38
rs978479407 Chr.6: 72967480-72967480 on Build GRCh38
rs887060455 Chr.6: 72967680-72967680 on Build GRCh38
rs779241143 Chr.6: 72967387-72967387 on Build GRCh38
rs967417461 Chr.6: 72967467-72967467 on Build GRCh38
rs1057352323 Chr.6: 72967623-72967623 on Build GRCh38
rs1044709462 Chr.6: 72967634-72967634 on Build GRCh38
rs563731704 Chr.6: 72967452-72967452 on Build GRCh38
rs192578171 Chr.6: 72967523-72967523 on Build GRCh38
rs1046441415 Chr.6: 72967567-72967567 on Build GRCh38
rs947633948 Chr.6: 72967563-72967563 on Build GRCh38
rs548777309 Chr.6: 72967495-72967495 on Build GRCh38
rs758937370 Chr.6: 72967379-72967380 on Build GRCh38
rs975915707 Chr.6: 72967742-72967742 on Build GRCh38
rs368837298 Chr.6: 72967586-72967586 on Build GRCh38
rs947816188 Chr.6: 72967633-72967633 on Build GRCh38
rs888401160 Chr.6: 72967408-72967411 on Build GRCh38
rs1021603786 Chr.6: 72967747-72967747 on Build GRCh38
rs764782943 Chr.6: 72967324-72967324 on Build GRCh38
rs976414564 Chr.6: 72967310-72967310 on Build GRCh38
rs947649158 Chr.6: 72967308-72967308 on Build GRCh38
rs542299073 Chr.6: 72967671-72967671 on Build GRCh38
rs757102382 Chr.6: 72967431-72967431 on Build GRCh38
rs965213886 Chr.6: 72967429-72967429 on Build GRCh38
rs924762053 Chr.6: 72967578-72967578 on Build GRCh38
rs1056226887 Chr.6: 72967384-72967384 on Build GRCh38
rs548722465 Chr.6: 72967496-72967496 on Build GRCh38
rs964082668 Chr.6: 72967729-72967729 on Build GRCh38
rs1048416606 Chr.6: 72967667-72967667 on Build GRCh38
rs75974559 Chr.6: 72967400-72967400 on Build GRCh38
rs759622522 Chr.6: 72967639-72967639 on Build GRCh38
rs775317107 Chr.6: 72967387-72967387 on Build GRCh38
rs530566146 Chr.6: 72967479-72967479 on Build GRCh38
rs935044512 Chr.6: 72967351-72967351 on Build GRCh38
rs1020048857 Chr.6: 72967450-72967450 on Build GRCh38

View Full Product Details