Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 163342427-163342951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZp451B082
Entrez Gene ID
401282
Gene Name
uncharacterized LOC401282
Gene Aliases
-
UniGene
Hs.444817
Gene Chromosome Location
Chr.6: 163338342-163347033 on Build GRCh38
Transcripts
NR_033862.1

SNP Detail

SNP ID Chromosome Location
rs924823864 Chr.6: 163342507-163342507 on Build GRCh38
rs766942350 Chr.6: 163342688-163342688 on Build GRCh38
rs906634428 Chr.6: 163342870-163342870 on Build GRCh38
rs1106872 Chr.6: 163342863-163342863 on Build GRCh38
rs186371242 Chr.6: 163342476-163342476 on Build GRCh38
rs1038118818 Chr.6: 163342580-163342580 on Build GRCh38
rs986268984 Chr.6: 163342486-163342486 on Build GRCh38
rs924369232 Chr.6: 163342724-163342724 on Build GRCh38
rs567509165 Chr.6: 163342843-163342843 on Build GRCh38
rs1002678890 Chr.6: 163342902-163342902 on Build GRCh38
rs954839901 Chr.6: 163342730-163342730 on Build GRCh38
rs987620011 Chr.6: 163342743-163342743 on Build GRCh38
rs962208105 Chr.6: 163342677-163342677 on Build GRCh38
rs776400252 Chr.6: 163342502-163342502 on Build GRCh38
rs943348006 Chr.6: 163342568-163342568 on Build GRCh38
rs977610036 Chr.6: 163342721-163342721 on Build GRCh38
rs760886 Chr.6: 163342619-163342619 on Build GRCh38
rs563592949 Chr.6: 163342658-163342658 on Build GRCh38
rs894604408 Chr.6: 163342663-163342663 on Build GRCh38
rs541835586 Chr.6: 163342494-163342494 on Build GRCh38
rs1021734662 Chr.6: 163342695-163342695 on Build GRCh38
rs933014368 Chr.6: 163342484-163342484 on Build GRCh38
rs758047959 Chr.6: 163342701-163342701 on Build GRCh38
rs191623285 Chr.6: 163342603-163342603 on Build GRCh38
rs760889 Chr.6: 163342814-163342814 on Build GRCh38
rs564095692 Chr.6: 163342651-163342651 on Build GRCh38
rs1051927962 Chr.6: 163342525-163342525 on Build GRCh38
rs551819715 Chr.6: 163342759-163342759 on Build GRCh38
rs1006386341 Chr.6: 163342671-163342671 on Build GRCh38
rs760887 Chr.6: 163342620-163342620 on Build GRCh38
rs910805226 Chr.6: 163342532-163342532 on Build GRCh38
rs76719030 Chr.6: 163342477-163342477 on Build GRCh38
rs749755956 Chr.6: 163342900-163342900 on Build GRCh38
rs4376332 Chr.6: 163342683-163342683 on Build GRCh38
rs545068165 Chr.6: 163342604-163342604 on Build GRCh38
rs945254883 Chr.6: 163342573-163342573 on Build GRCh38
rs760888 Chr.6: 163342789-163342789 on Build GRCh38
rs552924628 Chr.6: 163342594-163342594 on Build GRCh38
rs892307384 Chr.6: 163342566-163342566 on Build GRCh38
rs879172992 Chr.6: 163342499-163342499 on Build GRCh38
rs553872897 Chr.6: 163342524-163342524 on Build GRCh38
rs182265128 Chr.6: 163342750-163342750 on Build GRCh38
rs566047859 Chr.6: 163342737-163342737 on Build GRCh38
rs11962117 Chr.6: 163342818-163342818 on Build GRCh38
rs1035582519 Chr.6: 163342637-163342637 on Build GRCh38
rs148641402 Chr.6: 163342593-163342593 on Build GRCh38
rs568124792 Chr.6: 163342832-163342832 on Build GRCh38
rs371566205 Chr.6: 163342879-163342879 on Build GRCh38
rs972695749 Chr.6: 163342692-163342692 on Build GRCh38
rs529537158 Chr.6: 163342674-163342674 on Build GRCh38
rs771624572 Chr.6: 163342826-163342826 on Build GRCh38
rs954507941 Chr.6: 163342706-163342706 on Build GRCh38
rs1053581743 Chr.6: 163342560-163342560 on Build GRCh38
rs936388367 Chr.6: 163342769-163342769 on Build GRCh38
rs141187330 Chr.6: 163342905-163342905 on Build GRCh38

View Full Product Details