Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 163346587-163347130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZp451B082
Entrez Gene ID
401282
Gene Name
uncharacterized LOC401282
Gene Aliases
-
UniGene
Hs.444817
Gene Chromosome Location
Chr.6: 163338342-163347033 on Build GRCh38
Transcripts
NR_033862.1

SNP Detail

SNP ID Chromosome Location
rs957768700 Chr.6: 163346795-163346795 on Build GRCh38
rs769276782 Chr.6: 163347007-163347007 on Build GRCh38
rs80251108 Chr.6: 163347076-163347076 on Build GRCh38
rs1036687646 Chr.6: 163346755-163346755 on Build GRCh38
rs576315384 Chr.6: 163346672-163346672 on Build GRCh38
rs981633586 Chr.6: 163346898-163346898 on Build GRCh38
rs918329643 Chr.6: 163346816-163346816 on Build GRCh38
rs747826179 Chr.6: 163346998-163346998 on Build GRCh38
rs1041723470 Chr.6: 163346994-163346994 on Build GRCh38
rs538458258 Chr.6: 163346650-163346650 on Build GRCh38
rs985515655 Chr.6: 163346616-163346616 on Build GRCh38
rs926915974 Chr.6: 163346854-163346854 on Build GRCh38
rs187149088 Chr.6: 163346705-163346705 on Build GRCh38
rs981708527 Chr.6: 163346849-163346849 on Build GRCh38
rs1041754160 Chr.6: 163346750-163346750 on Build GRCh38
rs981068050 Chr.6: 163346650-163346650 on Build GRCh38
rs541209038 Chr.6: 163346760-163346760 on Build GRCh38
rs971599359 Chr.6: 163346840-163346840 on Build GRCh38
rs957575469 Chr.6: 163346637-163346637 on Build GRCh38
rs541270770 Chr.6: 163347088-163347088 on Build GRCh38
rs765093579 Chr.6: 163346656-163346656 on Build GRCh38
rs577999873 Chr.6: 163346796-163346796 on Build GRCh38
rs1036164026 Chr.6: 163346957-163346957 on Build GRCh38
rs78121609 Chr.6: 163346642-163346642 on Build GRCh38
rs890912239 Chr.6: 163347053-163347053 on Build GRCh38
rs4370339 Chr.6: 163346792-163346792 on Build GRCh38
rs901689483 Chr.6: 163346962-163346962 on Build GRCh38
rs545108672 Chr.6: 163347070-163347070 on Build GRCh38
rs914845774 Chr.6: 163346991-163346991 on Build GRCh38
rs114073352 Chr.6: 163346785-163346785 on Build GRCh38
rs112082537 Chr.6: 163346615-163346615 on Build GRCh38
rs1020470234 Chr.6: 163346834-163346834 on Build GRCh38
rs1006740971 Chr.6: 163347063-163347063 on Build GRCh38
rs928887533 Chr.6: 163346678-163346678 on Build GRCh38
rs1029695103 Chr.6: 163346614-163346614 on Build GRCh38
rs1035448813 Chr.6: 163347098-163347098 on Build GRCh38
rs898445814 Chr.6: 163347072-163347072 on Build GRCh38
rs984930407 Chr.6: 163346910-163346910 on Build GRCh38
rs937533818 Chr.6: 163346921-163346921 on Build GRCh38
rs1003112908 Chr.6: 163347074-163347074 on Build GRCh38
rs572561877 Chr.6: 163347079-163347079 on Build GRCh38
rs191120781 Chr.6: 163346744-163346744 on Build GRCh38
rs973777438 Chr.6: 163346635-163346635 on Build GRCh38
rs1007184474 Chr.6: 163346777-163346777 on Build GRCh38
rs1006930444 Chr.6: 163347019-163347019 on Build GRCh38
rs930101286 Chr.6: 163346675-163346675 on Build GRCh38
rs1046351363 Chr.6: 163346676-163346676 on Build GRCh38
rs150825705 Chr.6: 163346767-163346767 on Build GRCh38
rs999347455 Chr.6: 163346761-163346761 on Build GRCh38
rs1054939056 Chr.6: 163347023-163347023 on Build GRCh38
rs1020440743 Chr.6: 163347064-163347064 on Build GRCh38
rs910910559 Chr.6: 163346622-163346622 on Build GRCh38
rs780776609 Chr.6: 163346920-163346920 on Build GRCh38
rs111674553 Chr.6: 163346933-163346933 on Build GRCh38
rs940511177 Chr.6: 163346925-163346925 on Build GRCh38
rs772590745 Chr.6: 163347086-163347086 on Build GRCh38
rs150020782 Chr.6: 163346993-163346993 on Build GRCh38
rs754504349 Chr.6: 163346868-163346868 on Build GRCh38
rs981564199 Chr.6: 163347097-163347097 on Build GRCh38
rs893105589 Chr.6: 163346743-163346743 on Build GRCh38
rs1051862854 Chr.6: 163347033-163347033 on Build GRCh38
rs1011536141 Chr.6: 163346746-163346746 on Build GRCh38
rs537389418 Chr.6: 163346949-163346949 on Build GRCh38
rs989448618 Chr.6: 163346639-163346639 on Build GRCh38
rs139233115 Chr.6: 163346970-163346970 on Build GRCh38
rs951158069 Chr.6: 163347092-163347092 on Build GRCh38
rs951136147 Chr.6: 163346880-163346880 on Build GRCh38
rs780087066 Chr.6: 163346770-163346770 on Build GRCh38
rs1055791395 Chr.6: 163346731-163346731 on Build GRCh38
rs958613186 Chr.6: 163347077-163347077 on Build GRCh38
rs962376554 Chr.6: 163346623-163346623 on Build GRCh38
rs1018205971 Chr.6: 163346778-163346778 on Build GRCh38
rs1048816188 Chr.6: 163346688-163346688 on Build GRCh38

View Full Product Details