Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96979252-96979775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX6-AS1
Entrez Gene ID
285987
Gene Name
DLX6 antisense RNA 1
Gene Aliases
DLX6-AS, DLX6AS, Evf-2, NCRNA00212
UniGene
Hs.34969
Gene Chromosome Location
Chr.7: 96968515-97014065 on Build GRCh38
Transcripts
NR_015448.1

SNP Detail

SNP ID Chromosome Location
rs963827291 Chr.7: 96979400-96979402 on Build GRCh38
rs5012357 Chr.7: 96979747-96979747 on Build GRCh38
rs141602263 Chr.7: 96979702-96979702 on Build GRCh38
rs572645060 Chr.7: 96979466-96979466 on Build GRCh38
rs897843570 Chr.7: 96979645-96979645 on Build GRCh38
rs866315789 Chr.7: 96979721-96979721 on Build GRCh38
rs917346980 Chr.7: 96979428-96979428 on Build GRCh38
rs142486814 Chr.7: 96979677-96979677 on Build GRCh38
rs188855350 Chr.7: 96979547-96979547 on Build GRCh38
rs567365005 Chr.7: 96979408-96979408 on Build GRCh38
rs535920117 Chr.7: 96979419-96979419 on Build GRCh38
rs577996303 Chr.7: 96979514-96979514 on Build GRCh38
rs542362973 Chr.7: 96979541-96979541 on Build GRCh38
rs967966428 Chr.7: 96979333-96979333 on Build GRCh38
rs991972179 Chr.7: 96979426-96979426 on Build GRCh38
rs549334034 Chr.7: 96979378-96979378 on Build GRCh38
rs977718733 Chr.7: 96979461-96979461 on Build GRCh38
rs556820745 Chr.7: 96979347-96979347 on Build GRCh38
rs1032673244 Chr.7: 96979739-96979739 on Build GRCh38
rs1005758845 Chr.7: 96979372-96979372 on Build GRCh38
rs925943842 Chr.7: 96979406-96979406 on Build GRCh38
rs991526338 Chr.7: 96979688-96979688 on Build GRCh38
rs556018598 Chr.7: 96979460-96979460 on Build GRCh38
rs146777571 Chr.7: 96979682-96979682 on Build GRCh38
rs890883292 Chr.7: 96979609-96979609 on Build GRCh38
rs950162653 Chr.7: 96979563-96979563 on Build GRCh38
rs987990011 Chr.7: 96979297-96979297 on Build GRCh38
rs941603134 Chr.7: 96979486-96979486 on Build GRCh38
rs191554340 Chr.7: 96979712-96979712 on Build GRCh38
rs775472605 Chr.7: 96979671-96979671 on Build GRCh38
rs78786258 Chr.7: 96979746-96979747 on Build GRCh38
rs1045780017 Chr.7: 96979599-96979604 on Build GRCh38
rs558469816 Chr.7: 96979513-96979513 on Build GRCh38
rs563487190 Chr.7: 96979679-96979679 on Build GRCh38
rs139337247 Chr.7: 96979693-96979693 on Build GRCh38
rs958930039 Chr.7: 96979686-96979686 on Build GRCh38
rs950198338 Chr.7: 96979441-96979441 on Build GRCh38
rs952594862 Chr.7: 96979277-96979277 on Build GRCh38
rs1020845007 Chr.7: 96979327-96979327 on Build GRCh38
rs1046977879 Chr.7: 96979675-96979675 on Build GRCh38
rs908695444 Chr.7: 96979709-96979709 on Build GRCh38
rs559156535 Chr.7: 96979694-96979694 on Build GRCh38
rs1050807660 Chr.7: 96979595-96979595 on Build GRCh38
rs976478328 Chr.7: 96979410-96979410 on Build GRCh38
rs1009370182 Chr.7: 96979664-96979664 on Build GRCh38
rs541782820 Chr.7: 96979496-96979496 on Build GRCh38

View Full Product Details