Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93700191-93700685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM92A1
Entrez Gene ID
137392
Gene Name
family with sequence similarity 92 member A1
Gene Aliases
-
UniGene
Hs.125038
Gene Chromosome Location
Chr.8: 93700252-93732041 on Build GRCh38
Transcripts
NM_001283034.1 NM_145269.4 NR_104267.1 NR_104268.1 XM_005250787.2 NR_033858.1

Gene Symbol
LINC00535
Entrez Gene ID
642924
Gene Name
long intergenic non-protein coding RNA 535
Gene Aliases
-
UniGene
Hs.434882
Gene Chromosome Location
Chr.8: 93346467-93700433 on Build GRCh38
Transcripts
NM_001283034.1 NM_145269.4 NR_104267.1 NR_104268.1 XM_005250787.2 NR_033858.1

SNP Detail

SNP ID Chromosome Location
rs991479159 Chr.8: 93700390-93700390 on Build GRCh38
rs923681918 Chr.8: 93700224-93700224 on Build GRCh38
rs979596946 Chr.8: 93700395-93700395 on Build GRCh38
rs927604541 Chr.8: 93700321-93700321 on Build GRCh38
rs748564678 Chr.8: 93700345-93700345 on Build GRCh38
rs756089143 Chr.8: 93700416-93700416 on Build GRCh38
rs1004911129 Chr.8: 93700378-93700378 on Build GRCh38
rs956493044 Chr.8: 93700555-93700555 on Build GRCh38
rs964061488 Chr.8: 93700578-93700578 on Build GRCh38
rs576456014 Chr.8: 93700393-93700393 on Build GRCh38
rs931233747 Chr.8: 93700601-93700601 on Build GRCh38
rs879932422 Chr.8: 93700394-93700394 on Build GRCh38
rs1057477156 Chr.8: 93700489-93700489 on Build GRCh38
rs1040087159 Chr.8: 93700645-93700645 on Build GRCh38
rs993100465 Chr.8: 93700607-93700607 on Build GRCh38
rs559101616 Chr.8: 93700427-93700427 on Build GRCh38
rs193214717 Chr.8: 93700247-93700247 on Build GRCh38
rs938456923 Chr.8: 93700623-93700623 on Build GRCh38
rs567564953 Chr.8: 93700355-93700355 on Build GRCh38
rs1001468178 Chr.8: 93700539-93700539 on Build GRCh38
rs867941541 Chr.8: 93700428-93700428 on Build GRCh38
rs539163540 Chr.8: 93700412-93700412 on Build GRCh38
rs776865567 Chr.8: 93700618-93700618 on Build GRCh38
rs900389230 Chr.8: 93700550-93700550 on Build GRCh38
rs1046966406 Chr.8: 93700608-93700608 on Build GRCh38
rs986513475 Chr.8: 93700561-93700561 on Build GRCh38
rs747744368 Chr.8: 93700384-93700384 on Build GRCh38
rs913404185 Chr.8: 93700519-93700519 on Build GRCh38
rs542257786 Chr.8: 93700594-93700594 on Build GRCh38
rs908850041 Chr.8: 93700478-93700478 on Build GRCh38
rs896493406 Chr.8: 93700514-93700514 on Build GRCh38
rs533739446 Chr.8: 93700297-93700297 on Build GRCh38
rs765351008 Chr.8: 93700629-93700629 on Build GRCh38
rs1057325743 Chr.8: 93700366-93700366 on Build GRCh38
rs886552610 Chr.8: 93700474-93700474 on Build GRCh38
rs975081918 Chr.8: 93700593-93700593 on Build GRCh38
rs1046875401 Chr.8: 93700473-93700473 on Build GRCh38
rs189067749 Chr.8: 93700568-93700568 on Build GRCh38
rs556108514 Chr.8: 93700604-93700604 on Build GRCh38
rs1030083555 Chr.8: 93700560-93700560 on Build GRCh38
rs997461791 Chr.8: 93700557-93700557 on Build GRCh38
rs963008249 Chr.8: 93700479-93700479 on Build GRCh38
rs35386317 Chr.8: 93700232-93700232 on Build GRCh38
rs769904121 Chr.8: 93700464-93700464 on Build GRCh38
rs986461172 Chr.8: 93700414-93700414 on Build GRCh38
rs935067928 Chr.8: 93700413-93700413 on Build GRCh38
rs1007284286 Chr.8: 93700532-93700532 on Build GRCh38
rs562547987 Chr.8: 93700653-93700653 on Build GRCh38
rs968306771 Chr.8: 93700538-93700538 on Build GRCh38
rs950448232 Chr.8: 93700471-93700471 on Build GRCh38
rs908373126 Chr.8: 93700615-93700615 on Build GRCh38
rs1031119617 Chr.8: 93700554-93700554 on Build GRCh38
rs878906720 Chr.8: 93700625-93700625 on Build GRCh38
rs891068860 Chr.8: 93700264-93700264 on Build GRCh38
rs185570402 Chr.8: 93700304-93700304 on Build GRCh38
rs942813204 Chr.8: 93700630-93700630 on Build GRCh38
rs1012218424 Chr.8: 93700524-93700524 on Build GRCh38
rs556549773 Chr.8: 93700392-93700392 on Build GRCh38
rs938957667 Chr.8: 93700484-93700484 on Build GRCh38
rs1004708543 Chr.8: 93700572-93700572 on Build GRCh38
rs572650937 Chr.8: 93700452-93700452 on Build GRCh38
rs898325203 Chr.8: 93700329-93700329 on Build GRCh38
rs913518467 Chr.8: 93700614-93700614 on Build GRCh38
rs972406782 Chr.8: 93700410-93700410 on Build GRCh38
rs762058635 Chr.8: 93700626-93700626 on Build GRCh38
rs536233636 Chr.8: 93700362-93700362 on Build GRCh38
rs571070150 Chr.8: 93700285-93700285 on Build GRCh38
rs1024322943 Chr.8: 93700530-93700530 on Build GRCh38
rs1039736428 Chr.8: 93700542-93700542 on Build GRCh38
rs886498584 Chr.8: 93700365-93700365 on Build GRCh38
rs562424475 Chr.8: 93700213-93700213 on Build GRCh38
rs1015966220 Chr.8: 93700573-93700573 on Build GRCh38
rs1017936563 Chr.8: 93700429-93700429 on Build GRCh38
rs541661034 Chr.8: 93700482-93700482 on Build GRCh38
rs368834447 Chr.8: 93700250-93700250 on Build GRCh38
rs768666867 Chr.8: 93700603-93700603 on Build GRCh38
rs997746771 Chr.8: 93700433-93700433 on Build GRCh38
rs547259313 Chr.8: 93700332-93700332 on Build GRCh38
rs1035057749 Chr.8: 93700498-93700498 on Build GRCh38
rs979834381 Chr.8: 93700480-93700480 on Build GRCh38
rs960982102 Chr.8: 93700385-93700385 on Build GRCh38
rs561550217 Chr.8: 93700525-93700525 on Build GRCh38
rs964681813 Chr.8: 93700430-93700430 on Build GRCh38
rs931156497 Chr.8: 93700454-93700454 on Build GRCh38
rs1040033667 Chr.8: 93700445-93700445 on Build GRCh38
rs1004963451 Chr.8: 93700487-93700487 on Build GRCh38

View Full Product Details