Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 74069943-74070442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTX
Entrez Gene ID
100302692
Gene Name
FTX transcript, XIST regulator (non-protein coding)
Gene Aliases
LINC00182, MIR374AHG, NCRNA00182
UniGene
Hs.349570
Gene Chromosome Location
Chr.X: 74028136-74293574 on Build GRCh38
Transcripts
NR_028379.1 NR_024582.1

Gene Symbol
JPX
Entrez Gene ID
554203
Gene Name
JPX transcript, XIST activator (non-protein coding)
Gene Aliases
DCBALD06, ENOX, LINC00183, NCRNA00183
UniGene
Hs.648316
Gene Chromosome Location
Chr.X: 73944324-74070384 on Build GRCh38
Transcripts
NR_028379.1 NR_024582.1

SNP Detail

SNP ID Chromosome Location
rs967002520 Chr.X: 74070361-74070386 on Build GRCh38
rs867317132 Chr.X: 74070412-74070412 on Build GRCh38
rs774559675 Chr.X: 74070232-74070232 on Build GRCh38
rs1022149543 Chr.X: 74070284-74070328 on Build GRCh38
rs751317744 Chr.X: 74070408-74070408 on Build GRCh38
rs749681207 Chr.X: 74070158-74070158 on Build GRCh38
rs59904201 Chr.X: 74070384-74070384 on Build GRCh38
rs948891657 Chr.X: 74070000-74070000 on Build GRCh38
rs750264975 Chr.X: 74070038-74070038 on Build GRCh38
rs1048937832 Chr.X: 74070335-74070335 on Build GRCh38
rs112560894 Chr.X: 74070338-74070338 on Build GRCh38
rs866194099 Chr.X: 74070165-74070165 on Build GRCh38
rs779455656 Chr.X: 74070339-74070340 on Build GRCh38
rs375593933 Chr.X: 74069978-74069978 on Build GRCh38
rs369877888 Chr.X: 74070381-74070381 on Build GRCh38
rs759748344 Chr.X: 74070237-74070237 on Build GRCh38
rs913429967 Chr.X: 74070238-74070238 on Build GRCh38
rs966307829 Chr.X: 74070386-74070386 on Build GRCh38
rs773547938 Chr.X: 74070142-74070142 on Build GRCh38
rs999303070 Chr.X: 74070415-74070415 on Build GRCh38
rs1008099357 Chr.X: 74070322-74070323 on Build GRCh38
rs72278733 Chr.X: 74070410-74070414 on Build GRCh38
rs765243039 Chr.X: 74070374-74070374 on Build GRCh38
rs201510084 Chr.X: 74070377-74070381 on Build GRCh38
rs368393998 Chr.X: 74070019-74070019 on Build GRCh38
rs746597490 Chr.X: 74070096-74070096 on Build GRCh38
rs779774299 Chr.X: 74070069-74070069 on Build GRCh38
rs780839136 Chr.X: 74070117-74070117 on Build GRCh38
rs1001456823 Chr.X: 74070030-74070030 on Build GRCh38
rs766086258 Chr.X: 74070400-74070400 on Build GRCh38
rs776269242 Chr.X: 74070381-74070386 on Build GRCh38
rs201934781 Chr.X: 74070383-74070384 on Build GRCh38
rs867405008 Chr.X: 74070157-74070157 on Build GRCh38
rs753837618 Chr.X: 74069973-74069973 on Build GRCh38
rs867173653 Chr.X: 74070112-74070112 on Build GRCh38
rs898512097 Chr.X: 74070209-74070209 on Build GRCh38
rs916114649 Chr.X: 74069965-74069965 on Build GRCh38
rs953205162 Chr.X: 74070202-74070202 on Build GRCh38
rs868659675 Chr.X: 74070197-74070197 on Build GRCh38
rs866263204 Chr.X: 74070385-74070385 on Build GRCh38
rs866740825 Chr.X: 74070193-74070193 on Build GRCh38
rs950155161 Chr.X: 74070297-74070297 on Build GRCh38
rs921609006 Chr.X: 74070169-74070169 on Build GRCh38
rs867976516 Chr.X: 74070210-74070210 on Build GRCh38
rs151057146 Chr.X: 74070057-74070057 on Build GRCh38
rs866511668 Chr.X: 74070137-74070137 on Build GRCh38
rs748711299 Chr.X: 74070125-74070125 on Build GRCh38
rs757749934 Chr.X: 74070379-74070380 on Build GRCh38
rs866095705 Chr.X: 74070198-74070198 on Build GRCh38
rs867186904 Chr.X: 74070236-74070236 on Build GRCh38
rs754657625 Chr.X: 74070410-74070410 on Build GRCh38
rs868534184 Chr.X: 74070213-74070213 on Build GRCh38
rs193106142 Chr.X: 74070109-74070109 on Build GRCh38
rs1046273203 Chr.X: 74070300-74070300 on Build GRCh38
rs868045998 Chr.X: 74070318-74070318 on Build GRCh38
rs926203543 Chr.X: 74070212-74070212 on Build GRCh38
rs760706992 Chr.X: 74070365-74070365 on Build GRCh38
rs867613094 Chr.X: 74070343-74070343 on Build GRCh38
rs749675441 Chr.X: 74070340-74070343 on Build GRCh38
rs767639755 Chr.X: 74070265-74070265 on Build GRCh38
rs868009376 Chr.X: 74070116-74070116 on Build GRCh38
rs1004782002 Chr.X: 74070376-74070376 on Build GRCh38
rs771142969 Chr.X: 74070208-74070208 on Build GRCh38
rs866390169 Chr.X: 74070187-74070187 on Build GRCh38
rs868786175 Chr.X: 74070185-74070185 on Build GRCh38
rs777972040 Chr.X: 74070343-74070344 on Build GRCh38
rs868520398 Chr.X: 74070233-74070233 on Build GRCh38
rs868188472 Chr.X: 74070108-74070108 on Build GRCh38
rs865797935 Chr.X: 74070224-74070224 on Build GRCh38
rs371642479 Chr.X: 74070391-74070391 on Build GRCh38
rs866834857 Chr.X: 74070168-74070168 on Build GRCh38
rs2182287 Chr.X: 74070327-74070327 on Build GRCh38
rs770301349 Chr.X: 74070126-74070126 on Build GRCh38
rs762814882 Chr.X: 74070389-74070389 on Build GRCh38
rs867182908 Chr.X: 74070152-74070152 on Build GRCh38
rs940093583 Chr.X: 74070135-74070135 on Build GRCh38
rs905825238 Chr.X: 74070303-74070303 on Build GRCh38

View Full Product Details