Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 74293254-74293761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTX
Entrez Gene ID
100302692
Gene Name
FTX transcript, XIST regulator (non-protein coding)
Gene Aliases
LINC00182, MIR374AHG, NCRNA00182
UniGene
Hs.349570
Gene Chromosome Location
Chr.X: 74028136-74293574 on Build GRCh38
Transcripts
NR_028379.1

SNP Detail

SNP ID Chromosome Location
rs174137 Chr.X: 74293640-74293640 on Build GRCh38
rs752350102 Chr.X: 74293603-74293603 on Build GRCh38
rs192433154 Chr.X: 74293505-74293505 on Build GRCh38
rs768730565 Chr.X: 74293402-74293402 on Build GRCh38
rs988310662 Chr.X: 74293497-74293497 on Build GRCh38
rs968398683 Chr.X: 74293311-74293311 on Build GRCh38
rs1031303034 Chr.X: 74293675-74293675 on Build GRCh38
rs951455565 Chr.X: 74293430-74293435 on Build GRCh38
rs754352300 Chr.X: 74293643-74293643 on Build GRCh38
rs1048358489 Chr.X: 74293578-74293578 on Build GRCh38
rs174136 Chr.X: 74293707-74293707 on Build GRCh38
rs931326374 Chr.X: 74293520-74293520 on Build GRCh38
rs866384704 Chr.X: 74293508-74293508 on Build GRCh38
rs766132993 Chr.X: 74293447-74293447 on Build GRCh38
rs757773600 Chr.X: 74293703-74293703 on Build GRCh38
rs904247384 Chr.X: 74293662-74293662 on Build GRCh38
rs943890358 Chr.X: 74293608-74293608 on Build GRCh38
rs776288891 Chr.X: 74293398-74293398 on Build GRCh38
rs190422282 Chr.X: 74293632-74293632 on Build GRCh38
rs935046032 Chr.X: 74293601-74293601 on Build GRCh38
rs956919756 Chr.X: 74293459-74293459 on Build GRCh38
rs918059380 Chr.X: 74293500-74293500 on Build GRCh38
rs1028489510 Chr.X: 74293303-74293303 on Build GRCh38
rs968992067 Chr.X: 74293540-74293540 on Build GRCh38
rs762912946 Chr.X: 74293430-74293437 on Build GRCh38
rs375192360 Chr.X: 74293449-74293449 on Build GRCh38
rs1008533890 Chr.X: 74293312-74293312 on Build GRCh38
rs899742015 Chr.X: 74293706-74293709 on Build GRCh38
rs924965693 Chr.X: 74293587-74293587 on Build GRCh38
rs757759768 Chr.X: 74293601-74293602 on Build GRCh38
rs773052363 Chr.X: 74293420-74293420 on Build GRCh38
rs996641439 Chr.X: 74293299-74293299 on Build GRCh38
rs1025645813 Chr.X: 74293735-74293735 on Build GRCh38
rs995467434 Chr.X: 74293428-74293429 on Build GRCh38
rs990403489 Chr.X: 74293623-74293623 on Build GRCh38
rs939904657 Chr.X: 74293597-74293597 on Build GRCh38
rs774181135 Chr.X: 74293503-74293503 on Build GRCh38
rs73624295 Chr.X: 74293607-74293607 on Build GRCh38
rs572582665 Chr.X: 74293606-74293606 on Build GRCh38
rs999883831 Chr.X: 74293663-74293663 on Build GRCh38
rs749388332 Chr.X: 74293430-74293431 on Build GRCh38
rs1018199100 Chr.X: 74293389-74293389 on Build GRCh38
rs767280520 Chr.X: 74293580-74293580 on Build GRCh38
rs965508708 Chr.X: 74293285-74293285 on Build GRCh38

View Full Product Details