Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149938673-149939192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf40B
Entrez Gene ID
541578
Gene Name
chromosome X open reading frame 40B
Gene Aliases
-
UniGene
Hs.545950
Gene Chromosome Location
Chr.X: 149924152-149938850 on Build GRCh38
Transcripts
XM_005274698.1 XM_005274699.1 XM_005274700.1 XM_005274701.2 XM_005274702.3 XM_006724826.3 XM_011531181.2 XM_017029585.1 XM_017029586.1 XM_017029587.1 XM_017029588.1 XM_017029590.1 XM_017029591.1 XM_017029592.1 XM_017029593.1 NR_027456.1

Gene Symbol
LINC00894
Entrez Gene ID
100272228
Gene Name
long intergenic non-protein coding RNA 894
Gene Aliases
-
UniGene
Hs.496916
Gene Chromosome Location
Chr.X: 149938548-150016787 on Build GRCh38
Transcripts
XM_005274698.1 XM_005274699.1 XM_005274700.1 XM_005274701.2 XM_005274702.3 XM_006724826.3 XM_011531181.2 XM_017029585.1 XM_017029586.1 XM_017029587.1 XM_017029588.1 XM_017029590.1 XM_017029591.1 XM_017029592.1 XM_017029593.1 NR_027456.1

SNP Detail

SNP ID Chromosome Location
rs782095264 Chr.X: 149938862-149938862 on Build GRCh38
rs367975574 Chr.X: 149938800-149938800 on Build GRCh38
rs181761861 Chr.X: 149938804-149938804 on Build GRCh38
rs371332620 Chr.X: 149938715-149938715 on Build GRCh38
rs5987388 Chr.X: 149938710-149938710 on Build GRCh38
rs782697501 Chr.X: 149938881-149938881 on Build GRCh38
rs782540610 Chr.X: 149938877-149938877 on Build GRCh38
rs112429616 Chr.X: 149938764-149938764 on Build GRCh38
rs781829976 Chr.X: 149938842-149938842 on Build GRCh38
rs370714133 Chr.X: 149938788-149938788 on Build GRCh38
rs782026383 Chr.X: 149938791-149938791 on Build GRCh38
rs186505299 Chr.X: 149938816-149938816 on Build GRCh38
rs782765625 Chr.X: 149938871-149938871 on Build GRCh38
rs781878418 Chr.X: 149938874-149938874 on Build GRCh38
rs112864494 Chr.X: 149938835-149938835 on Build GRCh38

View Full Product Details