Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39314536-39315080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
UniGene
Hs.369765
Gene Chromosome Location
Chr.8: 39314663-39417226 on Build GRCh38
Transcripts
NR_001448.1

SNP Detail

SNP ID Chromosome Location
rs1029732012 Chr.8: 39314936-39314936 on Build GRCh38
rs944755095 Chr.8: 39314715-39314715 on Build GRCh38
rs551237844 Chr.8: 39314694-39314694 on Build GRCh38
rs979396079 Chr.8: 39314622-39314622 on Build GRCh38
rs1000343810 Chr.8: 39314895-39314895 on Build GRCh38
rs1041638922 Chr.8: 39314590-39314590 on Build GRCh38
rs80279060 Chr.8: 39315036-39315036 on Build GRCh38
rs533242896 Chr.8: 39314657-39314657 on Build GRCh38
rs1010135021 Chr.8: 39314589-39314589 on Build GRCh38
rs765032037 Chr.8: 39314724-39314724 on Build GRCh38
rs1056106104 Chr.8: 39314812-39314812 on Build GRCh38
rs945682754 Chr.8: 39314769-39314769 on Build GRCh38
rs73610518 Chr.8: 39314930-39314930 on Build GRCh38
rs192009007 Chr.8: 39315040-39315040 on Build GRCh38
rs951362423 Chr.8: 39314700-39314700 on Build GRCh38
rs1048989255 Chr.8: 39314751-39314751 on Build GRCh38
rs762878484 Chr.8: 39314839-39314839 on Build GRCh38
rs896153905 Chr.8: 39314855-39314855 on Build GRCh38
rs549244005 Chr.8: 39314736-39314736 on Build GRCh38
rs1056818668 Chr.8: 39314760-39314760 on Build GRCh38
rs768416607 Chr.8: 39314619-39314619 on Build GRCh38
rs930468399 Chr.8: 39315021-39315021 on Build GRCh38
rs904440165 Chr.8: 39314795-39314795 on Build GRCh38
rs868069244 Chr.8: 39314640-39314640 on Build GRCh38
rs960478232 Chr.8: 39314592-39314592 on Build GRCh38
rs1039911799 Chr.8: 39314877-39314877 on Build GRCh38
rs1004542985 Chr.8: 39314785-39314785 on Build GRCh38
rs577018092 Chr.8: 39315002-39315002 on Build GRCh38
rs75164723 Chr.8: 39314658-39314658 on Build GRCh38
rs528528172 Chr.8: 39314609-39314609 on Build GRCh38
rs760377548 Chr.8: 39314579-39314579 on Build GRCh38
rs1034764813 Chr.8: 39314587-39314587 on Build GRCh38
rs930521323 Chr.8: 39314742-39314742 on Build GRCh38
rs112096846 Chr.8: 39315052-39315052 on Build GRCh38
rs962888076 Chr.8: 39314741-39314741 on Build GRCh38
rs865798336 Chr.8: 39314575-39314575 on Build GRCh38
rs181560673 Chr.8: 39314586-39314586 on Build GRCh38
rs900342639 Chr.8: 39314561-39314561 on Build GRCh38
rs533697232 Chr.8: 39314782-39314782 on Build GRCh38
rs548350609 Chr.8: 39314941-39314941 on Build GRCh38
rs970618888 Chr.8: 39314892-39314892 on Build GRCh38
rs528232927 Chr.8: 39314954-39314954 on Build GRCh38
rs1018401093 Chr.8: 39314639-39314639 on Build GRCh38
rs886395372 Chr.8: 39314761-39314761 on Build GRCh38
rs571019512 Chr.8: 39314714-39314714 on Build GRCh38
rs911969402 Chr.8: 39314682-39314682 on Build GRCh38
rs750391863 Chr.8: 39314811-39314811 on Build GRCh38
rs921738723 Chr.8: 39314740-39314740 on Build GRCh38
rs983022172 Chr.8: 39314705-39314705 on Build GRCh38
rs1012384546 Chr.8: 39314856-39314856 on Build GRCh38
rs926238561 Chr.8: 39314636-39314636 on Build GRCh38
rs146140254 Chr.8: 39314955-39314955 on Build GRCh38
rs1017142951 Chr.8: 39314980-39314980 on Build GRCh38
rs559702453 Chr.8: 39314604-39314604 on Build GRCh38
rs560409193 Chr.8: 39314961-39314961 on Build GRCh38
rs187219847 Chr.8: 39314882-39314882 on Build GRCh38
rs922062307 Chr.8: 39315020-39315020 on Build GRCh38
rs990590010 Chr.8: 39314603-39314603 on Build GRCh38
rs138096477 Chr.8: 39314676-39314676 on Build GRCh38
rs368592423 Chr.8: 39314564-39314564 on Build GRCh38
rs559566502 Chr.8: 39314570-39314570 on Build GRCh38
rs980976548 Chr.8: 39315003-39315003 on Build GRCh38
rs751820989 Chr.8: 39314904-39314904 on Build GRCh38
rs962898969 Chr.8: 39314993-39314993 on Build GRCh38
rs1039122424 Chr.8: 39314734-39314734 on Build GRCh38
rs937025441 Chr.8: 39314559-39314559 on Build GRCh38
rs1030050319 Chr.8: 39314663-39314663 on Build GRCh38
rs886092703 Chr.8: 39314585-39314585 on Build GRCh38
rs551840445 Chr.8: 39314881-39314881 on Build GRCh38
rs1039313599 Chr.8: 39314558-39314558 on Build GRCh38

View Full Product Details