Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39323688-39324241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
UniGene
Hs.369765
Gene Chromosome Location
Chr.8: 39314663-39417226 on Build GRCh38
Transcripts
NR_001448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834707 c.35C>T p.P12L Chr.8: 39324209-39324209 on Build GRCh38
5713160 c.46_47insT p.S17fs*9 Chr.8: 39324220-39324221 on Build GRCh38
5427895 c.37A>T p.M13L Chr.8: 39324211-39324211 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs9969447 Chr.8: 39323893-39323893 on Build GRCh38
rs932005434 Chr.8: 39323792-39323792 on Build GRCh38
rs896982438 Chr.8: 39323923-39323923 on Build GRCh38
rs976324933 Chr.8: 39323911-39323911 on Build GRCh38
rs559369297 Chr.8: 39324045-39324045 on Build GRCh38
rs1025409963 Chr.8: 39323975-39323975 on Build GRCh38
rs77143648 Chr.8: 39323749-39323749 on Build GRCh38
rs923441900 Chr.8: 39324070-39324070 on Build GRCh38
rs919379864 Chr.8: 39324116-39324116 on Build GRCh38
rs535558636 Chr.8: 39323882-39323882 on Build GRCh38
rs34525881 Chr.8: 39323737-39323737 on Build GRCh38
rs980442860 Chr.8: 39323872-39323872 on Build GRCh38
rs1054175466 Chr.8: 39323845-39323845 on Build GRCh38
rs949472422 Chr.8: 39324151-39324151 on Build GRCh38
rs932419165 Chr.8: 39324019-39324019 on Build GRCh38
rs1002082720 Chr.8: 39323989-39323989 on Build GRCh38
rs191121116 Chr.8: 39323818-39323818 on Build GRCh38
rs112324096 Chr.8: 39323965-39323965 on Build GRCh38
rs751496476 Chr.8: 39323795-39323795 on Build GRCh38
rs918200191 Chr.8: 39323770-39323770 on Build GRCh38
rs1013405896 Chr.8: 39323982-39323982 on Build GRCh38
rs188436445 Chr.8: 39323954-39323954 on Build GRCh38
rs987929008 Chr.8: 39323890-39323890 on Build GRCh38
rs112994586 Chr.8: 39323889-39323889 on Build GRCh38
rs566577147 Chr.8: 39323878-39323878 on Build GRCh38
rs573140074 Chr.8: 39324152-39324152 on Build GRCh38
rs1015142545 Chr.8: 39323825-39323825 on Build GRCh38
rs530523488 Chr.8: 39324090-39324090 on Build GRCh38
rs905077149 Chr.8: 39323988-39323988 on Build GRCh38
rs868133935 Chr.8: 39323974-39323974 on Build GRCh38
rs185048320 Chr.8: 39324086-39324086 on Build GRCh38
rs398086518 Chr.8: 39323749-39323749 on Build GRCh38
rs1009720149 Chr.8: 39324051-39324051 on Build GRCh38
rs569280448 Chr.8: 39324072-39324072 on Build GRCh38
rs905045783 Chr.8: 39324179-39324179 on Build GRCh38
rs971405884 Chr.8: 39323772-39323772 on Build GRCh38
rs867807334 Chr.8: 39323856-39323856 on Build GRCh38
rs35898753 Chr.8: 39324114-39324115 on Build GRCh38
rs1032392887 Chr.8: 39323998-39323998 on Build GRCh38
rs13263225 Chr.8: 39324107-39324107 on Build GRCh38
rs946227680 Chr.8: 39323909-39323909 on Build GRCh38
rs373331411 Chr.8: 39324212-39324213 on Build GRCh38
rs192636923 Chr.8: 39324083-39324083 on Build GRCh38
rs796229871 Chr.8: 39323750-39323750 on Build GRCh38
rs912220055 Chr.8: 39324040-39324040 on Build GRCh38
rs1020737693 Chr.8: 39324056-39324056 on Build GRCh38
rs1050824048 Chr.8: 39323952-39323952 on Build GRCh38
rs932224818 Chr.8: 39323922-39323922 on Build GRCh38
rs772692346 Chr.8: 39323713-39323713 on Build GRCh38
rs867594697 Chr.8: 39323964-39323964 on Build GRCh38
rs570351173 Chr.8: 39324099-39324099 on Build GRCh38
rs9969625 Chr.8: 39323976-39323976 on Build GRCh38
rs550708580 Chr.8: 39324097-39324097 on Build GRCh38
rs150270951 Chr.8: 39324099-39324100 on Build GRCh38
rs971355818 Chr.8: 39323883-39323883 on Build GRCh38
rs551522955 Chr.8: 39324156-39324156 on Build GRCh38
rs976210358 Chr.8: 39324066-39324066 on Build GRCh38
rs1046294209 Chr.8: 39323986-39323986 on Build GRCh38
rs894952722 Chr.8: 39324085-39324085 on Build GRCh38
rs544765386 Chr.8: 39323940-39323940 on Build GRCh38
rs923558084 Chr.8: 39323912-39323912 on Build GRCh38
rs957752484 Chr.8: 39324044-39324044 on Build GRCh38
rs557465217 Chr.8: 39324058-39324058 on Build GRCh38
rs765130999 Chr.8: 39324091-39324091 on Build GRCh38
rs199577980 Chr.8: 39323723-39323724 on Build GRCh38
rs985144632 Chr.8: 39324028-39324028 on Build GRCh38
rs969017421 Chr.8: 39323868-39323868 on Build GRCh38
rs1013437038 Chr.8: 39323851-39323851 on Build GRCh38
rs763628620 Chr.8: 39323835-39323835 on Build GRCh38
rs991980311 Chr.8: 39323871-39323871 on Build GRCh38
rs989666394 Chr.8: 39324209-39324209 on Build GRCh38
rs760267925 Chr.8: 39323819-39323819 on Build GRCh38
rs986417255 Chr.8: 39324074-39324074 on Build GRCh38
rs1023070631 Chr.8: 39323874-39323874 on Build GRCh38
rs546521073 Chr.8: 39324178-39324178 on Build GRCh38
rs371435477 Chr.8: 39324011-39324011 on Build GRCh38
rs571730173 Chr.8: 39323960-39323960 on Build GRCh38

View Full Product Details