Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34835968-34836524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM205BP
Entrez Gene ID
389715
Gene Name
transmembrane protein C9orf144B pseudogene
Gene Aliases
C9orf144, FAM205B
UniGene
Hs.713965
Gene Chromosome Location
Chr.9: 34830267-34838586 on Build GRCh38
Transcripts
NR_024481.1 XM_011517827.1

Gene Symbol
PHF24
Entrez Gene ID
23349
Gene Name
PHD finger protein 24
Gene Aliases
KIAA1045
UniGene
Hs.7989
Gene Chromosome Location
Chr.9: 34810040-34982544 on Build GRCh38
Transcripts
NR_024481.1 XM_011517827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5998663 c.283G>A p.E95K Chr.9: 34836017-34836017 on Build GRCh38
5043705 c.252-3C>T p.? Chr.9: 34836051-34836051 on Build GRCh38
7239149 c.301C>A p.P101T Chr.9: 34835999-34835999 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757540364 Chr.9: 34836041-34836042 on Build GRCh38
rs755986233 Chr.9: 34835998-34835998 on Build GRCh38
rs1046731547 Chr.9: 34836285-34836285 on Build GRCh38
rs1008026944 Chr.9: 34836405-34836405 on Build GRCh38
rs1022284953 Chr.9: 34836136-34836136 on Build GRCh38
rs557661353 Chr.9: 34836091-34836091 on Build GRCh38
rs922438181 Chr.9: 34836460-34836460 on Build GRCh38
rs190180670 Chr.9: 34836006-34836006 on Build GRCh38
rs769739881 Chr.9: 34836419-34836419 on Build GRCh38
rs572706079 Chr.9: 34836313-34836313 on Build GRCh38
rs538363727 Chr.9: 34836286-34836286 on Build GRCh38
rs976606844 Chr.9: 34836458-34836458 on Build GRCh38
rs181016742 Chr.9: 34836033-34836033 on Build GRCh38
rs576485256 Chr.9: 34836224-34836224 on Build GRCh38
rs757702967 Chr.9: 34836249-34836249 on Build GRCh38
rs760137430 Chr.9: 34836085-34836085 on Build GRCh38
rs1017674749 Chr.9: 34836416-34836416 on Build GRCh38
rs749043515 Chr.9: 34836017-34836017 on Build GRCh38
rs200297071 Chr.9: 34836350-34836350 on Build GRCh38
rs966549659 Chr.9: 34836440-34836440 on Build GRCh38
rs111514649 Chr.9: 34836270-34836270 on Build GRCh38
rs746158493 Chr.9: 34836319-34836319 on Build GRCh38
rs986232443 Chr.9: 34836165-34836165 on Build GRCh38
rs909522058 Chr.9: 34836500-34836500 on Build GRCh38
rs535247720 Chr.9: 34836020-34836020 on Build GRCh38
rs929913094 Chr.9: 34836278-34836278 on Build GRCh38
rs922493746 Chr.9: 34836271-34836271 on Build GRCh38
rs745808346 Chr.9: 34835992-34835992 on Build GRCh38
rs142600471 Chr.9: 34836046-34836046 on Build GRCh38
rs1044835657 Chr.9: 34836305-34836305 on Build GRCh38
rs942561789 Chr.9: 34836173-34836173 on Build GRCh38
rs956977044 Chr.9: 34836117-34836117 on Build GRCh38
rs751916723 Chr.9: 34836097-34836097 on Build GRCh38
rs867673432 Chr.9: 34836007-34836007 on Build GRCh38
rs577541428 Chr.9: 34836118-34836118 on Build GRCh38
rs747752395 Chr.9: 34836022-34836022 on Build GRCh38
rs1022211337 Chr.9: 34836386-34836386 on Build GRCh38
rs549180463 Chr.9: 34836021-34836021 on Build GRCh38
rs779516269 Chr.9: 34836274-34836274 on Build GRCh38
rs1046804524 Chr.9: 34836018-34836018 on Build GRCh38
rs555917051 Chr.9: 34836079-34836079 on Build GRCh38
rs541741871 Chr.9: 34836321-34836321 on Build GRCh38
rs111929144 Chr.9: 34836011-34836011 on Build GRCh38
rs951238994 Chr.9: 34836472-34836472 on Build GRCh38
rs554625573 Chr.9: 34836301-34836301 on Build GRCh38
rs911021965 Chr.9: 34836167-34836167 on Build GRCh38
rs771482295 Chr.9: 34836025-34836025 on Build GRCh38
rs77147479 Chr.9: 34836111-34836111 on Build GRCh38
rs1003697928 Chr.9: 34836320-34836320 on Build GRCh38
rs892710126 Chr.9: 34836333-34836333 on Build GRCh38
rs185936504 Chr.9: 34836121-34836121 on Build GRCh38
rs968094169 Chr.9: 34836398-34836398 on Build GRCh38
rs988098141 Chr.9: 34836123-34836123 on Build GRCh38
rs950048 Chr.9: 34836499-34836499 on Build GRCh38
rs151287943 Chr.9: 34836156-34836156 on Build GRCh38
rs905379170 Chr.9: 34836036-34836036 on Build GRCh38
rs1009780849 Chr.9: 34836377-34836377 on Build GRCh38

View Full Product Details