Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35444277-35444841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
UniGene
Hs.575661
Gene Chromosome Location
Chr.9: 35406755-35483029 on Build GRCh38
Transcripts
NR_003581.2 NR_003582.1 NR_110320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4626448 c.54G>A p.M18I Chr.9: 35444457-35444457 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs947118362 Chr.9: 35444476-35444476 on Build GRCh38
rs754593689 Chr.9: 35444647-35444647 on Build GRCh38
rs562741795 Chr.9: 35444360-35444360 on Build GRCh38
rs10972478 Chr.9: 35444673-35444673 on Build GRCh38
rs934519508 Chr.9: 35444434-35444434 on Build GRCh38
rs781622280 Chr.9: 35444319-35444319 on Build GRCh38
rs868587870 Chr.9: 35444760-35444760 on Build GRCh38
rs7864635 Chr.9: 35444485-35444485 on Build GRCh38
rs777445747 Chr.9: 35444593-35444593 on Build GRCh38
rs975992443 Chr.9: 35444400-35444400 on Build GRCh38
rs969004840 Chr.9: 35444807-35444807 on Build GRCh38
rs1057382141 Chr.9: 35444502-35444502 on Build GRCh38
rs988869879 Chr.9: 35444333-35444333 on Build GRCh38
rs10972475 Chr.9: 35444334-35444334 on Build GRCh38
rs886363794 Chr.9: 35444602-35444602 on Build GRCh38
rs531942974 Chr.9: 35444776-35444776 on Build GRCh38
rs964656805 Chr.9: 35444369-35444369 on Build GRCh38
rs978955704 Chr.9: 35444423-35444423 on Build GRCh38
rs959722810 Chr.9: 35444323-35444323 on Build GRCh38
rs888564368 Chr.9: 35444661-35444661 on Build GRCh38
rs531924263 Chr.9: 35444380-35444380 on Build GRCh38
rs1006519436 Chr.9: 35444614-35444614 on Build GRCh38
rs989020383 Chr.9: 35444436-35444436 on Build GRCh38
rs922661077 Chr.9: 35444532-35444532 on Build GRCh38
rs10972477 Chr.9: 35444359-35444359 on Build GRCh38
rs10972476 Chr.9: 35444354-35444354 on Build GRCh38
rs41277045 Chr.9: 35444404-35444404 on Build GRCh38
rs905744684 Chr.9: 35444639-35444639 on Build GRCh38
rs1049017066 Chr.9: 35444644-35444644 on Build GRCh38
rs570563230 Chr.9: 35444563-35444563 on Build GRCh38
rs549645049 Chr.9: 35444489-35444489 on Build GRCh38
rs879045130 Chr.9: 35444616-35444616 on Build GRCh38
rs959752269 Chr.9: 35444772-35444772 on Build GRCh38
rs1037616435 Chr.9: 35444638-35444638 on Build GRCh38
rs1018270969 Chr.9: 35444347-35444347 on Build GRCh38
rs930009011 Chr.9: 35444537-35444537 on Build GRCh38
rs994360454 Chr.9: 35444592-35444592 on Build GRCh38
rs753629467 Chr.9: 35444512-35444512 on Build GRCh38
rs540493995 Chr.9: 35444486-35444486 on Build GRCh38
rs979455010 Chr.9: 35444809-35444809 on Build GRCh38
rs567994434 Chr.9: 35444450-35444450 on Build GRCh38
rs914342986 Chr.9: 35444468-35444468 on Build GRCh38
rs1020331449 Chr.9: 35444785-35444785 on Build GRCh38
rs911357840 Chr.9: 35444431-35444431 on Build GRCh38
rs1006967557 Chr.9: 35444668-35444668 on Build GRCh38

View Full Product Details