Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35449353-35449934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
UniGene
Hs.575661
Gene Chromosome Location
Chr.9: 35406755-35483029 on Build GRCh38
Transcripts
NR_003581.2 NR_003582.1 NR_110320.1

SNP Detail

SNP ID Chromosome Location
rs747277007 Chr.9: 35449885-35449885 on Build GRCh38
rs749863601 Chr.9: 35449517-35449517 on Build GRCh38
rs555603198 Chr.9: 35449649-35449649 on Build GRCh38
rs897554583 Chr.9: 35449833-35449833 on Build GRCh38
rs755514529 Chr.9: 35449524-35449524 on Build GRCh38
rs1045338561 Chr.9: 35449801-35449801 on Build GRCh38
rs758901011 Chr.9: 35449738-35449738 on Build GRCh38
rs986731856 Chr.9: 35449557-35449557 on Build GRCh38
rs878989186 Chr.9: 35449869-35449869 on Build GRCh38
rs549149277 Chr.9: 35449675-35449675 on Build GRCh38
rs80220865 Chr.9: 35449402-35449402 on Build GRCh38
rs567870683 Chr.9: 35449910-35449910 on Build GRCh38
rs536091305 Chr.9: 35449727-35449727 on Build GRCh38
rs117528895 Chr.9: 35449824-35449824 on Build GRCh38
rs1054363739 Chr.9: 35449720-35449720 on Build GRCh38
rs961711283 Chr.9: 35449503-35449503 on Build GRCh38
rs529959739 Chr.9: 35449637-35449637 on Build GRCh38
rs753245080 Chr.9: 35449690-35449690 on Build GRCh38
rs916514217 Chr.9: 35449527-35449527 on Build GRCh38
rs1014641495 Chr.9: 35449500-35449500 on Build GRCh38
rs150067444 Chr.9: 35449571-35449571 on Build GRCh38
rs994474132 Chr.9: 35449516-35449516 on Build GRCh38
rs4557787 Chr.9: 35449644-35449644 on Build GRCh38
rs898019541 Chr.9: 35449844-35449844 on Build GRCh38
rs61744965 Chr.9: 35449898-35449898 on Build GRCh38
rs182585756 Chr.9: 35449827-35449827 on Build GRCh38
rs1052205497 Chr.9: 35449683-35449683 on Build GRCh38
rs1019079165 Chr.9: 35449424-35449424 on Build GRCh38
rs969291107 Chr.9: 35449547-35449547 on Build GRCh38
rs924664833 Chr.9: 35449670-35449670 on Build GRCh38
rs138510617 Chr.9: 35449802-35449802 on Build GRCh38
rs1050049662 Chr.9: 35449776-35449776 on Build GRCh38
rs962517551 Chr.9: 35449447-35449447 on Build GRCh38
rs4461983 Chr.9: 35449632-35449632 on Build GRCh38
rs993638230 Chr.9: 35449498-35449498 on Build GRCh38
rs1003290803 Chr.9: 35449405-35449405 on Build GRCh38
rs112548347 Chr.9: 35449843-35449843 on Build GRCh38
rs1045938213 Chr.9: 35449742-35449755 on Build GRCh38
rs537661806 Chr.9: 35449834-35449834 on Build GRCh38
rs966567490 Chr.9: 35449571-35449571 on Build GRCh38
rs939145939 Chr.9: 35449820-35449820 on Build GRCh38
rs115942390 Chr.9: 35449404-35449404 on Build GRCh38
rs779505378 Chr.9: 35449546-35449546 on Build GRCh38
rs912055384 Chr.9: 35449570-35449570 on Build GRCh38
rs565895175 Chr.9: 35449578-35449579 on Build GRCh38
rs977537099 Chr.9: 35449624-35449624 on Build GRCh38

View Full Product Details