Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35450207-35450755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
UniGene
Hs.575661
Gene Chromosome Location
Chr.9: 35406755-35483029 on Build GRCh38
Transcripts
NR_003581.2 NR_003582.1 NR_110320.1

SNP Detail

SNP ID Chromosome Location
rs1022171758 Chr.9: 35450397-35450398 on Build GRCh38
rs550942863 Chr.9: 35450356-35450356 on Build GRCh38
rs78366996 Chr.9: 35450247-35450247 on Build GRCh38
rs909424449 Chr.9: 35450474-35450474 on Build GRCh38
rs566797150 Chr.9: 35450508-35450508 on Build GRCh38
rs773971028 Chr.9: 35450392-35450392 on Build GRCh38
rs1053496745 Chr.9: 35450660-35450660 on Build GRCh38
rs146952089 Chr.9: 35450443-35450445 on Build GRCh38
rs1044514304 Chr.9: 35450535-35450535 on Build GRCh38
rs1040427846 Chr.9: 35450673-35450673 on Build GRCh38
rs930425035 Chr.9: 35450594-35450594 on Build GRCh38
rs538251250 Chr.9: 35450564-35450564 on Build GRCh38
rs536580523 Chr.9: 35450380-35450380 on Build GRCh38
rs944403081 Chr.9: 35450422-35450423 on Build GRCh38
rs929524030 Chr.9: 35450466-35450466 on Build GRCh38
rs971924872 Chr.9: 35450548-35450548 on Build GRCh38
rs56234627 Chr.9: 35450425-35450425 on Build GRCh38
rs981823730 Chr.9: 35450448-35450448 on Build GRCh38
rs1019207413 Chr.9: 35450279-35450279 on Build GRCh38
rs776344449 Chr.9: 35450329-35450329 on Build GRCh38
rs1048850666 Chr.9: 35450623-35450623 on Build GRCh38
rs907223542 Chr.9: 35450284-35450284 on Build GRCh38
rs367796177 Chr.9: 35450705-35450705 on Build GRCh38
rs530983133 Chr.9: 35450347-35450347 on Build GRCh38
rs767015482 Chr.9: 35450557-35450557 on Build GRCh38
rs891793601 Chr.9: 35450668-35450668 on Build GRCh38
rs188749247 Chr.9: 35450624-35450624 on Build GRCh38
rs943068684 Chr.9: 35450645-35450645 on Build GRCh38
rs999005346 Chr.9: 35450321-35450321 on Build GRCh38
rs545085350 Chr.9: 35450637-35450637 on Build GRCh38
rs547771520 Chr.9: 35450413-35450414 on Build GRCh38
rs138503109 Chr.9: 35450713-35450713 on Build GRCh38
rs183902540 Chr.9: 35450298-35450298 on Build GRCh38
rs927926369 Chr.9: 35450459-35450459 on Build GRCh38
rs565379433 Chr.9: 35450337-35450337 on Build GRCh38
rs976129494 Chr.9: 35450436-35450436 on Build GRCh38
rs1023467824 Chr.9: 35450253-35450253 on Build GRCh38
rs1049291506 Chr.9: 35450467-35450467 on Build GRCh38
rs765984411 Chr.9: 35450684-35450684 on Build GRCh38
rs985924766 Chr.9: 35450412-35450412 on Build GRCh38
rs970095918 Chr.9: 35450447-35450447 on Build GRCh38
rs145876528 Chr.9: 35450362-35450362 on Build GRCh38
rs547290021 Chr.9: 35450400-35450400 on Build GRCh38

View Full Product Details