Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35450626-35451172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
UniGene
Hs.575661
Gene Chromosome Location
Chr.9: 35406755-35483029 on Build GRCh38
Transcripts
NR_003581.2 NR_003582.1 NR_110320.1

SNP Detail

SNP ID Chromosome Location
rs1007393737 Chr.9: 35451132-35451132 on Build GRCh38
rs903750534 Chr.9: 35451106-35451107 on Build GRCh38
rs1011670025 Chr.9: 35450777-35450777 on Build GRCh38
rs150656776 Chr.9: 35450893-35450893 on Build GRCh38
rs1023065848 Chr.9: 35450786-35450786 on Build GRCh38
rs1053496745 Chr.9: 35450660-35450660 on Build GRCh38
rs778312663 Chr.9: 35451122-35451122 on Build GRCh38
rs950383038 Chr.9: 35450932-35450932 on Build GRCh38
rs193302020 Chr.9: 35450894-35450894 on Build GRCh38
rs141941471 Chr.9: 35450791-35450791 on Build GRCh38
rs1040427846 Chr.9: 35450673-35450673 on Build GRCh38
rs910326153 Chr.9: 35451027-35451027 on Build GRCh38
rs75666643 Chr.9: 35451104-35451104 on Build GRCh38
rs771209031 Chr.9: 35450826-35450826 on Build GRCh38
rs951873351 Chr.9: 35451008-35451008 on Build GRCh38
rs758923642 Chr.9: 35450934-35450934 on Build GRCh38
rs934290390 Chr.9: 35451135-35451135 on Build GRCh38
rs943101697 Chr.9: 35451084-35451084 on Build GRCh38
rs966145067 Chr.9: 35450861-35450861 on Build GRCh38
rs530939238 Chr.9: 35450975-35450975 on Build GRCh38
rs367796177 Chr.9: 35450705-35450705 on Build GRCh38
rs1053311200 Chr.9: 35451140-35451140 on Build GRCh38
rs891828512 Chr.9: 35451081-35451082 on Build GRCh38
rs764748269 Chr.9: 35450821-35450821 on Build GRCh38
rs891793601 Chr.9: 35450668-35450668 on Build GRCh38
rs938799674 Chr.9: 35451011-35451011 on Build GRCh38
rs574559056 Chr.9: 35450776-35450776 on Build GRCh38
rs561193749 Chr.9: 35451080-35451080 on Build GRCh38
rs551040418 Chr.9: 35451026-35451026 on Build GRCh38
rs1043678492 Chr.9: 35451105-35451105 on Build GRCh38
rs1002903980 Chr.9: 35450874-35450874 on Build GRCh38
rs971977097 Chr.9: 35450948-35450948 on Build GRCh38
rs186920137 Chr.9: 35450954-35450954 on Build GRCh38
rs138503109 Chr.9: 35450713-35450713 on Build GRCh38
rs1053258404 Chr.9: 35450732-35450732 on Build GRCh38
rs184218849 Chr.9: 35450909-35450909 on Build GRCh38
rs1053207007 Chr.9: 35451019-35451019 on Build GRCh38
rs909454156 Chr.9: 35450947-35450947 on Build GRCh38
rs922940115 Chr.9: 35451130-35451130 on Build GRCh38
rs1017819604 Chr.9: 35451145-35451145 on Build GRCh38
rs1017374362 Chr.9: 35450831-35450831 on Build GRCh38
rs765984411 Chr.9: 35450684-35450684 on Build GRCh38
rs918954470 Chr.9: 35450906-35450906 on Build GRCh38

View Full Product Details