Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35478036-35478594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
UniGene
Hs.575661
Gene Chromosome Location
Chr.9: 35406755-35483029 on Build GRCh38
Transcripts
NR_003581.2

SNP Detail

SNP ID Chromosome Location
rs77879542 Chr.9: 35478480-35478480 on Build GRCh38
rs1002320596 Chr.9: 35478362-35478362 on Build GRCh38
rs887005086 Chr.9: 35478347-35478347 on Build GRCh38
rs901438587 Chr.9: 35478073-35478073 on Build GRCh38
rs956413706 Chr.9: 35478116-35478116 on Build GRCh38
rs969181465 Chr.9: 35478484-35478484 on Build GRCh38
rs926247592 Chr.9: 35478145-35478145 on Build GRCh38
rs35402033 Chr.9: 35478134-35478134 on Build GRCh38
rs145023916 Chr.9: 35478557-35478557 on Build GRCh38
rs914144765 Chr.9: 35478157-35478157 on Build GRCh38
rs1048488194 Chr.9: 35478299-35478299 on Build GRCh38
rs991806427 Chr.9: 35478421-35478421 on Build GRCh38
rs888556408 Chr.9: 35478348-35478348 on Build GRCh38
rs998391784 Chr.9: 35478398-35478398 on Build GRCh38
rs927844383 Chr.9: 35478213-35478213 on Build GRCh38
rs892119280 Chr.9: 35478474-35478474 on Build GRCh38
rs7023494 Chr.9: 35478260-35478260 on Build GRCh38
rs1030772771 Chr.9: 35478123-35478123 on Build GRCh38
rs933934451 Chr.9: 35478304-35478304 on Build GRCh38
rs752030411 Chr.9: 35478245-35478245 on Build GRCh38
rs1039038108 Chr.9: 35478275-35478275 on Build GRCh38
rs960562616 Chr.9: 35478532-35478532 on Build GRCh38
rs901381663 Chr.9: 35478360-35478360 on Build GRCh38
rs939292009 Chr.9: 35478218-35478218 on Build GRCh38
rs962773151 Chr.9: 35478087-35478087 on Build GRCh38
rs191660962 Chr.9: 35478193-35478193 on Build GRCh38
rs1031249607 Chr.9: 35478433-35478433 on Build GRCh38
rs548005983 Chr.9: 35478366-35478366 on Build GRCh38
rs980980578 Chr.9: 35478499-35478499 on Build GRCh38
rs971960606 Chr.9: 35478549-35478549 on Build GRCh38
rs960990373 Chr.9: 35478154-35478154 on Build GRCh38
rs960682130 Chr.9: 35478396-35478396 on Build GRCh38
rs1030044716 Chr.9: 35478143-35478143 on Build GRCh38
rs1050992449 Chr.9: 35478306-35478306 on Build GRCh38
rs956401673 Chr.9: 35478129-35478129 on Build GRCh38
rs945550926 Chr.9: 35478158-35478158 on Build GRCh38
rs969609675 Chr.9: 35478167-35478167 on Build GRCh38
rs997772600 Chr.9: 35478100-35478100 on Build GRCh38
rs568418568 Chr.9: 35478357-35478357 on Build GRCh38
rs1004040084 Chr.9: 35478355-35478355 on Build GRCh38
rs918582055 Chr.9: 35478249-35478249 on Build GRCh38
rs943487213 Chr.9: 35478528-35478528 on Build GRCh38
rs545830106 Chr.9: 35478155-35478155 on Build GRCh38
rs556861223 Chr.9: 35478514-35478514 on Build GRCh38
rs933345503 Chr.9: 35478563-35478563 on Build GRCh38
rs371215947 Chr.9: 35478141-35478141 on Build GRCh38
rs548663618 Chr.9: 35478356-35478356 on Build GRCh38
rs780106610 Chr.9: 35478538-35478538 on Build GRCh38
rs571189501 Chr.9: 35478469-35478469 on Build GRCh38
rs1010552977 Chr.9: 35478476-35478476 on Build GRCh38
rs1021256951 Chr.9: 35478456-35478456 on Build GRCh38
rs1030626772 Chr.9: 35478112-35478112 on Build GRCh38
rs527924380 Chr.9: 35478363-35478363 on Build GRCh38
rs980650978 Chr.9: 35478176-35478176 on Build GRCh38

View Full Product Details