Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97287559-97288114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100499484
Entrez Gene ID
100499484
Gene Name
SUGT1-1300002K09Rik pseudogene
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97297314 on Build GRCh38
Transcripts
NR_036526.1 NR_036527.1 NR_036528.1 NR_036529.1

Gene Symbol
LOC100499484-C9ORF174
Entrez Gene ID
57653
Gene Name
LOC100499484-C9orf174 readthrough
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97377295 on Build GRCh38
Transcripts
NR_036526.1 NR_036527.1 NR_036528.1 NR_036529.1

SNP Detail

SNP ID Chromosome Location
rs956783410 Chr.9: 97288077-97288077 on Build GRCh38
rs762647418 Chr.9: 97287990-97287990 on Build GRCh38
rs896863396 Chr.9: 97287886-97287886 on Build GRCh38
rs956923881 Chr.9: 97287708-97287708 on Build GRCh38
rs749783861 Chr.9: 97287619-97287619 on Build GRCh38
rs140186708 Chr.9: 97287830-97287830 on Build GRCh38
rs1002420940 Chr.9: 97287941-97287941 on Build GRCh38
rs1027585647 Chr.9: 97288034-97288034 on Build GRCh38
rs779177923 Chr.9: 97287639-97287639 on Build GRCh38
rs1012507759 Chr.9: 97287687-97287687 on Build GRCh38
rs777070471 Chr.9: 97287868-97287868 on Build GRCh38
rs371477373 Chr.9: 97287666-97287666 on Build GRCh38
rs558339303 Chr.9: 97287979-97287979 on Build GRCh38
rs994246873 Chr.9: 97287902-97287902 on Build GRCh38
rs1050586305 Chr.9: 97287611-97287611 on Build GRCh38
rs78881544 Chr.9: 97287928-97287928 on Build GRCh38
rs983336041 Chr.9: 97288064-97288064 on Build GRCh38
rs761371551 Chr.9: 97287878-97287878 on Build GRCh38
rs189749792 Chr.9: 97287781-97287781 on Build GRCh38
rs769137275 Chr.9: 97287733-97287733 on Build GRCh38
rs904330364 Chr.9: 97287767-97287767 on Build GRCh38
rs143119785 Chr.9: 97287686-97287686 on Build GRCh38
rs1023548639 Chr.9: 97287933-97287933 on Build GRCh38
rs1052904588 Chr.9: 97287826-97287826 on Build GRCh38
rs386736611 Chr.9: 97287688-97287690 on Build GRCh38
rs2493444 Chr.9: 97287688-97287688 on Build GRCh38
rs932148081 Chr.9: 97287597-97287597 on Build GRCh38
rs528573887 Chr.9: 97287685-97287685 on Build GRCh38
rs762749219 Chr.9: 97287723-97287723 on Build GRCh38
rs772888726 Chr.9: 97287971-97287971 on Build GRCh38
rs895553354 Chr.9: 97287916-97287916 on Build GRCh38
rs767339285 Chr.9: 97287780-97287780 on Build GRCh38
rs1038074225 Chr.9: 97287644-97287644 on Build GRCh38
rs74473798 Chr.9: 97287652-97287652 on Build GRCh38
rs562065716 Chr.9: 97288087-97288087 on Build GRCh38
rs116286944 Chr.9: 97288044-97288044 on Build GRCh38
rs186533426 Chr.9: 97287667-97287667 on Build GRCh38
rs976073500 Chr.9: 97287710-97287710 on Build GRCh38
rs769274239 Chr.9: 97287767-97287767 on Build GRCh38
rs552551897 Chr.9: 97287672-97287672 on Build GRCh38
rs1050617818 Chr.9: 97287751-97287751 on Build GRCh38
rs573367736 Chr.9: 97288038-97288038 on Build GRCh38
rs534801997 Chr.9: 97287801-97287801 on Build GRCh38
rs746555550 Chr.9: 97287680-97287680 on Build GRCh38
rs1000661503 Chr.9: 97287992-97287992 on Build GRCh38
rs1043283633 Chr.9: 97287766-97287766 on Build GRCh38
rs967912195 Chr.9: 97287962-97287962 on Build GRCh38
rs959672496 Chr.9: 97287651-97287651 on Build GRCh38
rs944042920 Chr.9: 97287706-97287706 on Build GRCh38
rs904374589 Chr.9: 97287593-97287593 on Build GRCh38
rs1005283339 Chr.9: 97287638-97287638 on Build GRCh38
rs1056105504 Chr.9: 97287912-97287912 on Build GRCh38
rs923529576 Chr.9: 97287729-97287729 on Build GRCh38
rs148259002 Chr.9: 97287697-97287697 on Build GRCh38
rs758887272 Chr.9: 97288013-97288013 on Build GRCh38
rs1012193925 Chr.9: 97287929-97287929 on Build GRCh38
rs2487874 Chr.9: 97287690-97287690 on Build GRCh38
rs921215041 Chr.9: 97287717-97287717 on Build GRCh38
rs546293533 Chr.9: 97287616-97287616 on Build GRCh38
rs908665178 Chr.9: 97288066-97288066 on Build GRCh38
rs979545646 Chr.9: 97287696-97287696 on Build GRCh38
rs375024643 Chr.9: 97287594-97287594 on Build GRCh38
rs904277310 Chr.9: 97287600-97287600 on Build GRCh38

View Full Product Details