Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97296247-97296791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100499484
Entrez Gene ID
100499484
Gene Name
SUGT1-1300002K09Rik pseudogene
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97297314 on Build GRCh38
Transcripts
NR_036526.1 NR_036527.1 NR_036528.1 NR_036529.1

Gene Symbol
LOC100499484-C9ORF174
Entrez Gene ID
57653
Gene Name
LOC100499484-C9orf174 readthrough
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97377295 on Build GRCh38
Transcripts
NR_036526.1 NR_036527.1 NR_036528.1 NR_036529.1

SNP Detail

SNP ID Chromosome Location
rs534107427 Chr.9: 97296758-97296758 on Build GRCh38
rs927485399 Chr.9: 97296706-97296706 on Build GRCh38
rs1011748879 Chr.9: 97296709-97296709 on Build GRCh38
rs950724388 Chr.9: 97296631-97296631 on Build GRCh38
rs1003398045 Chr.9: 97296731-97296731 on Build GRCh38
rs771456129 Chr.9: 97296496-97296496 on Build GRCh38
rs958734808 Chr.9: 97296755-97296755 on Build GRCh38
rs936163203 Chr.9: 97296725-97296725 on Build GRCh38
rs1051454843 Chr.9: 97296567-97296567 on Build GRCh38
rs774886726 Chr.9: 97296558-97296558 on Build GRCh38
rs150838636 Chr.9: 97296538-97296538 on Build GRCh38
rs983739851 Chr.9: 97296293-97296293 on Build GRCh38
rs893354194 Chr.9: 97296694-97296694 on Build GRCh38
rs909127503 Chr.9: 97296527-97296527 on Build GRCh38
rs981483105 Chr.9: 97296368-97296368 on Build GRCh38
rs763232125 Chr.9: 97296453-97296453 on Build GRCh38
rs890351819 Chr.9: 97296539-97296539 on Build GRCh38
rs941878096 Chr.9: 97296537-97296537 on Build GRCh38
rs1020846891 Chr.9: 97296725-97296726 on Build GRCh38
rs1009179210 Chr.9: 97296533-97296533 on Build GRCh38
rs185737003 Chr.9: 97296310-97296310 on Build GRCh38
rs1009203154 Chr.9: 97296552-97296552 on Build GRCh38
rs1053907490 Chr.9: 97296466-97296466 on Build GRCh38
rs752224510 Chr.9: 97296730-97296730 on Build GRCh38
rs937311226 Chr.9: 97296430-97296430 on Build GRCh38
rs1053572594 Chr.9: 97296741-97296741 on Build GRCh38
rs917231878 Chr.9: 97296672-97296672 on Build GRCh38
rs143221663 Chr.9: 97296417-97296417 on Build GRCh38
rs917493140 Chr.9: 97296311-97296311 on Build GRCh38
rs964864428 Chr.9: 97296581-97296581 on Build GRCh38
rs1035760734 Chr.9: 97296752-97296753 on Build GRCh38
rs2290933 Chr.9: 97296593-97296593 on Build GRCh38
rs372967940 Chr.9: 97296708-97296708 on Build GRCh38
rs980726999 Chr.9: 97296684-97296684 on Build GRCh38
rs933043730 Chr.9: 97296561-97296561 on Build GRCh38
rs76549934 Chr.9: 97296557-97296557 on Build GRCh38
rs950220396 Chr.9: 97296363-97296363 on Build GRCh38
rs73559813 Chr.9: 97296676-97296676 on Build GRCh38
rs983433120 Chr.9: 97296640-97296640 on Build GRCh38
rs1027733617 Chr.9: 97296601-97296601 on Build GRCh38
rs773566765 Chr.9: 97296441-97296441 on Build GRCh38

View Full Product Details