Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130495191-130495743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM45BP
Entrez Gene ID
55855
Gene Name
family with sequence similarity 45, member A pseudogene
Gene Aliases
FAM45B, HT011
UniGene
Hs.567120
Gene Chromosome Location
Chr.X: 130494941-130497448 on Build GRCh38
Transcripts
NR_027141.2

SNP Detail

SNP ID Chromosome Location
rs143809204 Chr.X: 130495597-130495597 on Build GRCh38
rs770268167 Chr.X: 130495220-130495220 on Build GRCh38
rs779416711 Chr.X: 130495593-130495593 on Build GRCh38
rs746637655 Chr.X: 130495300-130495300 on Build GRCh38
rs745309373 Chr.X: 130495307-130495307 on Build GRCh38
rs778325279 Chr.X: 130495621-130495621 on Build GRCh38
rs377221061 Chr.X: 130495238-130495238 on Build GRCh38
rs758083899 Chr.X: 130495291-130495291 on Build GRCh38
rs1015770323 Chr.X: 130495235-130495235 on Build GRCh38
rs113434089 Chr.X: 130495437-130495437 on Build GRCh38
rs955890661 Chr.X: 130495431-130495431 on Build GRCh38
rs1044370765 Chr.X: 130495706-130495706 on Build GRCh38
rs780673174 Chr.X: 130495306-130495306 on Build GRCh38
rs757046734 Chr.X: 130495405-130495405 on Build GRCh38
rs760993320 Chr.X: 130495528-130495528 on Build GRCh38
rs760240367 Chr.X: 130495699-130495699 on Build GRCh38
rs749737563 Chr.X: 130495446-130495446 on Build GRCh38
rs750291601 Chr.X: 130495693-130495693 on Build GRCh38
rs766341713 Chr.X: 130495536-130495536 on Build GRCh38
rs762167629 Chr.X: 130495239-130495239 on Build GRCh38
rs752334037 Chr.X: 130495554-130495554 on Build GRCh38
rs764540946 Chr.X: 130495234-130495234 on Build GRCh38
rs976403992 Chr.X: 130495438-130495438 on Build GRCh38
rs763948830 Chr.X: 130495561-130495561 on Build GRCh38
rs772594024 Chr.X: 130495688-130495688 on Build GRCh38
rs766506400 Chr.X: 130495376-130495376 on Build GRCh38
rs756958777 Chr.X: 130495301-130495301 on Build GRCh38
rs773520882 Chr.X: 130495524-130495524 on Build GRCh38
rs773575964 Chr.X: 130495363-130495363 on Build GRCh38
rs753228932 Chr.X: 130495719-130495719 on Build GRCh38
rs755337734 Chr.X: 130495384-130495385 on Build GRCh38
rs779507562 Chr.X: 130495609-130495609 on Build GRCh38
rs775811919 Chr.X: 130495310-130495310 on Build GRCh38
rs761778144 Chr.X: 130495655-130495655 on Build GRCh38
rs778600857 Chr.X: 130495712-130495712 on Build GRCh38
rs199958944 Chr.X: 130495344-130495344 on Build GRCh38
rs750589805 Chr.X: 130495263-130495263 on Build GRCh38
rs769358091 Chr.X: 130495308-130495308 on Build GRCh38
rs774853469 Chr.X: 130495474-130495474 on Build GRCh38
rs753448016 Chr.X: 130495701-130495701 on Build GRCh38
rs370215306 Chr.X: 130495622-130495622 on Build GRCh38
rs1014180852 Chr.X: 130495262-130495262 on Build GRCh38
rs745555482 Chr.X: 130495420-130495420 on Build GRCh38
rs970297216 Chr.X: 130495264-130495264 on Build GRCh38
rs754939422 Chr.X: 130495267-130495267 on Build GRCh38
rs769264053 Chr.X: 130495464-130495464 on Build GRCh38
rs751510340 Chr.X: 130495562-130495562 on Build GRCh38
rs758053897 Chr.X: 130495578-130495578 on Build GRCh38
rs769883413 Chr.X: 130495646-130495646 on Build GRCh38
rs754492139 Chr.X: 130495711-130495711 on Build GRCh38
rs755840949 Chr.X: 130495587-130495587 on Build GRCh38
rs763167404 Chr.X: 130495224-130495224 on Build GRCh38
rs753699695 Chr.X: 130495349-130495349 on Build GRCh38
rs373545516 Chr.X: 130495538-130495538 on Build GRCh38
rs989967521 Chr.X: 130495435-130495435 on Build GRCh38
rs776720515 Chr.X: 130495628-130495628 on Build GRCh38
rs373657948 Chr.X: 130495411-130495411 on Build GRCh38
rs371540123 Chr.X: 130495299-130495299 on Build GRCh38
rs924551052 Chr.X: 130495378-130495378 on Build GRCh38
rs1000780780 Chr.X: 130495272-130495272 on Build GRCh38
rs752540662 Chr.X: 130495390-130495390 on Build GRCh38
rs763858811 Chr.X: 130495397-130495397 on Build GRCh38
rs372438369 Chr.X: 130495216-130495216 on Build GRCh38
rs41301495 Chr.X: 130495246-130495246 on Build GRCh38
rs768872165 Chr.X: 130495338-130495338 on Build GRCh38
rs766356342 Chr.X: 130495286-130495286 on Build GRCh38
rs765019912 Chr.X: 130495281-130495281 on Build GRCh38
rs866529241 Chr.X: 130495373-130495373 on Build GRCh38
rs772528514 Chr.X: 130495615-130495615 on Build GRCh38
rs376392342 Chr.X: 130495610-130495610 on Build GRCh38
rs774230816 Chr.X: 130495647-130495647 on Build GRCh38
rs776067196 Chr.X: 130495223-130495223 on Build GRCh38
rs755539444 Chr.X: 130495426-130495426 on Build GRCh38
rs370371782 Chr.X: 130495670-130495670 on Build GRCh38
rs377194384 Chr.X: 130495633-130495633 on Build GRCh38
rs369187336 Chr.X: 130495391-130495391 on Build GRCh38
rs749832230 Chr.X: 130495334-130495334 on Build GRCh38
rs754541151 Chr.X: 130495595-130495595 on Build GRCh38
rs751420353 Chr.X: 130495400-130495400 on Build GRCh38
rs200882990 Chr.X: 130495494-130495494 on Build GRCh38
rs772581518 Chr.X: 130495522-130495522 on Build GRCh38
rs759356233 Chr.X: 130495362-130495362 on Build GRCh38
rs201967377 Chr.X: 130495625-130495625 on Build GRCh38
rs767330995 Chr.X: 130495579-130495579 on Build GRCh38

View Full Product Details