Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130496524-130497115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM45BP
Entrez Gene ID
55855
Gene Name
family with sequence similarity 45, member A pseudogene
Gene Aliases
FAM45B, HT011
UniGene
Hs.567120
Gene Chromosome Location
Chr.X: 130494941-130497448 on Build GRCh38
Transcripts
NR_027141.2

SNP Detail

SNP ID Chromosome Location
rs763743814 Chr.X: 130496580-130496580 on Build GRCh38
rs761615572 Chr.X: 130496994-130496995 on Build GRCh38
rs906936905 Chr.X: 130496629-130496629 on Build GRCh38
rs1021470335 Chr.X: 130496812-130496812 on Build GRCh38
rs955951622 Chr.X: 130496730-130496730 on Build GRCh38
rs750974488 Chr.X: 130496781-130496781 on Build GRCh38
rs1012368671 Chr.X: 130496572-130496572 on Build GRCh38
rs141480347 Chr.X: 130496759-130496759 on Build GRCh38
rs985302421 Chr.X: 130496962-130496962 on Build GRCh38
rs866064801 Chr.X: 130496778-130496778 on Build GRCh38
rs879242773 Chr.X: 130496756-130496756 on Build GRCh38
rs751089104 Chr.X: 130496589-130496589 on Build GRCh38
rs187726226 Chr.X: 130497018-130497018 on Build GRCh38
rs529619093 Chr.X: 130496778-130496778 on Build GRCh38
rs780646806 Chr.X: 130496692-130496692 on Build GRCh38
rs1002975194 Chr.X: 130496665-130496665 on Build GRCh38
rs1011464999 Chr.X: 130496788-130496788 on Build GRCh38
rs199851627 Chr.X: 130496998-130496998 on Build GRCh38
rs868754601 Chr.X: 130496749-130496749 on Build GRCh38
rs149339159 Chr.X: 130496759-130496760 on Build GRCh38
rs756711979 Chr.X: 130496671-130496671 on Build GRCh38
rs113195090 Chr.X: 130496897-130496897 on Build GRCh38
rs953898027 Chr.X: 130496836-130496836 on Build GRCh38

View Full Product Details