Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16626658-16627206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCCP2
Entrez Gene ID
84809
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 2
Gene Aliases
CROCCL1
UniGene
Hs.631865
Gene Chromosome Location
Chr.1: 16618256-16630906 on Build GRCh38
Transcripts
NR_026752.1

SNP Detail

SNP ID Chromosome Location
rs878936412 Chr.1: 16626895-16626895 on Build GRCh38
rs889514229 Chr.1: 16626693-16626693 on Build GRCh38
rs3738597 Chr.1: 16627177-16627177 on Build GRCh38
rs2005328 Chr.1: 16626917-16626917 on Build GRCh38
rs536911713 Chr.1: 16626752-16626753 on Build GRCh38
rs547559585 Chr.1: 16626942-16626942 on Build GRCh38
rs527877590 Chr.1: 16626832-16626832 on Build GRCh38
rs377729531 Chr.1: 16627187-16627187 on Build GRCh38
rs3738598 Chr.1: 16626929-16626929 on Build GRCh38
rs571710778 Chr.1: 16626702-16626702 on Build GRCh38
rs879123901 Chr.1: 16627171-16627171 on Build GRCh38
rs113836035 Chr.1: 16627169-16627169 on Build GRCh38
rs878916320 Chr.1: 16627154-16627154 on Build GRCh38

View Full Product Details