Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16702497-16703052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPNP
Entrez Gene ID
284729
Gene Name
espin pseudogene
Gene Aliases
dJ1182A14.1
UniGene
Hs.548239
Gene Chromosome Location
Chr.1: 16691218-16720157 on Build GRCh38
Transcripts
NR_026567.1

SNP Detail

SNP ID Chromosome Location
rs773353585 Chr.1: 16702816-16702816 on Build GRCh38
rs756150958 Chr.1: 16702752-16702752 on Build GRCh38
rs745882569 Chr.1: 16702696-16702696 on Build GRCh38
rs750116704 Chr.1: 16702965-16702965 on Build GRCh38
rs780673173 Chr.1: 16702809-16702809 on Build GRCh38
rs763342386 Chr.1: 16702734-16702734 on Build GRCh38
rs523936 Chr.1: 16703018-16703018 on Build GRCh38
rs773623759 Chr.1: 16702862-16702862 on Build GRCh38
rs771611021 Chr.1: 16702772-16702772 on Build GRCh38
rs759632127 Chr.1: 16702834-16702834 on Build GRCh38
rs1019821844 Chr.1: 16702719-16702719 on Build GRCh38
rs780069272 Chr.1: 16703002-16703002 on Build GRCh38
rs747903603 Chr.1: 16702765-16702765 on Build GRCh38
rs750028366 Chr.1: 16702882-16702882 on Build GRCh38
rs754868132 Chr.1: 16702761-16702761 on Build GRCh38
rs752448799 Chr.1: 16702796-16702796 on Build GRCh38
rs755989139 Chr.1: 16702697-16702697 on Build GRCh38
rs771383215 Chr.1: 16702908-16702908 on Build GRCh38
rs776927948 Chr.1: 16702867-16702867 on Build GRCh38
rs761706241 Chr.1: 16703011-16703011 on Build GRCh38
rs758708055 Chr.1: 16702670-16702670 on Build GRCh38
rs766543506 Chr.1: 16702788-16702788 on Build GRCh38
rs746163707 Chr.1: 16702996-16702996 on Build GRCh38
rs779309735 Chr.1: 16702853-16702853 on Build GRCh38
rs774320763 Chr.1: 16703009-16703009 on Build GRCh38
rs753046653 Chr.1: 16702664-16702664 on Build GRCh38
rs778086766 Chr.1: 16702896-16702896 on Build GRCh38
rs760330284 Chr.1: 16703021-16703021 on Build GRCh38
rs530756384 Chr.1: 16702790-16702790 on Build GRCh38
rs752643489 Chr.1: 16702837-16702837 on Build GRCh38
rs778909820 Chr.1: 16702792-16702792 on Build GRCh38
rs769073244 Chr.1: 16702780-16702780 on Build GRCh38
rs751817340 Chr.1: 16702742-16702742 on Build GRCh38
rs761438579 Chr.1: 16702656-16702656 on Build GRCh38
rs781099224 Chr.1: 16702684-16702684 on Build GRCh38
rs774770014 Chr.1: 16702814-16702814 on Build GRCh38
rs137866947 Chr.1: 16702786-16702786 on Build GRCh38
rs766633583 Chr.1: 16702823-16702823 on Build GRCh38
rs914859376 Chr.1: 16702869-16702869 on Build GRCh38
rs761614617 Chr.1: 16702954-16702954 on Build GRCh38
rs777019831 Chr.1: 16702917-16702917 on Build GRCh38
rs768567372 Chr.1: 16703007-16703007 on Build GRCh38
rs768529381 Chr.1: 16702713-16702713 on Build GRCh38
rs764308867 Chr.1: 16702674-16702674 on Build GRCh38
rs770076320 Chr.1: 16702676-16702676 on Build GRCh38
rs748873386 Chr.1: 16702931-16702931 on Build GRCh38
rs776001190 Chr.1: 16702777-16702777 on Build GRCh38
rs746626650 Chr.1: 16702800-16702800 on Build GRCh38
rs750256970 Chr.1: 16702933-16702934 on Build GRCh38
rs777247146 Chr.1: 16702659-16702659 on Build GRCh38
rs760350752 Chr.1: 16702945-16702945 on Build GRCh38
rs557311536 Chr.1: 16702698-16702698 on Build GRCh38
rs756887076 Chr.1: 16702841-16702841 on Build GRCh38
rs758932929 Chr.1: 16702894-16702894 on Build GRCh38
rs777427596 Chr.1: 16702773-16702773 on Build GRCh38
rs757414756 Chr.1: 16702746-16702746 on Build GRCh38
rs760918835 Chr.1: 16702820-16702820 on Build GRCh38
rs746069444 Chr.1: 16702927-16702927 on Build GRCh38
rs765363887 Chr.1: 16702873-16702873 on Build GRCh38
rs759931468 Chr.1: 16702661-16702661 on Build GRCh38
rs761848727 Chr.1: 16702873-16702873 on Build GRCh38
rs775341699 Chr.1: 16702874-16702874 on Build GRCh38
rs768350164 Chr.1: 16702931-16702931 on Build GRCh38
rs772593312 Chr.1: 16702728-16702728 on Build GRCh38
rs764396743 Chr.1: 16702738-16702738 on Build GRCh38
rs749850132 Chr.1: 16702812-16702812 on Build GRCh38
rs747294667 Chr.1: 16702865-16702865 on Build GRCh38
rs947610454 Chr.1: 16702872-16702872 on Build GRCh38
rs762174666 Chr.1: 16702782-16702782 on Build GRCh38
rs774497780 Chr.1: 16702864-16702875 on Build GRCh38
rs774080153 Chr.1: 16702654-16702654 on Build GRCh38
rs778322691 Chr.1: 16702771-16702771 on Build GRCh38
rs759725119 Chr.1: 16702868-16702868 on Build GRCh38
rs572960295 Chr.1: 16702760-16702760 on Build GRCh38
rs12125112 Chr.1: 16702762-16702762 on Build GRCh38
rs200159413 Chr.1: 16702708-16702708 on Build GRCh38
rs781559659 Chr.1: 16702813-16702813 on Build GRCh38
rs753403403 Chr.1: 16702970-16702970 on Build GRCh38
rs764094435 Chr.1: 16702952-16702952 on Build GRCh38
rs778377568 Chr.1: 16702976-16702976 on Build GRCh38
rs772433221 Chr.1: 16702815-16702815 on Build GRCh38
rs766886867 Chr.1: 16702657-16702657 on Build GRCh38
rs777604293 Chr.1: 16702839-16702839 on Build GRCh38
rs748666580 Chr.1: 16702854-16702854 on Build GRCh38
rs568440358 Chr.1: 16702562-16702562 on Build GRCh38
rs774167861 Chr.1: 16702715-16702715 on Build GRCh38
rs772977851 Chr.1: 16703019-16703019 on Build GRCh38
rs766100738 Chr.1: 16702975-16702975 on Build GRCh38
rs544457949 Chr.1: 16702849-16702849 on Build GRCh38
rs780763054 Chr.1: 16702847-16702847 on Build GRCh38
rs1030673170 Chr.1: 16702845-16702845 on Build GRCh38
rs768843950 Chr.1: 16702860-16702861 on Build GRCh38
rs765701231 Chr.1: 16702662-16702662 on Build GRCh38
rs777326080 Chr.1: 16702799-16702799 on Build GRCh38
rs752102171 Chr.1: 16702779-16702779 on Build GRCh38
rs745383287 Chr.1: 16702848-16702848 on Build GRCh38
rs758149682 Chr.1: 16702797-16702797 on Build GRCh38
rs537745814 Chr.1: 16702683-16702683 on Build GRCh38
rs751743927 Chr.1: 16702678-16702678 on Build GRCh38
rs757748114 Chr.1: 16702993-16702993 on Build GRCh38
rs753462117 Chr.1: 16703022-16703023 on Build GRCh38
rs763164923 Chr.1: 16702816-16702839 on Build GRCh38
rs564132501 Chr.1: 16702942-16702942 on Build GRCh38
rs770064504 Chr.1: 16702998-16702998 on Build GRCh38
rs751214763 Chr.1: 16702840-16702840 on Build GRCh38
rs760692087 Chr.1: 16702787-16702787 on Build GRCh38
rs553071177 Chr.1: 16702733-16702733 on Build GRCh38
rs989074761 Chr.1: 16702863-16702863 on Build GRCh38
rs771291397 Chr.1: 16702866-16702866 on Build GRCh38
rs749414335 Chr.1: 16703003-16703003 on Build GRCh38
rs759855515 Chr.1: 16702530-16702530 on Build GRCh38
rs758155165 Chr.1: 16702838-16702838 on Build GRCh38
rs746509299 Chr.1: 16702776-16702776 on Build GRCh38
rs767689713 Chr.1: 16702748-16702748 on Build GRCh38
rs747782522 Chr.1: 16702727-16702727 on Build GRCh38
rs752067737 Chr.1: 16702990-16702990 on Build GRCh38
rs770480283 Chr.1: 16702805-16702805 on Build GRCh38
rs756975096 Chr.1: 16702879-16702879 on Build GRCh38
rs754381268 Chr.1: 16702972-16702972 on Build GRCh38
rs774269835 Chr.1: 16702994-16702994 on Build GRCh38
rs753956079 Chr.1: 16702832-16702832 on Build GRCh38
rs778487349 Chr.1: 16702961-16702961 on Build GRCh38
rs11260891 Chr.1: 16702967-16702967 on Build GRCh38
rs780170989 Chr.1: 16702756-16702756 on Build GRCh38
rs750479540 Chr.1: 16702749-16702749 on Build GRCh38
rs747382642 Chr.1: 16702906-16702906 on Build GRCh38
rs754963363 Chr.1: 16702791-16702791 on Build GRCh38
rs755669145 Chr.1: 16702884-16702884 on Build GRCh38
rs767183979 Chr.1: 16702880-16702880 on Build GRCh38
rs763849981 Chr.1: 16702877-16702877 on Build GRCh38
rs192986203 Chr.1: 16702731-16702731 on Build GRCh38
rs775533992 Chr.1: 16702686-16702687 on Build GRCh38
rs772361164 Chr.1: 16702855-16702855 on Build GRCh38
rs374854214 Chr.1: 16702893-16702893 on Build GRCh38
rs765679529 Chr.1: 16702732-16702732 on Build GRCh38

View Full Product Details