Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16719844-16720396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPNP
Entrez Gene ID
284729
Gene Name
espin pseudogene
Gene Aliases
dJ1182A14.1
UniGene
Hs.548239
Gene Chromosome Location
Chr.1: 16691218-16720157 on Build GRCh38
Transcripts
NR_026567.1

SNP Detail

SNP ID Chromosome Location
rs796886283 Chr.1: 16720162-16720162 on Build GRCh38
rs765878080 Chr.1: 16719980-16719980 on Build GRCh38
rs9661747 Chr.1: 16720336-16720336 on Build GRCh38
rs779240598 Chr.1: 16720057-16720058 on Build GRCh38
rs1755333 Chr.1: 16720301-16720301 on Build GRCh38
rs774878928 Chr.1: 16720053-16720053 on Build GRCh38
rs776867165 Chr.1: 16720101-16720101 on Build GRCh38
rs767661519 Chr.1: 16720033-16720033 on Build GRCh38
rs542513825 Chr.1: 16720290-16720290 on Build GRCh38
rs3962312 Chr.1: 16720121-16720121 on Build GRCh38
rs762674670 Chr.1: 16719927-16719927 on Build GRCh38
rs373523038 Chr.1: 16720200-16720200 on Build GRCh38
rs780107919 Chr.1: 16720036-16720036 on Build GRCh38
rs757413661 Chr.1: 16719951-16719951 on Build GRCh38
rs3982088 Chr.1: 16720276-16720276 on Build GRCh38
rs776678514 Chr.1: 16719922-16719922 on Build GRCh38
rs551134197 Chr.1: 16720363-16720363 on Build GRCh38
rs573087715 Chr.1: 16719908-16719908 on Build GRCh38
rs774024194 Chr.1: 16720017-16720017 on Build GRCh38
rs749125869 Chr.1: 16719963-16719963 on Build GRCh38
rs780018598 Chr.1: 16719999-16719999 on Build GRCh38
rs56383686 Chr.1: 16720220-16720220 on Build GRCh38
rs756143559 Chr.1: 16719997-16719997 on Build GRCh38
rs577267024 Chr.1: 16720234-16720234 on Build GRCh38
rs762186705 Chr.1: 16720032-16720032 on Build GRCh38
rs771870998 Chr.1: 16720041-16720041 on Build GRCh38
rs59873084 Chr.1: 16720131-16720131 on Build GRCh38
rs769732171 Chr.1: 16719962-16719962 on Build GRCh38
rs748281848 Chr.1: 16720070-16720071 on Build GRCh38
rs3962311 Chr.1: 16720139-16720139 on Build GRCh38
rs753851920 Chr.1: 16720037-16720037 on Build GRCh38
rs3982089 Chr.1: 16720203-16720203 on Build GRCh38
rs756980082 Chr.1: 16720078-16720078 on Build GRCh38
rs555102291 Chr.1: 16719926-16719926 on Build GRCh38
rs752960715 Chr.1: 16719923-16719923 on Build GRCh38
rs771647755 Chr.1: 16720019-16720019 on Build GRCh38
rs3856294 Chr.1: 16720118-16720118 on Build GRCh38
rs772397268 Chr.1: 16720086-16720086 on Build GRCh38
rs754436029 Chr.1: 16719938-16719938 on Build GRCh38
rs60861463 Chr.1: 16719967-16719967 on Build GRCh38
rs760979433 Chr.1: 16720087-16720087 on Build GRCh38
rs550371505 Chr.1: 16720070-16720070 on Build GRCh38
rs376263829 Chr.1: 16719907-16719907 on Build GRCh38
rs531165436 Chr.1: 16720321-16720321 on Build GRCh38
rs772611562 Chr.1: 16719974-16719974 on Build GRCh38
rs535304766 Chr.1: 16719925-16719925 on Build GRCh38
rs745954684 Chr.1: 16719996-16719996 on Build GRCh38
rs769062081 Chr.1: 16720052-16720052 on Build GRCh38
rs745399449 Chr.1: 16720079-16720079 on Build GRCh38
rs770631202 Chr.1: 16720044-16720044 on Build GRCh38
rs776197257 Chr.1: 16720025-16720025 on Build GRCh38
rs761439235 Chr.1: 16719933-16719933 on Build GRCh38
rs1755332 Chr.1: 16720054-16720054 on Build GRCh38
rs747175379 Chr.1: 16719950-16719950 on Build GRCh38
rs755446986 Chr.1: 16719939-16719939 on Build GRCh38
rs758880268 Chr.1: 16719981-16719981 on Build GRCh38
rs774781294 Chr.1: 16720085-16720085 on Build GRCh38
rs546565323 Chr.1: 16720115-16720115 on Build GRCh38
rs777620236 Chr.1: 16720071-16720071 on Build GRCh38
rs570513357 Chr.1: 16720097-16720097 on Build GRCh38
rs769313065 Chr.1: 16720083-16720083 on Build GRCh38
rs545919362 Chr.1: 16720264-16720264 on Build GRCh38
rs559843119 Chr.1: 16720010-16720010 on Build GRCh38
rs3856291 Chr.1: 16719866-16719866 on Build GRCh38
rs747919928 Chr.1: 16720018-16720018 on Build GRCh38
rs753134957 Chr.1: 16719941-16719941 on Build GRCh38
rs750640031 Chr.1: 16720034-16720034 on Build GRCh38
rs369885829 Chr.1: 16720272-16720272 on Build GRCh38
rs557200413 Chr.1: 16720206-16720206 on Build GRCh38
rs764540273 Chr.1: 16719982-16719982 on Build GRCh38
rs773965715 Chr.1: 16719932-16719932 on Build GRCh38
rs766533913 Chr.1: 16720058-16720058 on Build GRCh38
rs577200669 Chr.1: 16719988-16719988 on Build GRCh38
rs563714163 Chr.1: 16719968-16719968 on Build GRCh38
rs747927466 Chr.1: 16720038-16720038 on Build GRCh38
rs759054909 Chr.1: 16720051-16720051 on Build GRCh38
rs539108298 Chr.1: 16720114-16720114 on Build GRCh38
rs763549045 Chr.1: 16720026-16720026 on Build GRCh38
rs755275820 Chr.1: 16719991-16719992 on Build GRCh38
rs575058338 Chr.1: 16720173-16720173 on Build GRCh38
rs778757428 Chr.1: 16720061-16720061 on Build GRCh38
rs3982077 Chr.1: 16719976-16719976 on Build GRCh38
rs3856292 Chr.1: 16719961-16719961 on Build GRCh38
rs530645198 Chr.1: 16720060-16720060 on Build GRCh38
rs756569556 Chr.1: 16719947-16719949 on Build GRCh38
rs9728204 Chr.1: 16719944-16719944 on Build GRCh38
rs2235689 Chr.1: 16719977-16719977 on Build GRCh38
rs760301017 Chr.1: 16720020-16720020 on Build GRCh38
rs756247666 Chr.1: 16720035-16720035 on Build GRCh38
rs766925799 Chr.1: 16719937-16719937 on Build GRCh38
rs746644648 Chr.1: 16720043-16720043 on Build GRCh38
rs767131714 Chr.1: 16719969-16719969 on Build GRCh38
rs537631931 Chr.1: 16720190-16720190 on Build GRCh38
rs555161738 Chr.1: 16720142-16720142 on Build GRCh38
rs763754644 Chr.1: 16719928-16719928 on Build GRCh38
rs3962313 Chr.1: 16720110-16720110 on Build GRCh38
rs562217888 Chr.1: 16720059-16720059 on Build GRCh38
rs781124070 Chr.1: 16719956-16719956 on Build GRCh38
rs3962314 Chr.1: 16719889-16719889 on Build GRCh38
rs768509206 Chr.1: 16720007-16720007 on Build GRCh38
rs764591869 Chr.1: 16720030-16720030 on Build GRCh38
rs746732733 Chr.1: 16720073-16720073 on Build GRCh38
rs758794327 Chr.1: 16719942-16719942 on Build GRCh38
rs749609221 Chr.1: 16719968-16719969 on Build GRCh38
rs3982078 Chr.1: 16719887-16719887 on Build GRCh38
rs546155204 Chr.1: 16720002-16720002 on Build GRCh38
rs776285029 Chr.1: 16720048-16720048 on Build GRCh38
rs757421184 Chr.1: 16719991-16719991 on Build GRCh38
rs199750242 Chr.1: 16720166-16720166 on Build GRCh38
rs760895136 Chr.1: 16720056-16720056 on Build GRCh38
rs765782378 Chr.1: 16719940-16719940 on Build GRCh38
rs780287758 Chr.1: 16719966-16719967 on Build GRCh38
rs781413947 Chr.1: 16719995-16719995 on Build GRCh38
rs3856293 Chr.1: 16720100-16720100 on Build GRCh38
rs573583297 Chr.1: 16720280-16720280 on Build GRCh38

View Full Product Details