Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112922895-112923462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR7A2P1
Entrez Gene ID
246182
Gene Name
aldo-keto reductase family 7 member A2 pseudogene 1
Gene Aliases
AFARP1
UniGene
Hs.647691
Gene Chromosome Location
Chr.1: 112923350-112924673 on Build GRCh38
Transcripts
NR_002796.2 NM_001166496.1 NM_003051.3 XM_011542026.2 XM_011542027.2

Gene Symbol
SLC16A1
Entrez Gene ID
6566
Gene Name
solute carrier family 16 member 1
Gene Aliases
HHF7, MCT, MCT1, MCT1D
UniGene
Hs.75231
Gene Chromosome Location
Chr.1: 112911847-112956353 on Build GRCh38
Transcripts
NR_002796.2 NM_001166496.1 NM_003051.3 XM_011542026.2 XM_011542027.2

SNP Detail

SNP ID Chromosome Location
rs113726551 Chr.1: 112923401-112923401 on Build GRCh38
rs968208251 Chr.1: 112923187-112923187 on Build GRCh38
rs970227916 Chr.1: 112922981-112922981 on Build GRCh38
rs377210192 Chr.1: 112923150-112923150 on Build GRCh38
rs60507342 Chr.1: 112923267-112923268 on Build GRCh38
rs561189617 Chr.1: 112922955-112922955 on Build GRCh38
rs1023847803 Chr.1: 112923394-112923394 on Build GRCh38
rs1038424361 Chr.1: 112923121-112923121 on Build GRCh38
rs886452749 Chr.1: 112923222-112923222 on Build GRCh38
rs897198750 Chr.1: 112923128-112923128 on Build GRCh38
rs192930312 Chr.1: 112922935-112922935 on Build GRCh38
rs978482320 Chr.1: 112923209-112923209 on Build GRCh38
rs1001745475 Chr.1: 112923293-112923293 on Build GRCh38
rs869142794 Chr.1: 112923262-112923263 on Build GRCh38
rs11273193 Chr.1: 112923261-112923262 on Build GRCh38
rs182785871 Chr.1: 112923137-112923137 on Build GRCh38
rs369665337 Chr.1: 112923298-112923298 on Build GRCh38
rs116757457 Chr.1: 112923025-112923025 on Build GRCh38
rs889600547 Chr.1: 112922963-112922963 on Build GRCh38
rs925628649 Chr.1: 112923011-112923011 on Build GRCh38
rs151182701 Chr.1: 112923430-112923430 on Build GRCh38
rs530218368 Chr.1: 112922996-112922996 on Build GRCh38
rs142170475 Chr.1: 112923303-112923303 on Build GRCh38
rs1037962368 Chr.1: 112923268-112923268 on Build GRCh38
rs938365646 Chr.1: 112922929-112922929 on Build GRCh38
rs1056496720 Chr.1: 112923365-112923365 on Build GRCh38
rs1037699904 Chr.1: 112923421-112923421 on Build GRCh38
rs1050044147 Chr.1: 112923068-112923068 on Build GRCh38
rs955736191 Chr.1: 112923261-112923261 on Build GRCh38
rs960052381 Chr.1: 112923132-112923132 on Build GRCh38
rs911049250 Chr.1: 112923340-112923340 on Build GRCh38
rs754515533 Chr.1: 112923006-112923006 on Build GRCh38
rs114230280 Chr.1: 112923317-112923317 on Build GRCh38
rs548185731 Chr.1: 112923217-112923217 on Build GRCh38
rs928352099 Chr.1: 112922922-112922922 on Build GRCh38
rs922512042 Chr.1: 112923057-112923057 on Build GRCh38
rs1012475299 Chr.1: 112923175-112923175 on Build GRCh38
rs886717478 Chr.1: 112923054-112923054 on Build GRCh38
rs933383743 Chr.1: 112923205-112923205 on Build GRCh38
rs906082351 Chr.1: 112923269-112923269 on Build GRCh38
rs372796513 Chr.1: 112923117-112923117 on Build GRCh38
rs778277758 Chr.1: 112923031-112923031 on Build GRCh38
rs188723492 Chr.1: 112923383-112923383 on Build GRCh38
rs1016496700 Chr.1: 112923108-112923108 on Build GRCh38
rs1041242336 Chr.1: 112923191-112923191 on Build GRCh38
rs976651229 Chr.1: 112923369-112923369 on Build GRCh38
rs988491715 Chr.1: 112923440-112923440 on Build GRCh38
rs1032749032 Chr.1: 112923416-112923416 on Build GRCh38
rs752549406 Chr.1: 112923342-112923342 on Build GRCh38
rs889599095 Chr.1: 112923093-112923093 on Build GRCh38
rs961228538 Chr.1: 112923171-112923171 on Build GRCh38
rs57421275 Chr.1: 112922980-112922980 on Build GRCh38
rs552542478 Chr.1: 112923129-112923129 on Build GRCh38
rs372346907 Chr.1: 112923107-112923107 on Build GRCh38
rs897229964 Chr.1: 112923357-112923357 on Build GRCh38
rs923413572 Chr.1: 112923194-112923194 on Build GRCh38
rs371267867 Chr.1: 112923429-112923429 on Build GRCh38
rs988850330 Chr.1: 112922930-112922930 on Build GRCh38
rs912870433 Chr.1: 112923139-112923139 on Build GRCh38
rs925698483 Chr.1: 112923242-112923242 on Build GRCh38
rs546428183 Chr.1: 112923003-112923003 on Build GRCh38
rs980228698 Chr.1: 112923115-112923115 on Build GRCh38
rs1039136284 Chr.1: 112922985-112922985 on Build GRCh38
rs866286087 Chr.1: 112923346-112923346 on Build GRCh38
rs753882919 Chr.1: 112923414-112923416 on Build GRCh38
rs79422765 Chr.1: 112923051-112923051 on Build GRCh38
rs533314801 Chr.1: 112923179-112923179 on Build GRCh38
rs970232410 Chr.1: 112923110-112923110 on Build GRCh38
rs558797131 Chr.1: 112923396-112923396 on Build GRCh38
rs1020001350 Chr.1: 112923311-112923311 on Build GRCh38
rs933915245 Chr.1: 112923304-112923304 on Build GRCh38
rs994193824 Chr.1: 112923364-112923364 on Build GRCh38
rs933865237 Chr.1: 112923061-112923061 on Build GRCh38
rs538923193 Chr.1: 112923391-112923391 on Build GRCh38
rs746061135 Chr.1: 112923395-112923395 on Build GRCh38
rs913184149 Chr.1: 112922934-112922934 on Build GRCh38
rs1038497058 Chr.1: 112923347-112923347 on Build GRCh38
rs755798822 Chr.1: 112923392-112923392 on Build GRCh38

View Full Product Details