Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171138000-171138547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO6P
Entrez Gene ID
388714
Gene Name
flavin containing monooxygenase 6 pseudogene
Gene Aliases
FMO6
UniGene
Hs.448988
Gene Chromosome Location
Chr.1: 171137740-171161563 on Build GRCh38
Transcripts
NR_002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7268977 c.102C>T p.S34S Chr.1: 171138313-171138313 on Build GRCh38
4943465 c.51C>T p.A17A Chr.1: 171138262-171138262 on Build GRCh38
7310729 c.103G>A p.D35N Chr.1: 171138314-171138314 on Build GRCh38
1207317 c.46G>T p.A16S Chr.1: 171138257-171138257 on Build GRCh38
5917185 c.62G>T p.C21F Chr.1: 171138273-171138273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs985033331 Chr.1: 171138154-171138155 on Build GRCh38
rs564082510 Chr.1: 171138239-171138239 on Build GRCh38
rs577205914 Chr.1: 171138084-171138084 on Build GRCh38
rs759356098 Chr.1: 171138346-171138346 on Build GRCh38
rs188740267 Chr.1: 171138294-171138294 on Build GRCh38
rs575318914 Chr.1: 171138235-171138235 on Build GRCh38
rs1002972021 Chr.1: 171138520-171138520 on Build GRCh38
rs545606640 Chr.1: 171138279-171138279 on Build GRCh38
rs937262754 Chr.1: 171138486-171138486 on Build GRCh38
rs766641345 Chr.1: 171138059-171138059 on Build GRCh38
rs34123755 Chr.1: 171138145-171138145 on Build GRCh38
rs774299429 Chr.1: 171138072-171138072 on Build GRCh38
rs911312678 Chr.1: 171138338-171138338 on Build GRCh38
rs541120927 Chr.1: 171138143-171138143 on Build GRCh38
rs918324916 Chr.1: 171138356-171138356 on Build GRCh38
rs745841821 Chr.1: 171138256-171138256 on Build GRCh38
rs972498450 Chr.1: 171138348-171138348 on Build GRCh38
rs748243601 Chr.1: 171138269-171138270 on Build GRCh38
rs1010289763 Chr.1: 171138225-171138225 on Build GRCh38
rs776375612 Chr.1: 171138236-171138236 on Build GRCh38
rs968887872 Chr.1: 171138260-171138260 on Build GRCh38
rs961846015 Chr.1: 171138204-171138204 on Build GRCh38
rs1042015458 Chr.1: 171138413-171138413 on Build GRCh38
rs1036102807 Chr.1: 171138152-171138152 on Build GRCh38
rs995319754 Chr.1: 171138071-171138071 on Build GRCh38
rs923175501 Chr.1: 171138266-171138266 on Build GRCh38
rs979331882 Chr.1: 171138273-171138273 on Build GRCh38
rs142328551 Chr.1: 171138160-171138160 on Build GRCh38
rs191606878 Chr.1: 171138133-171138133 on Build GRCh38
rs1002524001 Chr.1: 171138115-171138115 on Build GRCh38
rs539067375 Chr.1: 171138074-171138074 on Build GRCh38
rs946194870 Chr.1: 171138399-171138399 on Build GRCh38
rs561648335 Chr.1: 171138471-171138471 on Build GRCh38
rs775687698 Chr.1: 171138268-171138268 on Build GRCh38
rs958459698 Chr.1: 171138153-171138153 on Build GRCh38
rs905655639 Chr.1: 171138451-171138451 on Build GRCh38
rs575040404 Chr.1: 171138154-171138154 on Build GRCh38
rs546867178 Chr.1: 171138432-171138432 on Build GRCh38
rs535320824 Chr.1: 171138090-171138090 on Build GRCh38
rs764405582 Chr.1: 171138322-171138322 on Build GRCh38
rs185276884 Chr.1: 171138184-171138184 on Build GRCh38
rs558890586 Chr.1: 171138066-171138066 on Build GRCh38
rs868244007 Chr.1: 171138211-171138211 on Build GRCh38
rs993651686 Chr.1: 171138047-171138047 on Build GRCh38
rs893392844 Chr.1: 171138032-171138032 on Build GRCh38
rs755630960 Chr.1: 171138511-171138511 on Build GRCh38
rs1057018212 Chr.1: 171138521-171138521 on Build GRCh38
rs951751391 Chr.1: 171138435-171138435 on Build GRCh38
rs769723558 Chr.1: 171138259-171138259 on Build GRCh38
rs762022013 Chr.1: 171138350-171138350 on Build GRCh38
rs537025442 Chr.1: 171138037-171138037 on Build GRCh38
rs55762694 Chr.1: 171138138-171138138 on Build GRCh38
rs971997075 Chr.1: 171138242-171138242 on Build GRCh38
rs151282009 Chr.1: 171138391-171138391 on Build GRCh38
rs532912147 Chr.1: 171138320-171138320 on Build GRCh38
rs767943544 Chr.1: 171138412-171138412 on Build GRCh38
rs1029287505 Chr.1: 171138289-171138289 on Build GRCh38
rs752649554 Chr.1: 171138438-171138438 on Build GRCh38
rs78902800 Chr.1: 171138202-171138202 on Build GRCh38
rs774718545 Chr.1: 171138342-171138342 on Build GRCh38

View Full Product Details