Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171161169-171161714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO6P
Entrez Gene ID
388714
Gene Name
flavin containing monooxygenase 6 pseudogene
Gene Aliases
FMO6
UniGene
Hs.448988
Gene Chromosome Location
Chr.1: 171137740-171161563 on Build GRCh38
Transcripts
NR_002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5423318 c.1445G>T p.G482V Chr.1: 171161221-171161221 on Build GRCh38
6317063 c.1474G>T p.A492S Chr.1: 171161250-171161250 on Build GRCh38
1320761 c.1401G>A p.E467E Chr.1: 171161177-171161177 on Build GRCh38
5928082 c.1590C>A p.L530L Chr.1: 171161366-171161366 on Build GRCh38
7295522 c.1538A>G p.Q513R Chr.1: 171161314-171161314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761789859 Chr.1: 171161211-171161211 on Build GRCh38
rs749848644 Chr.1: 171161454-171161457 on Build GRCh38
rs879942476 Chr.1: 171161536-171161536 on Build GRCh38
rs767006797 Chr.1: 171161672-171161672 on Build GRCh38
rs987432060 Chr.1: 171161231-171161231 on Build GRCh38
rs759142391 Chr.1: 171161644-171161644 on Build GRCh38
rs368304191 Chr.1: 171161460-171161460 on Build GRCh38
rs560011106 Chr.1: 171161629-171161629 on Build GRCh38
rs766141120 Chr.1: 171161550-171161550 on Build GRCh38
rs201910707 Chr.1: 171161455-171161455 on Build GRCh38
rs1019954867 Chr.1: 171161512-171161512 on Build GRCh38
rs910832692 Chr.1: 171161579-171161579 on Build GRCh38
rs143026647 Chr.1: 171161528-171161528 on Build GRCh38
rs567543460 Chr.1: 171161223-171161223 on Build GRCh38
rs1053435375 Chr.1: 171161327-171161327 on Build GRCh38
rs1056661727 Chr.1: 171161445-171161445 on Build GRCh38
rs372433069 Chr.1: 171161461-171161461 on Build GRCh38
rs762830601 Chr.1: 171161541-171161541 on Build GRCh38
rs775282075 Chr.1: 171161272-171161272 on Build GRCh38
rs72714112 Chr.1: 171161400-171161400 on Build GRCh38
rs920465568 Chr.1: 171161653-171161653 on Build GRCh38
rs76012774 Chr.1: 171161578-171161578 on Build GRCh38
rs376197625 Chr.1: 171161469-171161470 on Build GRCh38
rs549989513 Chr.1: 171161348-171161348 on Build GRCh38
rs984854712 Chr.1: 171161212-171161212 on Build GRCh38
rs962134178 Chr.1: 171161516-171161516 on Build GRCh38
rs777242236 Chr.1: 171161426-171161426 on Build GRCh38
rs571510730 Chr.1: 171161362-171161362 on Build GRCh38
rs952725112 Chr.1: 171161470-171161470 on Build GRCh38
rs972088662 Chr.1: 171161520-171161521 on Build GRCh38
rs528146821 Chr.1: 171161452-171161457 on Build GRCh38
rs1036418580 Chr.1: 171161294-171161294 on Build GRCh38
rs953407055 Chr.1: 171161210-171161210 on Build GRCh38
rs202213595 Chr.1: 171161316-171161316 on Build GRCh38
rs557982447 Chr.1: 171161381-171161381 on Build GRCh38
rs768459674 Chr.1: 171161290-171161290 on Build GRCh38
rs987768710 Chr.1: 171161577-171161577 on Build GRCh38
rs778362147 Chr.1: 171161457-171161461 on Build GRCh38
rs527303272 Chr.1: 171161202-171161202 on Build GRCh38
rs945113007 Chr.1: 171161680-171161680 on Build GRCh38
rs761704161 Chr.1: 171161324-171161324 on Build GRCh38
rs1006202711 Chr.1: 171161495-171161495 on Build GRCh38
rs768746780 Chr.1: 171161191-171161191 on Build GRCh38
rs910227304 Chr.1: 171161304-171161304 on Build GRCh38
rs961839049 Chr.1: 171161634-171161634 on Build GRCh38
rs751902476 Chr.1: 171161264-171161264 on Build GRCh38
rs746338553 Chr.1: 171161459-171161460 on Build GRCh38
rs145126383 Chr.1: 171161460-171161464 on Build GRCh38
rs3841668 Chr.1: 171161467-171161470 on Build GRCh38
rs566824904 Chr.1: 171161386-171161386 on Build GRCh38
rs892608203 Chr.1: 171161468-171161468 on Build GRCh38
rs769321553 Chr.1: 171161271-171161271 on Build GRCh38
rs544545113 Chr.1: 171161502-171161502 on Build GRCh38
rs906372361 Chr.1: 171161686-171161686 on Build GRCh38
rs748976205 Chr.1: 171161285-171161285 on Build GRCh38
rs16864125 Chr.1: 171161207-171161207 on Build GRCh38
rs923432675 Chr.1: 171161270-171161270 on Build GRCh38
rs962085288 Chr.1: 171161227-171161227 on Build GRCh38
rs1033572666 Chr.1: 171161451-171161452 on Build GRCh38
rs968322387 Chr.1: 171161195-171161195 on Build GRCh38
rs892261083 Chr.1: 171161333-171161333 on Build GRCh38
rs113298887 Chr.1: 171161681-171161681 on Build GRCh38
rs774784917 Chr.1: 171161457-171161457 on Build GRCh38
rs578116199 Chr.1: 171161565-171161565 on Build GRCh38
rs113351195 Chr.1: 171161471-171161471 on Build GRCh38
rs911708011 Chr.1: 171161235-171161235 on Build GRCh38
rs972242731 Chr.1: 171161266-171161266 on Build GRCh38
rs577520016 Chr.1: 171161218-171161218 on Build GRCh38
rs773150942 Chr.1: 171161353-171161353 on Build GRCh38
rs60402482 Chr.1: 171161398-171161398 on Build GRCh38
rs1009314529 Chr.1: 171161355-171161355 on Build GRCh38
rs930709126 Chr.1: 171161661-171161661 on Build GRCh38
rs539878011 Chr.1: 171161378-171161378 on Build GRCh38

View Full Product Details