Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220266415-220266963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKAPS1
Entrez Gene ID
6791
Gene Name
aurora kinase A pseudogene 1
Gene Aliases
AurAps1, STK6P
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220266179-220267715 on Build GRCh38
Transcripts
NR_001587.1 NM_012414.3

Gene Symbol
RAB3GAP2
Entrez Gene ID
25782
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Gene Aliases
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220148268-220272501 on Build GRCh38
Transcripts
NR_001587.1 NM_012414.3

SNP Detail

SNP ID Chromosome Location
rs1003322993 Chr.1: 220266897-220266897 on Build GRCh38
rs111744215 Chr.1: 220266461-220266461 on Build GRCh38
rs945996429 Chr.1: 220266888-220266888 on Build GRCh38
rs939188752 Chr.1: 220266762-220266762 on Build GRCh38
rs186633128 Chr.1: 220266574-220266574 on Build GRCh38
rs111976509 Chr.1: 220266660-220266660 on Build GRCh38
rs1055330726 Chr.1: 220266776-220266776 on Build GRCh38
rs146370576 Chr.1: 220266622-220266622 on Build GRCh38
rs778861436 Chr.1: 220266638-220266638 on Build GRCh38
rs1043721541 Chr.1: 220266843-220266843 on Build GRCh38
rs566548264 Chr.1: 220266858-220266858 on Build GRCh38
rs1030274268 Chr.1: 220266497-220266497 on Build GRCh38
rs955763575 Chr.1: 220266922-220266922 on Build GRCh38
rs758601726 Chr.1: 220266714-220266714 on Build GRCh38
rs1016511613 Chr.1: 220266657-220266657 on Build GRCh38
rs893092346 Chr.1: 220266498-220266498 on Build GRCh38
rs1010757522 Chr.1: 220266812-220266812 on Build GRCh38
rs966786911 Chr.1: 220266457-220266457 on Build GRCh38
rs1029745183 Chr.1: 220266903-220266903 on Build GRCh38
rs1055879903 Chr.1: 220266901-220266901 on Build GRCh38
rs1051345157 Chr.1: 220266437-220266437 on Build GRCh38
rs185315084 Chr.1: 220266479-220266479 on Build GRCh38
rs1034477044 Chr.1: 220266613-220266613 on Build GRCh38
rs1042074034 Chr.1: 220266891-220266891 on Build GRCh38
rs929068454 Chr.1: 220266691-220266691 on Build GRCh38
rs999590173 Chr.1: 220266894-220266894 on Build GRCh38
rs972787301 Chr.1: 220266667-220266667 on Build GRCh38
rs187728196 Chr.1: 220266698-220266698 on Build GRCh38
rs895068179 Chr.1: 220266799-220266799 on Build GRCh38
rs780317476 Chr.1: 220266856-220266856 on Build GRCh38
rs907250722 Chr.1: 220266579-220266579 on Build GRCh38
rs942582695 Chr.1: 220266451-220266451 on Build GRCh38
rs565065884 Chr.1: 220266721-220266721 on Build GRCh38
rs984990313 Chr.1: 220266655-220266655 on Build GRCh38
rs191009432 Chr.1: 220266499-220266499 on Build GRCh38
rs191743749 Chr.1: 220266650-220266650 on Build GRCh38
rs1002966071 Chr.1: 220266609-220266609 on Build GRCh38
rs546730657 Chr.1: 220266736-220266736 on Build GRCh38
rs1006374995 Chr.1: 220266928-220266928 on Build GRCh38
rs894591472 Chr.1: 220266921-220266921 on Build GRCh38
rs958857248 Chr.1: 220266623-220266623 on Build GRCh38
rs750582594 Chr.1: 220266673-220266673 on Build GRCh38
rs911470246 Chr.1: 220266582-220266582 on Build GRCh38
rs181426640 Chr.1: 220266549-220266549 on Build GRCh38
rs574275059 Chr.1: 220266450-220266450 on Build GRCh38
rs942531343 Chr.1: 220266828-220266828 on Build GRCh38
rs868504208 Chr.1: 220266682-220266682 on Build GRCh38

View Full Product Details