Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220267253-220267804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKAPS1
Entrez Gene ID
6791
Gene Name
aurora kinase A pseudogene 1
Gene Aliases
AurAps1, STK6P
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220266179-220267715 on Build GRCh38
Transcripts
NR_001587.1 NM_012414.3

Gene Symbol
RAB3GAP2
Entrez Gene ID
25782
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Gene Aliases
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220148268-220272501 on Build GRCh38
Transcripts
NR_001587.1 NM_012414.3

SNP Detail

SNP ID Chromosome Location
rs772836849 Chr.1: 220267310-220267310 on Build GRCh38
rs559208538 Chr.1: 220267728-220267728 on Build GRCh38
rs955160360 Chr.1: 220267440-220267440 on Build GRCh38
rs754302845 Chr.1: 220267664-220267664 on Build GRCh38
rs75939080 Chr.1: 220267509-220267509 on Build GRCh38
rs769292505 Chr.1: 220267301-220267301 on Build GRCh38
rs148415227 Chr.1: 220267403-220267403 on Build GRCh38
rs45626633 Chr.1: 220267644-220267644 on Build GRCh38
rs367570459 Chr.1: 220267639-220267639 on Build GRCh38
rs979352139 Chr.1: 220267513-220267513 on Build GRCh38
rs983139369 Chr.1: 220267423-220267423 on Build GRCh38
rs991458967 Chr.1: 220267570-220267570 on Build GRCh38
rs1037839323 Chr.1: 220267397-220267397 on Build GRCh38
rs567959611 Chr.1: 220267606-220267606 on Build GRCh38
rs1030575938 Chr.1: 220267413-220267413 on Build GRCh38
rs573507737 Chr.1: 220267706-220267706 on Build GRCh38
rs1055826791 Chr.1: 220267330-220267330 on Build GRCh38
rs547738054 Chr.1: 220267572-220267572 on Build GRCh38
rs774200583 Chr.1: 220267587-220267587 on Build GRCh38
rs144435739 Chr.1: 220267354-220267354 on Build GRCh38
rs770676065 Chr.1: 220267565-220267565 on Build GRCh38
rs34147613 Chr.1: 220267543-220267544 on Build GRCh38
rs979237843 Chr.1: 220267756-220267756 on Build GRCh38
rs1051113809 Chr.1: 220267563-220267563 on Build GRCh38
rs567153400 Chr.1: 220267502-220267502 on Build GRCh38
rs147577815 Chr.1: 220267711-220267711 on Build GRCh38
rs929480160 Chr.1: 220267620-220267620 on Build GRCh38
rs532188433 Chr.1: 220267501-220267501 on Build GRCh38
rs1007833293 Chr.1: 220267442-220267442 on Build GRCh38
rs1007780887 Chr.1: 220267669-220267669 on Build GRCh38
rs117698410 Chr.1: 220267750-220267750 on Build GRCh38
rs890692816 Chr.1: 220267658-220267658 on Build GRCh38
rs546571262 Chr.1: 220267389-220267389 on Build GRCh38
rs1035077444 Chr.1: 220267459-220267459 on Build GRCh38
rs759888311 Chr.1: 220267451-220267451 on Build GRCh38
rs938682090 Chr.1: 220267325-220267325 on Build GRCh38
rs1021831282 Chr.1: 220267508-220267508 on Build GRCh38
rs1039807635 Chr.1: 220267612-220267612 on Build GRCh38
rs1003509145 Chr.1: 220267666-220267666 on Build GRCh38
rs561474726 Chr.1: 220267776-220267776 on Build GRCh38
rs1007356249 Chr.1: 220267351-220267351 on Build GRCh38
rs960502283 Chr.1: 220267484-220267484 on Build GRCh38
rs531046539 Chr.1: 220267298-220267298 on Build GRCh38
rs995679789 Chr.1: 220267617-220267617 on Build GRCh38
rs897939214 Chr.1: 220267618-220267618 on Build GRCh38
rs397814336 Chr.1: 220267548-220267549 on Build GRCh38
rs999475913 Chr.1: 220267280-220267280 on Build GRCh38
rs923746935 Chr.1: 220267769-220267769 on Build GRCh38
rs748064813 Chr.1: 220267288-220267288 on Build GRCh38
rs529762651 Chr.1: 220267755-220267755 on Build GRCh38
rs867187038 Chr.1: 220267410-220267410 on Build GRCh38
rs928576703 Chr.1: 220267304-220267304 on Build GRCh38
rs2808017 Chr.1: 220267376-220267376 on Build GRCh38
rs113970850 Chr.1: 220267608-220267608 on Build GRCh38
rs963728331 Chr.1: 220267489-220267489 on Build GRCh38
rs938598320 Chr.1: 220267537-220267537 on Build GRCh38
rs529950879 Chr.1: 220267383-220267383 on Build GRCh38
rs1023333331 Chr.1: 220267751-220267751 on Build GRCh38
rs960635658 Chr.1: 220267720-220267720 on Build GRCh38
rs967228423 Chr.1: 220267512-220267512 on Build GRCh38
rs1021360937 Chr.1: 220267771-220267771 on Build GRCh38
rs1017916407 Chr.1: 220267473-220267473 on Build GRCh38
rs898697325 Chr.1: 220267613-220267613 on Build GRCh38
rs537558470 Chr.1: 220267540-220267540 on Build GRCh38
rs967737424 Chr.1: 220267752-220267752 on Build GRCh38
rs923869138 Chr.1: 220267533-220267533 on Build GRCh38

View Full Product Details