Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58716254-58716799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICC1
Entrez Gene ID
80114
Gene Name
BicC family RNA binding protein 1
Gene Aliases
BICC, CYSRD
UniGene
Hs.100261
Gene Chromosome Location
Chr.10: 58512970-58831437 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_017016677.1 XM_017016678.1 NR_027508.1

Gene Symbol
FAM133CP
Entrez Gene ID
728640
Gene Name
family with sequence similarity 133, member A pseudogene
Gene Aliases
-
UniGene
Hs.729209
Gene Chromosome Location
Chr.10: 58715015-58717534 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_017016677.1 XM_017016678.1 NR_027508.1

SNP Detail

SNP ID Chromosome Location
rs894424296 Chr.10: 58716419-58716419 on Build GRCh38
rs953786722 Chr.10: 58716568-58716568 on Build GRCh38
rs986410631 Chr.10: 58716587-58716587 on Build GRCh38
rs548395883 Chr.10: 58716742-58716742 on Build GRCh38
rs540206952 Chr.10: 58716614-58716614 on Build GRCh38
rs1003896160 Chr.10: 58716515-58716515 on Build GRCh38
rs935848409 Chr.10: 58716711-58716711 on Build GRCh38
rs182128814 Chr.10: 58716430-58716430 on Build GRCh38
rs911965807 Chr.10: 58716327-58716327 on Build GRCh38
rs986560424 Chr.10: 58716304-58716304 on Build GRCh38
rs765624369 Chr.10: 58716411-58716411 on Build GRCh38
rs935829344 Chr.10: 58716401-58716401 on Build GRCh38
rs1012567629 Chr.10: 58716427-58716427 on Build GRCh38
rs546084231 Chr.10: 58716505-58716505 on Build GRCh38
rs192750009 Chr.10: 58716713-58716713 on Build GRCh38
rs750963878 Chr.10: 58716624-58716624 on Build GRCh38
rs561862773 Chr.10: 58716693-58716693 on Build GRCh38
rs966035878 Chr.10: 58716646-58716646 on Build GRCh38
rs906890588 Chr.10: 58716455-58716455 on Build GRCh38
rs915819678 Chr.10: 58716715-58716715 on Build GRCh38
rs773058374 Chr.10: 58716743-58716743 on Build GRCh38
rs1054372199 Chr.10: 58716418-58716418 on Build GRCh38
rs763495313 Chr.10: 58716629-58716629 on Build GRCh38
rs1027888451 Chr.10: 58716535-58716535 on Build GRCh38
rs745990314 Chr.10: 58716361-58716361 on Build GRCh38
rs574419056 Chr.10: 58716290-58716290 on Build GRCh38
rs944741508 Chr.10: 58716346-58716346 on Build GRCh38
rs911870083 Chr.10: 58716622-58716622 on Build GRCh38
rs1045669353 Chr.10: 58716780-58716780 on Build GRCh38
rs553105627 Chr.10: 58716289-58716289 on Build GRCh38
rs962176574 Chr.10: 58716525-58716525 on Build GRCh38
rs58277560 Chr.10: 58716527-58716527 on Build GRCh38
rs557257571 Chr.10: 58716414-58716414 on Build GRCh38
rs1015300506 Chr.10: 58716524-58716524 on Build GRCh38
rs187716168 Chr.10: 58716578-58716578 on Build GRCh38
rs1041841452 Chr.10: 58716357-58716357 on Build GRCh38
rs948615560 Chr.10: 58716732-58716732 on Build GRCh38
rs536058614 Chr.10: 58716328-58716328 on Build GRCh38
rs977468237 Chr.10: 58716648-58716648 on Build GRCh38
rs931997943 Chr.10: 58716296-58716296 on Build GRCh38

View Full Product Details