Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87843602-87844153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFL1P1
Entrez Gene ID
142913
Gene Name
cofilin 1 pseudogene 1
Gene Aliases
CFLP1
UniGene
Hs.369431
Gene Chromosome Location
Chr.10: 87818313-87845612 on Build GRCh38
Transcripts
NR_028492.1

SNP Detail

SNP ID Chromosome Location
rs996377417 Chr.10: 87843858-87843858 on Build GRCh38
rs946678767 Chr.10: 87844041-87844041 on Build GRCh38
rs890585706 Chr.10: 87843956-87843956 on Build GRCh38
rs960472053 Chr.10: 87844124-87844124 on Build GRCh38
rs548175701 Chr.10: 87843846-87843846 on Build GRCh38
rs980443112 Chr.10: 87844109-87844109 on Build GRCh38
rs559888503 Chr.10: 87843684-87843684 on Build GRCh38
rs756035110 Chr.10: 87844127-87844127 on Build GRCh38
rs61737034 Chr.10: 87844053-87844053 on Build GRCh38
rs995391343 Chr.10: 87844040-87844040 on Build GRCh38
rs10887759 Chr.10: 87843667-87843667 on Build GRCh38
rs1016595506 Chr.10: 87843930-87843930 on Build GRCh38
rs79130126 Chr.10: 87843758-87843758 on Build GRCh38
rs567011209 Chr.10: 87844114-87844114 on Build GRCh38
rs980818335 Chr.10: 87843784-87843784 on Build GRCh38
rs755660927 Chr.10: 87843819-87843819 on Build GRCh38
rs80038176 Chr.10: 87843756-87843756 on Build GRCh38
rs750356240 Chr.10: 87843732-87843732 on Build GRCh38
rs773365490 Chr.10: 87843695-87843695 on Build GRCh38
rs1055412680 Chr.10: 87843669-87843669 on Build GRCh38
rs1029630800 Chr.10: 87843892-87843892 on Build GRCh38
rs188654586 Chr.10: 87843836-87843836 on Build GRCh38
rs568002272 Chr.10: 87844002-87844002 on Build GRCh38
rs927834101 Chr.10: 87843648-87843648 on Build GRCh38
rs1050377000 Chr.10: 87843946-87843946 on Build GRCh38
rs549428905 Chr.10: 87843638-87843638 on Build GRCh38
rs927726186 Chr.10: 87843804-87843804 on Build GRCh38
rs760178 Chr.10: 87844126-87844126 on Build GRCh38
rs531681303 Chr.10: 87843957-87843957 on Build GRCh38
rs916470175 Chr.10: 87843975-87843975 on Build GRCh38
rs1008979353 Chr.10: 87843963-87843963 on Build GRCh38
rs909622712 Chr.10: 87843633-87843633 on Build GRCh38
rs547516443 Chr.10: 87844064-87844064 on Build GRCh38
rs970010779 Chr.10: 87844091-87844091 on Build GRCh38
rs561015337 Chr.10: 87843945-87843945 on Build GRCh38
rs1027757780 Chr.10: 87843696-87843696 on Build GRCh38
rs267602609 Chr.10: 87843902-87843902 on Build GRCh38
rs180805177 Chr.10: 87843960-87843960 on Build GRCh38
rs902580942 Chr.10: 87843738-87843738 on Build GRCh38
rs961136370 Chr.10: 87843636-87843636 on Build GRCh38
rs916517785 Chr.10: 87843673-87843673 on Build GRCh38
rs542867082 Chr.10: 87843928-87843928 on Build GRCh38
rs779519257 Chr.10: 87843964-87843964 on Build GRCh38
rs987967397 Chr.10: 87843834-87843834 on Build GRCh38
rs1043664647 Chr.10: 87843898-87843898 on Build GRCh38
rs899716447 Chr.10: 87843937-87843937 on Build GRCh38
rs946678153 Chr.10: 87843708-87843708 on Build GRCh38
rs144519096 Chr.10: 87844019-87844019 on Build GRCh38

View Full Product Details