Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119117478-119118027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM45A
Entrez Gene ID
404636
Gene Name
family with sequence similarity 45 member A
Gene Aliases
-
UniGene
Hs.434241
Gene Chromosome Location
Chr.10: 119104065-119137984 on Build GRCh38
Transcripts
NM_001303111.1 NM_001303112.1 NM_001303113.1 NM_207009.3 NR_130122.1 XM_006717849.3 XM_006717850.3 XM_006717851.3 XM_006717852.3 XM_017016261.1 XM_017016262.1 XM_017016263.1 XM_017016264.1 XM_017016265.1 XM_017016266.1 XM_017016267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659630 c.352A>T p.S118C Chr.10: 119117538-119117538 on Build GRCh38
7166675 c.396G>A p.K132K Chr.10: 119117582-119117582 on Build GRCh38
5940026 c.467C>T p.A156V Chr.10: 119117653-119117653 on Build GRCh38
168638 c.452G>A p.R151Q Chr.10: 119117638-119117638 on Build GRCh38
3665530 c.357A>G p.P119P Chr.10: 119117543-119117543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901198573 Chr.10: 119117972-119117972 on Build GRCh38
rs967655146 Chr.10: 119117761-119117761 on Build GRCh38
rs146361483 Chr.10: 119117664-119117664 on Build GRCh38
rs762651117 Chr.10: 119117910-119117910 on Build GRCh38
rs541065305 Chr.10: 119117806-119117806 on Build GRCh38
rs1040006048 Chr.10: 119117840-119117840 on Build GRCh38
rs770300595 Chr.10: 119117687-119117687 on Build GRCh38
rs763107028 Chr.10: 119117632-119117632 on Build GRCh38
rs200500337 Chr.10: 119117521-119117521 on Build GRCh38
rs747751815 Chr.10: 119117585-119117585 on Build GRCh38
rs774596962 Chr.10: 119117703-119117703 on Build GRCh38
rs576339730 Chr.10: 119117667-119117667 on Build GRCh38
rs370576956 Chr.10: 119117965-119117966 on Build GRCh38
rs765570555 Chr.10: 119117518-119117518 on Build GRCh38
rs527268352 Chr.10: 119117937-119117937 on Build GRCh38
rs539141106 Chr.10: 119117735-119117735 on Build GRCh38
rs1035956279 Chr.10: 119117942-119117942 on Build GRCh38
rs766413534 Chr.10: 119117659-119117660 on Build GRCh38
rs760001345 Chr.10: 119117609-119117609 on Build GRCh38
rs372426195 Chr.10: 119117648-119117648 on Build GRCh38
rs746689298 Chr.10: 119117678-119117678 on Build GRCh38
rs778879082 Chr.10: 119117578-119117578 on Build GRCh38
rs775857929 Chr.10: 119117622-119117622 on Build GRCh38
rs967604437 Chr.10: 119117929-119117929 on Build GRCh38
rs550075195 Chr.10: 119117998-119117998 on Build GRCh38
rs931454557 Chr.10: 119117961-119117961 on Build GRCh38
rs1005256946 Chr.10: 119117864-119117864 on Build GRCh38
rs1029119275 Chr.10: 119117863-119117863 on Build GRCh38
rs767581508 Chr.10: 119117707-119117707 on Build GRCh38
rs749227931 Chr.10: 119117566-119117566 on Build GRCh38
rs545066574 Chr.10: 119117922-119117922 on Build GRCh38
rs768462892 Chr.10: 119117569-119117569 on Build GRCh38
rs148298113 Chr.10: 119117724-119117724 on Build GRCh38
rs765547136 Chr.10: 119117618-119117618 on Build GRCh38
rs547426989 Chr.10: 119117950-119117950 on Build GRCh38
rs111813037 Chr.10: 119117538-119117538 on Build GRCh38
rs1032354685 Chr.10: 119117587-119117587 on Build GRCh38
rs991474027 Chr.10: 119117728-119117728 on Build GRCh38
rs746338772 Chr.10: 119117685-119117685 on Build GRCh38
rs772779787 Chr.10: 119117591-119117591 on Build GRCh38
rs753320321 Chr.10: 119117693-119117693 on Build GRCh38
rs754892039 Chr.10: 119117649-119117649 on Build GRCh38
rs1005351736 Chr.10: 119118002-119118002 on Build GRCh38
rs531439943 Chr.10: 119117917-119117917 on Build GRCh38
rs777555319 Chr.10: 119117665-119117665 on Build GRCh38
rs759932544 Chr.10: 119117516-119117516 on Build GRCh38
rs937394526 Chr.10: 119117782-119117782 on Build GRCh38
rs750479762 Chr.10: 119117709-119117709 on Build GRCh38
rs889786266 Chr.10: 119117793-119117793 on Build GRCh38
rs895000240 Chr.10: 119117584-119117584 on Build GRCh38
rs553607462 Chr.10: 119117817-119117817 on Build GRCh38
rs747960783 Chr.10: 119117656-119117656 on Build GRCh38
rs374896450 Chr.10: 119117695-119117695 on Build GRCh38
rs11198785 Chr.10: 119117860-119117860 on Build GRCh38
rs542429471 Chr.10: 119117824-119117824 on Build GRCh38
rs879539993 Chr.10: 119117927-119117927 on Build GRCh38
rs751710597 Chr.10: 119117801-119117801 on Build GRCh38
rs778047011 Chr.10: 119117738-119117738 on Build GRCh38
rs760750943 Chr.10: 119117656-119117657 on Build GRCh38
rs923991553 Chr.10: 119117749-119117749 on Build GRCh38
rs556523562 Chr.10: 119117960-119117960 on Build GRCh38
rs928643218 Chr.10: 119117781-119117781 on Build GRCh38
rs769410679 Chr.10: 119117859-119117859 on Build GRCh38
rs755942165 Chr.10: 119117558-119117558 on Build GRCh38
rs28521990 Chr.10: 119117983-119117983 on Build GRCh38
rs761968959 Chr.10: 119117705-119117705 on Build GRCh38
rs113751314 Chr.10: 119117681-119117681 on Build GRCh38
rs753438840 Chr.10: 119117678-119117678 on Build GRCh38
rs995876646 Chr.10: 119117862-119117862 on Build GRCh38
rs766913112 Chr.10: 119117502-119117502 on Build GRCh38
rs901585034 Chr.10: 119117689-119117689 on Build GRCh38
rs113906863 Chr.10: 119117834-119117834 on Build GRCh38
rs892892570 Chr.10: 119117898-119117898 on Build GRCh38
rs1012149007 Chr.10: 119117899-119117899 on Build GRCh38
rs758652618 Chr.10: 119117534-119117534 on Build GRCh38
rs1020501417 Chr.10: 119117915-119117915 on Build GRCh38
rs770444619 Chr.10: 119117680-119117680 on Build GRCh38
rs981438358 Chr.10: 119117764-119117764 on Build GRCh38
rs780638360 Chr.10: 119117763-119117763 on Build GRCh38
rs775768028 Chr.10: 119117688-119117688 on Build GRCh38
rs1000843138 Chr.10: 119117524-119117524 on Build GRCh38
rs919998766 Chr.10: 119117953-119117953 on Build GRCh38
rs767530687 Chr.10: 119117641-119117641 on Build GRCh38
rs750175342 Chr.10: 119117650-119117651 on Build GRCh38
rs920026240 Chr.10: 119117805-119117805 on Build GRCh38
rs751655777 Chr.10: 119117552-119117552 on Build GRCh38
rs763305998 Chr.10: 119117694-119117694 on Build GRCh38
rs994080955 Chr.10: 119117977-119117980 on Build GRCh38
rs940637664 Chr.10: 119117839-119117839 on Build GRCh38
rs752886933 Chr.10: 119117533-119117533 on Build GRCh38
rs747818257 Chr.10: 119117822-119117822 on Build GRCh38
rs371560694 Chr.10: 119117684-119117684 on Build GRCh38
rs1047007588 Chr.10: 119117988-119117988 on Build GRCh38
rs113664509 Chr.10: 119117637-119117637 on Build GRCh38
rs758334742 Chr.10: 119117661-119117661 on Build GRCh38
rs2901173 Chr.10: 119117736-119117736 on Build GRCh38
rs971014805 Chr.10: 119117948-119117948 on Build GRCh38
rs777784734 Chr.10: 119117544-119117544 on Build GRCh38
rs561521835 Chr.10: 119117952-119117952 on Build GRCh38
rs983866126 Chr.10: 119117704-119117704 on Build GRCh38
rs568000111 Chr.10: 119117601-119117601 on Build GRCh38
rs1041069433 Chr.10: 119117969-119117970 on Build GRCh38
rs991952757 Chr.10: 119117794-119117794 on Build GRCh38
rs139716316 Chr.10: 119117651-119117651 on Build GRCh38
rs777173813 Chr.10: 119117607-119117607 on Build GRCh38
rs71480780 Chr.10: 119117975-119117975 on Build GRCh38
rs148807437 Chr.10: 119117606-119117606 on Build GRCh38
rs111727288 Chr.10: 119117823-119117823 on Build GRCh38
rs942656955 Chr.10: 119117966-119117966 on Build GRCh38
rs146864091 Chr.10: 119117638-119117638 on Build GRCh38
rs572102435 Chr.10: 119117777-119117777 on Build GRCh38
rs73444357 Chr.10: 119117512-119117512 on Build GRCh38
rs565020669 Chr.10: 119117936-119117936 on Build GRCh38
rs780040162 Chr.10: 119117559-119117559 on Build GRCh38
rs992512454 Chr.10: 119117932-119117932 on Build GRCh38
rs754894622 Chr.10: 119117719-119117719 on Build GRCh38
rs1020918916 Chr.10: 119117741-119117741 on Build GRCh38

View Full Product Details