Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9420432-9420981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX12P
Entrez Gene ID
440081
Gene Name
DEAD/H-box helicase 12, pseudogene
Gene Aliases
CHLR2, DDX12
UniGene
Hs.447869
Gene Chromosome Location
Chr.12: 9417691-9448172 on Build GRCh38
Transcripts
NR_033399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6138228 c.2137C>T p.L713L Chr.12: 9420580-9420580 on Build GRCh38
1721195 c.2181C>T p.G727G Chr.12: 9420536-9420536 on Build GRCh38
432014 c.2090G>T p.G697V Chr.12: 9420627-9420627 on Build GRCh38
432013 c.2183G>A p.R728H Chr.12: 9420534-9420534 on Build GRCh38
944655 c.2098T>C p.S700P Chr.12: 9420619-9420619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771724363 Chr.12: 9420696-9420699 on Build GRCh38
rs907342512 Chr.12: 9420893-9420893 on Build GRCh38
rs780965092 Chr.12: 9420695-9420702 on Build GRCh38
rs34067753 Chr.12: 9420667-9420670 on Build GRCh38
rs760781126 Chr.12: 9420918-9420918 on Build GRCh38
rs941161011 Chr.12: 9420514-9420514 on Build GRCh38
rs750663581 Chr.12: 9420577-9420577 on Build GRCh38
rs770658821 Chr.12: 9420789-9420789 on Build GRCh38
rs760491211 Chr.12: 9420481-9420481 on Build GRCh38
rs755863220 Chr.12: 9420751-9420751 on Build GRCh38
rs955354855 Chr.12: 9420958-9420958 on Build GRCh38
rs934020015 Chr.12: 9420558-9420558 on Build GRCh38
rs4763950 Chr.12: 9420742-9420742 on Build GRCh38
rs749210718 Chr.12: 9420500-9420500 on Build GRCh38
rs751633507 Chr.12: 9420537-9420537 on Build GRCh38
rs4763341 Chr.12: 9420878-9420878 on Build GRCh38
rs1013018163 Chr.12: 9420629-9420629 on Build GRCh38
rs750539478 Chr.12: 9420883-9420883 on Build GRCh38
rs773164771 Chr.12: 9420839-9420847 on Build GRCh38
rs1034711938 Chr.12: 9420702-9420702 on Build GRCh38
rs201468985 Chr.12: 9420491-9420491 on Build GRCh38
rs764500288 Chr.12: 9420684-9420699 on Build GRCh38
rs886802344 Chr.12: 9420920-9420920 on Build GRCh38
rs796260596 Chr.12: 9420711-9420711 on Build GRCh38
rs780246954 Chr.12: 9420644-9420644 on Build GRCh38
rs1051108791 Chr.12: 9420580-9420580 on Build GRCh38
rs921066047 Chr.12: 9420782-9420782 on Build GRCh38
rs975351318 Chr.12: 9420532-9420532 on Build GRCh38
rs868340060 Chr.12: 9420892-9420892 on Build GRCh38
rs748090563 Chr.12: 9420840-9420840 on Build GRCh38
rs907455084 Chr.12: 9420648-9420648 on Build GRCh38
rs7358609 Chr.12: 9420628-9420628 on Build GRCh38
rs796591853 Chr.12: 9420713-9420713 on Build GRCh38
rs1003142060 Chr.12: 9420908-9420908 on Build GRCh38
rs1016727588 Chr.12: 9420926-9420926 on Build GRCh38
rs185399139 Chr.12: 9420901-9420901 on Build GRCh38
rs759361214 Chr.12: 9420515-9420517 on Build GRCh38
rs775835965 Chr.12: 9420928-9420928 on Build GRCh38
rs775463010 Chr.12: 9420848-9420849 on Build GRCh38
rs374156189 Chr.12: 9420659-9420659 on Build GRCh38
rs796487493 Chr.12: 9420712-9420712 on Build GRCh38
rs765710707 Chr.12: 9420667-9420674 on Build GRCh38
rs76089528 Chr.12: 9420472-9420472 on Build GRCh38
rs751744478 Chr.12: 9420709-9420710 on Build GRCh38
rs982583733 Chr.12: 9420739-9420739 on Build GRCh38
rs778487030 Chr.12: 9420811-9420835 on Build GRCh38
rs776773379 Chr.12: 9420568-9420568 on Build GRCh38
rs770656276 Chr.12: 9420484-9420484 on Build GRCh38
rs951092072 Chr.12: 9420730-9420730 on Build GRCh38
rs903143052 Chr.12: 9420581-9420581 on Build GRCh38
rs987097824 Chr.12: 9420807-9420807 on Build GRCh38
rs375388889 Chr.12: 9420778-9420778 on Build GRCh38
rs765085709 Chr.12: 9420712-9420713 on Build GRCh38
rs1054491906 Chr.12: 9420867-9420867 on Build GRCh38
rs6487908 Chr.12: 9420828-9420828 on Build GRCh38
rs754152268 Chr.12: 9420930-9420930 on Build GRCh38
rs1054440553 Chr.12: 9420624-9420624 on Build GRCh38
rs4763328 Chr.12: 9420699-9420699 on Build GRCh38
rs1023211509 Chr.12: 9420635-9420635 on Build GRCh38
rs975119720 Chr.12: 9420928-9420929 on Build GRCh38
rs934780131 Chr.12: 9420622-9420622 on Build GRCh38
rs4763936 Chr.12: 9420627-9420627 on Build GRCh38
rs774404990 Chr.12: 9420524-9420524 on Build GRCh38
rs375873947 Chr.12: 9420692-9420699 on Build GRCh38
rs369120887 Chr.12: 9420713-9420714 on Build GRCh38
rs982917879 Chr.12: 9420470-9420470 on Build GRCh38
rs764224720 Chr.12: 9420692-9420703 on Build GRCh38
rs367960137 Chr.12: 9420709-9420709 on Build GRCh38
rs893331481 Chr.12: 9420625-9420625 on Build GRCh38
rs933924652 Chr.12: 9420784-9420784 on Build GRCh38
rs7358608 Chr.12: 9420534-9420534 on Build GRCh38
rs4763977 Chr.12: 9420879-9420879 on Build GRCh38
rs746534507 Chr.12: 9420671-9420674 on Build GRCh38
rs761884248 Chr.12: 9420547-9420547 on Build GRCh38
rs769551105 Chr.12: 9420839-9420839 on Build GRCh38
rs770244334 Chr.12: 9420710-9420710 on Build GRCh38
rs749015078 Chr.12: 9420800-9420800 on Build GRCh38
rs765594379 Chr.12: 9420587-9420587 on Build GRCh38
rs962462181 Chr.12: 9420927-9420927 on Build GRCh38
rs796240779 Chr.12: 9420710-9420710 on Build GRCh38
rs1004284018 Chr.12: 9420925-9420925 on Build GRCh38
rs373943124 Chr.12: 9420695-9420695 on Build GRCh38
rs764528430 Chr.12: 9420935-9420935 on Build GRCh38
rs951166494 Chr.12: 9420462-9420462 on Build GRCh38
rs779544706 Chr.12: 9420688-9420699 on Build GRCh38
rs962575882 Chr.12: 9420746-9420746 on Build GRCh38
rs866960172 Chr.12: 9420961-9420961 on Build GRCh38

View Full Product Details