Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9425660-9426207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX12P
Entrez Gene ID
440081
Gene Name
DEAD/H-box helicase 12, pseudogene
Gene Aliases
CHLR2, DDX12
UniGene
Hs.447869
Gene Chromosome Location
Chr.12: 9417691-9448172 on Build GRCh38
Transcripts
NR_033399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
469097 c.1588T>G p.F530V Chr.12: 9425758-9425758 on Build GRCh38
6263833 c.1540C>T p.R514W Chr.12: 9425806-9425806 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745972924 Chr.12: 9425762-9425762 on Build GRCh38
rs1003317657 Chr.12: 9425831-9425831 on Build GRCh38
rs774941392 Chr.12: 9425903-9425903 on Build GRCh38
rs934709592 Chr.12: 9425938-9425938 on Build GRCh38
rs988308640 Chr.12: 9425872-9425872 on Build GRCh38
rs1034766251 Chr.12: 9425840-9425840 on Build GRCh38
rs886861310 Chr.12: 9426156-9426156 on Build GRCh38
rs757275417 Chr.12: 9425723-9425723 on Build GRCh38
rs866679948 Chr.12: 9425966-9425966 on Build GRCh38
rs1003943499 Chr.12: 9426172-9426172 on Build GRCh38
rs904898990 Chr.12: 9426143-9426143 on Build GRCh38
rs76934509 Chr.12: 9426023-9426023 on Build GRCh38
rs760931358 Chr.12: 9425782-9425782 on Build GRCh38
rs1017194541 Chr.12: 9426173-9426173 on Build GRCh38
rs765035286 Chr.12: 9426147-9426147 on Build GRCh38
rs893385707 Chr.12: 9425764-9425764 on Build GRCh38
rs962970120 Chr.12: 9425874-9425874 on Build GRCh38
rs878968003 Chr.12: 9425921-9425921 on Build GRCh38
rs1054747567 Chr.12: 9425760-9425760 on Build GRCh38
rs772222564 Chr.12: 9425737-9425737 on Build GRCh38
rs771120361 Chr.12: 9425806-9425806 on Build GRCh38
rs955354464 Chr.12: 9425906-9425906 on Build GRCh38
rs779922612 Chr.12: 9426178-9426178 on Build GRCh38
rs886975869 Chr.12: 9425846-9425846 on Build GRCh38
rs61914674 Chr.12: 9426100-9426100 on Build GRCh38
rs762330984 Chr.12: 9425925-9425925 on Build GRCh38
rs934093677 Chr.12: 9425711-9425711 on Build GRCh38
rs975329758 Chr.12: 9425892-9425892 on Build GRCh38
rs759988208 Chr.12: 9425882-9425882 on Build GRCh38
rs146796043 Chr.12: 9425917-9425917 on Build GRCh38
rs766049761 Chr.12: 9426032-9426032 on Build GRCh38
rs921253354 Chr.12: 9425682-9425682 on Build GRCh38
rs780826474 Chr.12: 9426093-9426093 on Build GRCh38
rs948999507 Chr.12: 9426061-9426061 on Build GRCh38
rs904830888 Chr.12: 9425829-9425829 on Build GRCh38
rs191531663 Chr.12: 9425679-9425679 on Build GRCh38
rs1004057605 Chr.12: 9425871-9425871 on Build GRCh38
rs1045147619 Chr.12: 9425805-9425805 on Build GRCh38
rs1056109358 Chr.12: 9426153-9426153 on Build GRCh38
rs10843759 Chr.12: 9425993-9425993 on Build GRCh38
rs924804200 Chr.12: 9425729-9425729 on Build GRCh38
rs554245775 Chr.12: 9425926-9425926 on Build GRCh38
rs148935434 Chr.12: 9425974-9425974 on Build GRCh38
rs990724946 Chr.12: 9425948-9425948 on Build GRCh38
rs1013128692 Chr.12: 9425781-9425781 on Build GRCh38
rs1051112606 Chr.12: 9425715-9425715 on Build GRCh38
rs749669747 Chr.12: 9425828-9425828 on Build GRCh38
rs879098536 Chr.12: 9425879-9425879 on Build GRCh38
rs1028773060 Chr.12: 9425893-9425893 on Build GRCh38

View Full Product Details