Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 18845222-18845795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A9P
Entrez Gene ID
284232
Gene Name
ankyrin repeat domain 20 family member A9, pseudogene
Gene Aliases
-
UniGene
Hs.679496
Gene Chromosome Location
Chr.13: 18834403-18871969 on Build GRCh38
Transcripts
NR_027995.1

SNP Detail

SNP ID Chromosome Location
rs372001711 Chr.13: 18845527-18845527 on Build GRCh38
rs530212276 Chr.13: 18845697-18845697 on Build GRCh38
rs559920770 Chr.13: 18845508-18845508 on Build GRCh38
rs901089880 Chr.13: 18845383-18845383 on Build GRCh38
rs71089005 Chr.13: 18845697-18845698 on Build GRCh38
rs75318892 Chr.13: 18845773-18845773 on Build GRCh38
rs531251326 Chr.13: 18845745-18845745 on Build GRCh38
rs960277394 Chr.13: 18845669-18845669 on Build GRCh38
rs199500066 Chr.13: 18845707-18845707 on Build GRCh38
rs931489409 Chr.13: 18845705-18845705 on Build GRCh38
rs548503589 Chr.13: 18845699-18845699 on Build GRCh38
rs879149732 Chr.13: 18845550-18845550 on Build GRCh38
rs369751253 Chr.13: 18845748-18845748 on Build GRCh38
rs577278872 Chr.13: 18845433-18845433 on Build GRCh38
rs879222867 Chr.13: 18845549-18845549 on Build GRCh38
rs398021718 Chr.13: 18845706-18845707 on Build GRCh38
rs928857514 Chr.13: 18845275-18845275 on Build GRCh38
rs111524581 Chr.13: 18845649-18845649 on Build GRCh38
rs182229460 Chr.13: 18845445-18845445 on Build GRCh38
rs755947648 Chr.13: 18845354-18845354 on Build GRCh38
rs867278717 Chr.13: 18845663-18845663 on Build GRCh38
rs368447108 Chr.13: 18845286-18845286 on Build GRCh38
rs549367330 Chr.13: 18845758-18845758 on Build GRCh38
rs74356655 Chr.13: 18845622-18845622 on Build GRCh38
rs1020697544 Chr.13: 18845451-18845451 on Build GRCh38
rs60594332 Chr.13: 18845321-18845321 on Build GRCh38
rs374744947 Chr.13: 18845399-18845399 on Build GRCh38
rs190051332 Chr.13: 18845325-18845325 on Build GRCh38
rs74349552 Chr.13: 18845707-18845707 on Build GRCh38
rs997170703 Chr.13: 18845405-18845405 on Build GRCh38
rs368107355 Chr.13: 18845521-18845521 on Build GRCh38
rs146090175 Chr.13: 18845300-18845300 on Build GRCh38
rs111461844 Chr.13: 18845704-18845704 on Build GRCh38
rs992157264 Chr.13: 18845701-18845702 on Build GRCh38
rs367865827 Chr.13: 18845473-18845473 on Build GRCh38
rs77964112 Chr.13: 18845772-18845772 on Build GRCh38
rs375031582 Chr.13: 18845633-18845633 on Build GRCh38
rs866944001 Chr.13: 18845463-18845463 on Build GRCh38
rs184634366 Chr.13: 18845367-18845367 on Build GRCh38
rs73435804 Chr.13: 18845668-18845668 on Build GRCh38
rs981796864 Chr.13: 18845272-18845272 on Build GRCh38
rs931377729 Chr.13: 18845313-18845313 on Build GRCh38
rs1004159504 Chr.13: 18845362-18845362 on Build GRCh38
rs927653190 Chr.13: 18845658-18845658 on Build GRCh38
rs201621686 Chr.13: 18845705-18845706 on Build GRCh38
rs571114658 Chr.13: 18845771-18845771 on Build GRCh38
rs781350837 Chr.13: 18845723-18845723 on Build GRCh38
rs908628220 Chr.13: 18845719-18845719 on Build GRCh38
rs371589349 Chr.13: 18845480-18845480 on Build GRCh38
rs373026548 Chr.13: 18845554-18845554 on Build GRCh38
rs1010637666 Chr.13: 18845449-18845449 on Build GRCh38
rs966384783 Chr.13: 18845244-18845244 on Build GRCh38
rs74692341 Chr.13: 18845643-18845643 on Build GRCh38
rs376265240 Chr.13: 18845763-18845763 on Build GRCh38
rs916050239 Chr.13: 18845703-18845703 on Build GRCh38
rs142880709 Chr.13: 18845279-18845279 on Build GRCh38
rs201384082 Chr.13: 18845579-18845579 on Build GRCh38
rs1007821767 Chr.13: 18845532-18845571 on Build GRCh38

View Full Product Details