Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 18849349-18849926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A9P
Entrez Gene ID
284232
Gene Name
ankyrin repeat domain 20 family member A9, pseudogene
Gene Aliases
-
UniGene
Hs.679496
Gene Chromosome Location
Chr.13: 18834403-18871969 on Build GRCh38
Transcripts
NR_027995.1

SNP Detail

SNP ID Chromosome Location
rs1033011532 Chr.13: 18849376-18849376 on Build GRCh38
rs918830827 Chr.13: 18849550-18849550 on Build GRCh38
rs893260249 Chr.13: 18849843-18849843 on Build GRCh38
rs561169548 Chr.13: 18849447-18849447 on Build GRCh38
rs550108079 Chr.13: 18849509-18849509 on Build GRCh38
rs1047959157 Chr.13: 18849519-18849519 on Build GRCh38
rs34123769 Chr.13: 18849719-18849719 on Build GRCh38
rs531962301 Chr.13: 18849434-18849434 on Build GRCh38
rs201184157 Chr.13: 18849659-18849659 on Build GRCh38
rs957376411 Chr.13: 18849610-18849610 on Build GRCh38
rs957240343 Chr.13: 18849403-18849404 on Build GRCh38
rs900546682 Chr.13: 18849590-18849590 on Build GRCh38
rs751703091 Chr.13: 18849530-18849530 on Build GRCh38
rs143698783 Chr.13: 18849451-18849451 on Build GRCh38
rs34315546 Chr.13: 18849684-18849684 on Build GRCh38
rs35149553 Chr.13: 18849768-18849768 on Build GRCh38
rs3963841 Chr.13: 18849796-18849796 on Build GRCh38
rs1020707673 Chr.13: 18849868-18849868 on Build GRCh38
rs759848791 Chr.13: 18849658-18849658 on Build GRCh38
rs35186508 Chr.13: 18849844-18849844 on Build GRCh38
rs544135938 Chr.13: 18849554-18849554 on Build GRCh38
rs927139400 Chr.13: 18849673-18849673 on Build GRCh38
rs1019568293 Chr.13: 18849551-18849551 on Build GRCh38
rs930875092 Chr.13: 18849513-18849513 on Build GRCh38
rs2997152 Chr.13: 18849560-18849560 on Build GRCh38
rs900684405 Chr.13: 18849835-18849835 on Build GRCh38
rs148259309 Chr.13: 18849660-18849660 on Build GRCh38
rs35513399 Chr.13: 18849764-18849765 on Build GRCh38
rs374809306 Chr.13: 18849424-18849424 on Build GRCh38
rs919702019 Chr.13: 18849766-18849766 on Build GRCh38
rs184874798 Chr.13: 18849695-18849695 on Build GRCh38
rs34068128 Chr.13: 18849902-18849903 on Build GRCh38
rs531925209 Chr.13: 18849897-18849897 on Build GRCh38
rs202106379 Chr.13: 18849584-18849584 on Build GRCh38
rs886718176 Chr.13: 18849524-18849524 on Build GRCh38
rs927043515 Chr.13: 18849481-18849481 on Build GRCh38
rs937065363 Chr.13: 18849689-18849689 on Build GRCh38
rs567885869 Chr.13: 18849845-18849845 on Build GRCh38
rs2343926 Chr.13: 18849841-18849841 on Build GRCh38
rs937094741 Chr.13: 18849492-18849492 on Build GRCh38
rs989249025 Chr.13: 18849613-18849613 on Build GRCh38
rs886689452 Chr.13: 18849805-18849805 on Build GRCh38
rs141254378 Chr.13: 18849784-18849784 on Build GRCh38
rs113371472 Chr.13: 18849630-18849630 on Build GRCh38
rs996270396 Chr.13: 18849596-18849596 on Build GRCh38
rs1040530550 Chr.13: 18849824-18849824 on Build GRCh38
rs913032026 Chr.13: 18849459-18849459 on Build GRCh38
rs573480539 Chr.13: 18849467-18849467 on Build GRCh38
rs34030388 Chr.13: 18849802-18849802 on Build GRCh38
rs981259979 Chr.13: 18849672-18849672 on Build GRCh38
rs981717279 Chr.13: 18849480-18849480 on Build GRCh38
rs898181984 Chr.13: 18849512-18849512 on Build GRCh38
rs944953898 Chr.13: 18849823-18849824 on Build GRCh38

View Full Product Details